Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 6560597-6561092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYAA
Entrez Gene ID
1409
Gene Name
crystallin alpha A
Gene Aliases
CRYA1, CTRCT9, HSPB4
UniGene
Hs.184085
Gene Chromosome Location
Chr.21: 43169008-43172810 on Build GRCh38
Transcripts
NM_000394.3 NM_001314050.2 XM_017028222.1 XM_017028224.1

Gene Symbol
LOC102724652
Entrez Gene ID
102724652
Gene Name
alpha-crystallin A chain
Gene Aliases
CRYAA
UniGene
Hs.184085
Gene Chromosome Location
Chr.21: 6560724-6564489 on Build GRCh38
Transcripts
NM_000394.3 NM_001314050.2 XM_017028222.1 XM_017028224.1

Gene Symbol
LOC102724843
Entrez Gene ID
102724843
Gene Name
uncharacterized LOC102724843
Gene Aliases
-
UniGene
Hs.743984
Gene Chromosome Location
Chr.21: 6318410-7087230 on Build GRCh38
Transcripts
NM_000394.3 NM_001314050.2 XM_017028222.1 XM_017028224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
226414 c.164C>T p.T55I Chr.21: 43169263-43169263 on Build GRCh38
5366177 c.76delT p.F27fs*51 Chr.21: 43169175-43169175 on Build GRCh38
1714058 c.172G>A p.D58N Chr.21: 43169271-43169271 on Build GRCh38
4630645 c.61C>T p.R21W Chr.21: 43169160-43169160 on Build GRCh38
478627 c.146G>A p.R49H Chr.21: 43169245-43169245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759943462 Chr.21: 43169138-43169138 on Build GRCh38
rs145489509 Chr.21: 43169098-43169098 on Build GRCh38
rs1026439128 Chr.21: 43168959-43168959 on Build GRCh38
rs771990490 Chr.21: 43169183-43169183 on Build GRCh38
rs397515626 Chr.21: 43169161-43169161 on Build GRCh38
rs776884812 Chr.21: 43169134-43169134 on Build GRCh38
rs151103202 Chr.21: 43169038-43169038 on Build GRCh38
rs765848159 Chr.21: 43169139-43169139 on Build GRCh38
rs768603130 Chr.21: 43169071-43169071 on Build GRCh38
rs755769949 Chr.21: 43169162-43169162 on Build GRCh38
rs779706903 Chr.21: 43169166-43169166 on Build GRCh38
rs769072627 Chr.21: 43169280-43169280 on Build GRCh38
rs926549818 Chr.21: 43169186-43169186 on Build GRCh38
rs544842669 Chr.21: 43169387-43169387 on Build GRCh38
rs74315441 Chr.21: 43169244-43169244 on Build GRCh38
rs867770612 Chr.21: 43169010-43169010 on Build GRCh38
rs749084824 Chr.21: 43169069-43169069 on Build GRCh38
rs752455701 Chr.21: 43169254-43169254 on Build GRCh38
rs528354296 Chr.21: 43169101-43169101 on Build GRCh38
rs756926049 Chr.21: 43169169-43169169 on Build GRCh38
rs777436397 Chr.21: 43169052-43169052 on Build GRCh38
rs564140269 Chr.21: 43169048-43169048 on Build GRCh38
rs148704068 Chr.21: 43169149-43169149 on Build GRCh38
rs764642890 Chr.21: 43169221-43169221 on Build GRCh38
rs973320061 Chr.21: 43169358-43169358 on Build GRCh38
rs397515623 Chr.21: 43169259-43169259 on Build GRCh38
rs759996565 Chr.21: 43169204-43169204 on Build GRCh38
rs773680017 Chr.21: 43169369-43169369 on Build GRCh38
rs993156477 Chr.21: 43168948-43168948 on Build GRCh38
rs74315440 Chr.21: 43169126-43169126 on Build GRCh38
rs529255065 Chr.21: 43168958-43168958 on Build GRCh38
rs370849818 Chr.21: 43169296-43169296 on Build GRCh38
rs781119593 Chr.21: 43169265-43169265 on Build GRCh38
rs746159831 Chr.21: 43169273-43169273 on Build GRCh38
rs937814934 Chr.21: 43169216-43169216 on Build GRCh38
rs377222721 Chr.21: 43169303-43169303 on Build GRCh38
rs747472068 Chr.21: 43169314-43169314 on Build GRCh38
rs374495173 Chr.21: 43169322-43169322 on Build GRCh38
rs61729442 Chr.21: 43169153-43169153 on Build GRCh38
rs151213687 Chr.21: 43169210-43169210 on Build GRCh38
rs73906469 Chr.21: 43169359-43169359 on Build GRCh38
rs754701447 Chr.21: 43169174-43169174 on Build GRCh38
rs397515625 Chr.21: 43169160-43169160 on Build GRCh38
rs569539136 Chr.21: 43169380-43169380 on Build GRCh38
rs777728814 Chr.21: 43169260-43169260 on Build GRCh38
rs749068174 Chr.21: 43169168-43169168 on Build GRCh38
rs778575205 Chr.21: 43169184-43169184 on Build GRCh38
rs760190388 Chr.21: 43169225-43169225 on Build GRCh38
rs758292459 Chr.21: 43169258-43169258 on Build GRCh38
rs188609121 Chr.21: 43169385-43169385 on Build GRCh38
rs550630746 Chr.21: 43169277-43169277 on Build GRCh38
rs867380092 Chr.21: 43169372-43169372 on Build GRCh38
rs56975193 Chr.21: 43169321-43169321 on Build GRCh38
rs747067279 Chr.21: 43169264-43169264 on Build GRCh38
rs776849025 Chr.21: 43169202-43169202 on Build GRCh38
rs866394406 Chr.21: 43169220-43169220 on Build GRCh38
rs397515624 Chr.21: 43169133-43169133 on Build GRCh38
rs766604319 Chr.21: 43169106-43169106 on Build GRCh38
rs779557208 Chr.21: 43169276-43169276 on Build GRCh38
rs183125538 Chr.21: 43169345-43169345 on Build GRCh38
rs547809161 Chr.21: 43169019-43169019 on Build GRCh38
rs765952577 Chr.21: 43169232-43169232 on Build GRCh38
rs954405425 Chr.21: 43168984-43168984 on Build GRCh38
rs370282382 Chr.21: 43169082-43169082 on Build GRCh38
rs763595704 Chr.21: 43169217-43169217 on Build GRCh38
rs749922517 Chr.21: 43169222-43169222 on Build GRCh38
rs771092064 Chr.21: 43169188-43169188 on Build GRCh38
rs760115864 Chr.21: 43169295-43169295 on Build GRCh38
rs551118846 Chr.21: 43169344-43169344 on Build GRCh38
rs774854230 Chr.21: 43169286-43169286 on Build GRCh38
rs373635187 Chr.21: 43169180-43169180 on Build GRCh38
rs996966230 Chr.21: 43169287-43169287 on Build GRCh38
rs779756218 Chr.21: 43169065-43169065 on Build GRCh38
rs368931928 Chr.21: 43169181-43169181 on Build GRCh38
rs868205458 Chr.21: 43169011-43169011 on Build GRCh38
rs191516889 Chr.21: 43169302-43169302 on Build GRCh38
rs754607706 Chr.21: 43169245-43169245 on Build GRCh38
rs775994219 Chr.21: 43169212-43169212 on Build GRCh38
rs201572637 Chr.21: 43169081-43169081 on Build GRCh38
rs530500347 Chr.21: 43169037-43169037 on Build GRCh38
rs752638416 Chr.21: 43169027-43169027 on Build GRCh38
rs770111576 Chr.21: 43169274-43169274 on Build GRCh38
rs536787336 Chr.21: 43169309-43169309 on Build GRCh38
rs146914780 Chr.21: 43169253-43169253 on Build GRCh38
rs745515700 Chr.21: 43169207-43169207 on Build GRCh38
rs1032431032 Chr.21: 43169288-43169288 on Build GRCh38
rs872331 Chr.21: 43169105-43169105 on Build GRCh38
rs751576823 Chr.21: 43169324-43169324 on Build GRCh38
rs864309685 Chr.21: 43169241-43169241 on Build GRCh38
rs75358372 Chr.21: 43169242-43169242 on Build GRCh38

View Full Product Details