Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43172533-43173026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYAA
Entrez Gene ID
1409
Gene Name
crystallin alpha A
Gene Aliases
CRYA1, CTRCT9, HSPB4
UniGene
Hs.184085
Gene Chromosome Location
Chr.21: 43169008-43172810 on Build GRCh38
Transcripts
NM_000394.3 XM_005261093.3 NM_001314050.2 NM_001320719.1 XM_017028222.1 XM_017028224.1

Gene Symbol
LOC102724652
Entrez Gene ID
102724652
Gene Name
alpha-crystallin A chain
Gene Aliases
CRYAA
UniGene
Hs.184085
Gene Chromosome Location
Chr.21: 6560724-6564489 on Build GRCh38
Transcripts
NM_000394.3 XM_005261093.3 NM_001314050.2 NM_001320719.1 XM_017028222.1 XM_017028224.1

Gene Symbol
LOC102724843
Entrez Gene ID
102724843
Gene Name
uncharacterized LOC102724843
Gene Aliases
-
UniGene
Hs.743984
Gene Chromosome Location
Chr.21: 6318410-7087230 on Build GRCh38
Transcripts
NM_000394.3 XM_005261093.3 NM_001314050.2 NM_001320719.1 XM_017028222.1 XM_017028224.1

SNP Detail

SNP ID Chromosome Location
rs942697582 Chr.21: 43172626-43172626 on Build GRCh38
rs898818657 Chr.21: 43172957-43172957 on Build GRCh38
rs538680993 Chr.21: 43172651-43172651 on Build GRCh38
rs1002219013 Chr.21: 43172574-43172574 on Build GRCh38
rs550642274 Chr.21: 43172835-43172835 on Build GRCh38
rs986869549 Chr.21: 43172784-43172784 on Build GRCh38
rs749514413 Chr.21: 43172842-43172842 on Build GRCh38
rs79614970 Chr.21: 43172787-43172787 on Build GRCh38
rs913762061 Chr.21: 43172856-43172856 on Build GRCh38
rs949177930 Chr.21: 43172892-43172892 on Build GRCh38
rs559509759 Chr.21: 43172812-43172812 on Build GRCh38
rs370015697 Chr.21: 43172660-43172660 on Build GRCh38
rs964403206 Chr.21: 43172789-43172789 on Build GRCh38
rs186003558 Chr.21: 43172719-43172719 on Build GRCh38
rs886057110 Chr.21: 43172678-43172678 on Build GRCh38
rs986838926 Chr.21: 43172615-43172615 on Build GRCh38
rs925193718 Chr.21: 43172642-43172642 on Build GRCh38
rs558009670 Chr.21: 43172815-43172815 on Build GRCh38
rs988047017 Chr.21: 43172845-43172845 on Build GRCh38
rs1046162095 Chr.21: 43172671-43172671 on Build GRCh38
rs372204836 Chr.21: 43172725-43172725 on Build GRCh38
rs1028797433 Chr.21: 43172627-43172627 on Build GRCh38
rs951534583 Chr.21: 43172774-43172774 on Build GRCh38
rs1034584781 Chr.21: 43172598-43172598 on Build GRCh38
rs112493206 Chr.21: 43172923-43172923 on Build GRCh38
rs886057108 Chr.21: 43172613-43172613 on Build GRCh38
rs562779423 Chr.21: 43172920-43172920 on Build GRCh38
rs577569114 Chr.21: 43172612-43172612 on Build GRCh38
rs555429491 Chr.21: 43172914-43172914 on Build GRCh38
rs543336377 Chr.21: 43172733-43172733 on Build GRCh38
rs963218071 Chr.21: 43172610-43172610 on Build GRCh38
rs951505233 Chr.21: 43172613-43172613 on Build GRCh38
rs563322743 Chr.21: 43172867-43172867 on Build GRCh38
rs904879500 Chr.21: 43172702-43172702 on Build GRCh38
rs1001849891 Chr.21: 43172750-43172750 on Build GRCh38
rs995801534 Chr.21: 43172965-43172966 on Build GRCh38
rs373483991 Chr.21: 43172929-43172929 on Build GRCh38
rs975752194 Chr.21: 43172631-43172631 on Build GRCh38
rs528273386 Chr.21: 43172994-43172994 on Build GRCh38
rs375948304 Chr.21: 43172974-43172974 on Build GRCh38
rs549300833 Chr.21: 43172935-43172935 on Build GRCh38
rs949145671 Chr.21: 43172663-43172663 on Build GRCh38
rs1037411492 Chr.21: 43172931-43172931 on Build GRCh38
rs1025801625 Chr.21: 43172773-43172773 on Build GRCh38
rs936506684 Chr.21: 43172656-43172656 on Build GRCh38
rs1035023592 Chr.21: 43172758-43172758 on Build GRCh38
rs544662221 Chr.21: 43172861-43172861 on Build GRCh38
rs886057109 Chr.21: 43172656-43172656 on Build GRCh38
rs573484482 Chr.21: 43172768-43172768 on Build GRCh38
rs13048089 Chr.21: 43172765-43172765 on Build GRCh38
rs879199143 Chr.21: 43172603-43172603 on Build GRCh38
rs530710796 Chr.21: 43172877-43172877 on Build GRCh38
rs926360077 Chr.21: 43172923-43172923 on Build GRCh38

View Full Product Details