Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43773581-43774074 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTB
Entrez Gene ID
1476
Gene Name
cystatin B
Gene Aliases
CPI-B, CST6, EPM1, EPM1A, PME, STFB, ULD
UniGene
Hs.695
Gene Chromosome Location
Chr.21: 43773665-43776375 on Build GRCh38
Transcripts
NM_000100.3

SNP Detail

SNP ID Chromosome Location
rs568581184 Chr.21: 43773959-43773959 on Build GRCh38
rs1015744457 Chr.21: 43773806-43773806 on Build GRCh38
rs201363469 Chr.21: 43773902-43773902 on Build GRCh38
rs202136987 Chr.21: 43773920-43773921 on Build GRCh38
rs894291820 Chr.21: 43773829-43773829 on Build GRCh38
rs951425686 Chr.21: 43773604-43773604 on Build GRCh38
rs548299534 Chr.21: 43773798-43773798 on Build GRCh38
rs1041474318 Chr.21: 43773952-43773952 on Build GRCh38
rs1055755692 Chr.21: 43773812-43773812 on Build GRCh38
rs536069557 Chr.21: 43773883-43773883 on Build GRCh38
rs1044429944 Chr.21: 43773760-43773760 on Build GRCh38
rs540274837 Chr.21: 43774006-43774006 on Build GRCh38
rs11553835 Chr.21: 43774010-43774010 on Build GRCh38
rs998254078 Chr.21: 43773848-43773848 on Build GRCh38
rs888243817 Chr.21: 43773788-43773788 on Build GRCh38
rs766872034 Chr.21: 43773661-43773661 on Build GRCh38
rs1014097979 Chr.21: 43773835-43773835 on Build GRCh38
rs528349054 Chr.21: 43773902-43773902 on Build GRCh38
rs547036879 Chr.21: 43773655-43773655 on Build GRCh38
rs754440032 Chr.21: 43773770-43773770 on Build GRCh38
rs935703178 Chr.21: 43773966-43773966 on Build GRCh38
rs888389444 Chr.21: 43773917-43773917 on Build GRCh38
rs1048300045 Chr.21: 43773783-43773783 on Build GRCh38
rs554739469 Chr.21: 43773996-43773996 on Build GRCh38
rs374963725 Chr.21: 43773725-43773725 on Build GRCh38
rs371799006 Chr.21: 43773656-43773656 on Build GRCh38
rs1016932344 Chr.21: 43773869-43773869 on Build GRCh38
rs149039598 Chr.21: 43773767-43773767 on Build GRCh38
rs571530840 Chr.21: 43773728-43773728 on Build GRCh38
rs900917625 Chr.21: 43774000-43774000 on Build GRCh38
rs572891070 Chr.21: 43773637-43773637 on Build GRCh38
rs917358088 Chr.21: 43773677-43773677 on Build GRCh38
rs577797482 Chr.21: 43773791-43773791 on Build GRCh38
rs950195472 Chr.21: 43773886-43773886 on Build GRCh38
rs1003876641 Chr.21: 43773609-43773609 on Build GRCh38
rs143062585 Chr.21: 43773847-43773847 on Build GRCh38
rs927256902 Chr.21: 43773901-43773901 on Build GRCh38
rs1015348453 Chr.21: 43773610-43773610 on Build GRCh38
rs556776865 Chr.21: 43773905-43773905 on Build GRCh38
rs978924042 Chr.21: 43773648-43773648 on Build GRCh38
rs533969154 Chr.21: 43773868-43773868 on Build GRCh38
rs532299150 Chr.21: 43773708-43773708 on Build GRCh38
rs576730070 Chr.21: 43774029-43774029 on Build GRCh38
rs572952209 Chr.21: 43774001-43774001 on Build GRCh38
rs951512967 Chr.21: 43773865-43773865 on Build GRCh38
rs1036902560 Chr.21: 43773989-43773989 on Build GRCh38
rs562549134 Chr.21: 43773773-43773773 on Build GRCh38
rs886057111 Chr.21: 43773901-43773901 on Build GRCh38
rs921411780 Chr.21: 43773671-43773671 on Build GRCh38
rs565205873 Chr.21: 43773657-43773657 on Build GRCh38
rs952460644 Chr.21: 43773603-43773603 on Build GRCh38
rs904228629 Chr.21: 43773799-43773799 on Build GRCh38
rs976471098 Chr.21: 43773624-43773624 on Build GRCh38
rs536022878 Chr.21: 43773969-43773969 on Build GRCh38
rs768173714 Chr.21: 43773915-43773915 on Build GRCh38
rs1004069458 Chr.21: 43773805-43773805 on Build GRCh38
rs950074811 Chr.21: 43773744-43773744 on Build GRCh38
rs925620723 Chr.21: 43773963-43773963 on Build GRCh38
rs3177026 Chr.21: 43773949-43773949 on Build GRCh38
rs939838421 Chr.21: 43773928-43773928 on Build GRCh38
rs1041379459 Chr.21: 43773775-43773775 on Build GRCh38
rs980505953 Chr.21: 43773889-43773889 on Build GRCh38
rs548101289 Chr.21: 43773708-43773709 on Build GRCh38
rs560936378 Chr.21: 43773615-43773615 on Build GRCh38
rs934212230 Chr.21: 43773706-43773706 on Build GRCh38
rs917307976 Chr.21: 43773884-43773884 on Build GRCh38
rs956168810 Chr.21: 43773880-43773880 on Build GRCh38
rs886057112 Chr.21: 43773975-43773975 on Build GRCh38
rs1033276231 Chr.21: 43773878-43773878 on Build GRCh38
rs945788757 Chr.21: 43773935-43773938 on Build GRCh38
rs955693007 Chr.21: 43773911-43773911 on Build GRCh38
rs954314276 Chr.21: 43773638-43773638 on Build GRCh38
rs28691645 Chr.21: 43773877-43773877 on Build GRCh38
rs558255323 Chr.21: 43774021-43774021 on Build GRCh38
rs1028031915 Chr.21: 43773857-43773857 on Build GRCh38

View Full Product Details