Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 43903868-43904367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT3
Entrez Gene ID
56894
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases
LPAAT-GAMMA1, LPAAT3
UniGene
Hs.248785
Gene Chromosome Location
Chr.21: 43865160-43987594 on Build GRCh38
Transcripts
NM_020132.4 XM_005261160.4 XM_006724031.3 XM_011529665.2 XM_017028409.1 XM_017028412.1 XM_017028413.1

SNP Detail

SNP ID Chromosome Location
rs987434658 Chr.21: 43904195-43904195 on Build GRCh38
rs894425357 Chr.21: 43904071-43904071 on Build GRCh38
rs773250374 Chr.21: 43904089-43904091 on Build GRCh38
rs1024552069 Chr.21: 43903971-43903971 on Build GRCh38
rs1038931156 Chr.21: 43903985-43903985 on Build GRCh38
rs767893516 Chr.21: 43904327-43904327 on Build GRCh38
rs989355210 Chr.21: 43904039-43904039 on Build GRCh38
rs796086630 Chr.21: 43904009-43904009 on Build GRCh38
rs945104245 Chr.21: 43903905-43903905 on Build GRCh38
rs1000080223 Chr.21: 43903898-43903898 on Build GRCh38
rs1027867239 Chr.21: 43904081-43904081 on Build GRCh38
rs370025346 Chr.21: 43904101-43904104 on Build GRCh38
rs980456448 Chr.21: 43904232-43904232 on Build GRCh38
rs973562421 Chr.21: 43904079-43904079 on Build GRCh38
rs113663742 Chr.21: 43903993-43904216 on Build GRCh38
rs930128610 Chr.21: 43904104-43904104 on Build GRCh38
rs1006840188 Chr.21: 43904014-43904014 on Build GRCh38
rs778682248 Chr.21: 43904273-43904273 on Build GRCh38
rs895399016 Chr.21: 43903969-43903969 on Build GRCh38
rs974739130 Chr.21: 43904321-43904321 on Build GRCh38
rs552515612 Chr.21: 43904314-43904314 on Build GRCh38
rs1011257255 Chr.21: 43904179-43904179 on Build GRCh38
rs1018216480 Chr.21: 43904127-43904127 on Build GRCh38
rs1051158318 Chr.21: 43904121-43904121 on Build GRCh38
rs966545863 Chr.21: 43904052-43904052 on Build GRCh38
rs938958652 Chr.21: 43903926-43903926 on Build GRCh38
rs2838427 Chr.21: 43904328-43904328 on Build GRCh38
rs34010891 Chr.21: 43904216-43904217 on Build GRCh38
rs1051299615 Chr.21: 43904039-43904040 on Build GRCh38
rs188033697 Chr.21: 43903906-43903906 on Build GRCh38
rs572362395 Chr.21: 43904045-43904045 on Build GRCh38
rs545997950 Chr.21: 43904333-43904333 on Build GRCh38
rs528827989 Chr.21: 43903991-43903991 on Build GRCh38
rs60807016 Chr.21: 43904100-43904103 on Build GRCh38
rs891209864 Chr.21: 43904125-43904125 on Build GRCh38
rs148975942 Chr.21: 43904126-43904126 on Build GRCh38
rs977911005 Chr.21: 43904212-43904212 on Build GRCh38
rs945511955 Chr.21: 43904244-43904244 on Build GRCh38
rs541958704 Chr.21: 43903919-43903919 on Build GRCh38
rs193270153 Chr.21: 43904148-43904148 on Build GRCh38
rs990467570 Chr.21: 43904075-43904075 on Build GRCh38
rs951473359 Chr.21: 43903972-43903972 on Build GRCh38
rs897320581 Chr.21: 43904102-43904102 on Build GRCh38
rs934235987 Chr.21: 43904024-43904024 on Build GRCh38
rs966953485 Chr.21: 43904202-43904202 on Build GRCh38
rs549204663 Chr.21: 43904266-43904266 on Build GRCh38
rs774563756 Chr.21: 43904270-43904270 on Build GRCh38
rs759858468 Chr.21: 43904274-43904274 on Build GRCh38
rs955671752 Chr.21: 43904168-43904168 on Build GRCh38
rs925744006 Chr.21: 43904269-43904269 on Build GRCh38
rs1017511991 Chr.21: 43904016-43904016 on Build GRCh38
rs1016998941 Chr.21: 43904131-43904131 on Build GRCh38
rs930177416 Chr.21: 43904345-43904345 on Build GRCh38
rs71322542 Chr.21: 43904064-43904064 on Build GRCh38
rs945816456 Chr.21: 43904055-43904055 on Build GRCh38
rs60694942 Chr.21: 43904088-43904103 on Build GRCh38
rs1055391799 Chr.21: 43903929-43903929 on Build GRCh38
rs569821174 Chr.21: 43904162-43904162 on Build GRCh38
rs77949124 Chr.21: 43903921-43903921 on Build GRCh38
rs975480546 Chr.21: 43904158-43904158 on Build GRCh38
rs982878817 Chr.21: 43904129-43904129 on Build GRCh38
rs964579057 Chr.21: 43904038-43904038 on Build GRCh38
rs904204309 Chr.21: 43904065-43904066 on Build GRCh38
rs897293901 Chr.21: 43903988-43903988 on Build GRCh38
rs1014575174 Chr.21: 43904090-43904090 on Build GRCh38
rs1022172477 Chr.21: 43904185-43904185 on Build GRCh38
rs904171483 Chr.21: 43903891-43903891 on Build GRCh38
rs372453870 Chr.21: 43904065-43904065 on Build GRCh38
rs146514344 Chr.21: 43904063-43904063 on Build GRCh38
rs1041450361 Chr.21: 43904264-43904264 on Build GRCh38
rs754733054 Chr.21: 43904089-43904104 on Build GRCh38
rs762002430 Chr.21: 43904068-43904069 on Build GRCh38
rs543262406 Chr.21: 43903922-43903922 on Build GRCh38
rs534947905 Chr.21: 43904285-43904285 on Build GRCh38
rs540241479 Chr.21: 43904011-43904011 on Build GRCh38
rs970052680 Chr.21: 43904088-43904088 on Build GRCh38
rs914082959 Chr.21: 43904215-43904215 on Build GRCh38
rs546103079 Chr.21: 43904315-43904315 on Build GRCh38
rs10577584 Chr.21: 43904066-43904067 on Build GRCh38
rs1034236203 Chr.21: 43903902-43903902 on Build GRCh38
rs530627540 Chr.21: 43904248-43904248 on Build GRCh38
rs529253502 Chr.21: 43904068-43904071 on Build GRCh38
rs994681299 Chr.21: 43904005-43904005 on Build GRCh38
rs139153986 Chr.21: 43904067-43904067 on Build GRCh38
rs565312184 Chr.21: 43904056-43904056 on Build GRCh38
rs567448083 Chr.21: 43904267-43904267 on Build GRCh38
rs921257915 Chr.21: 43904166-43904166 on Build GRCh38
rs145431293 Chr.21: 43904066-43904069 on Build GRCh38
rs8134347 Chr.21: 43903982-43903982 on Build GRCh38

View Full Product Details