Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44966907-44967398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM207A
Entrez Gene ID
85395
Gene Name
family with sequence similarity 207 member A
Gene Aliases
C21orf70, PRED56
UniGene
Hs.410830
Gene Chromosome Location
Chr.21: 44939697-44976989 on Build GRCh38
Transcripts
NM_001316983.1 NM_001316984.1 NM_001316985.1 NM_001316986.1 NM_001316987.1 NM_001316988.1 NM_058190.3 XM_017028481.1 XM_017028482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6411161 c.470T>A p.L157H Chr.21: 44967151-44967151 on Build GRCh38
1414467 c.494C>T p.A165V Chr.21: 44967175-44967175 on Build GRCh38
6659315 c.414G>A p.T138T Chr.21: 44967095-44967095 on Build GRCh38
4650564 c.488G>A p.R163H Chr.21: 44967169-44967169 on Build GRCh38
231901 c.496C>T p.R166C Chr.21: 44967177-44967177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964279991 Chr.21: 44967310-44967310 on Build GRCh38
rs773993904 Chr.21: 44967029-44967029 on Build GRCh38
rs371433764 Chr.21: 44967347-44967347 on Build GRCh38
rs562268598 Chr.21: 44967245-44967245 on Build GRCh38
rs778908774 Chr.21: 44967104-44967104 on Build GRCh38
rs555958536 Chr.21: 44967166-44967166 on Build GRCh38
rs77534538 Chr.21: 44967191-44967191 on Build GRCh38
rs768374665 Chr.21: 44967131-44967131 on Build GRCh38
rs954469085 Chr.21: 44967271-44967271 on Build GRCh38
rs747006698 Chr.21: 44967095-44967097 on Build GRCh38
rs1039996453 Chr.21: 44967162-44967162 on Build GRCh38
rs544052465 Chr.21: 44966938-44966938 on Build GRCh38
rs760038943 Chr.21: 44967247-44967247 on Build GRCh38
rs747585775 Chr.21: 44967186-44967186 on Build GRCh38
rs1020482771 Chr.21: 44967329-44967329 on Build GRCh38
rs767297798 Chr.21: 44967148-44967148 on Build GRCh38
rs1020416235 Chr.21: 44966997-44966997 on Build GRCh38
rs372628638 Chr.21: 44967079-44967079 on Build GRCh38
rs544087394 Chr.21: 44967242-44967242 on Build GRCh38
rs773402587 Chr.21: 44967038-44967038 on Build GRCh38
rs570001973 Chr.21: 44967354-44967354 on Build GRCh38
rs747319505 Chr.21: 44967118-44967118 on Build GRCh38
rs942949406 Chr.21: 44967157-44967157 on Build GRCh38
rs753648214 Chr.21: 44967221-44967221 on Build GRCh38
rs778351213 Chr.21: 44967114-44967114 on Build GRCh38
rs762440852 Chr.21: 44967050-44967050 on Build GRCh38
rs541577441 Chr.21: 44967265-44967265 on Build GRCh38
rs562230292 Chr.21: 44967015-44967015 on Build GRCh38
rs929784996 Chr.21: 44967372-44967372 on Build GRCh38
rs201080250 Chr.21: 44967028-44967028 on Build GRCh38
rs551508990 Chr.21: 44967332-44967332 on Build GRCh38
rs371356503 Chr.21: 44967022-44967022 on Build GRCh38
rs559575385 Chr.21: 44967288-44967288 on Build GRCh38
rs766442397 Chr.21: 44967218-44967218 on Build GRCh38
rs899094227 Chr.21: 44967368-44967368 on Build GRCh38
rs771346942 Chr.21: 44967126-44967126 on Build GRCh38
rs373517384 Chr.21: 44967355-44967355 on Build GRCh38
rs1041756400 Chr.21: 44967341-44967341 on Build GRCh38
rs781213044 Chr.21: 44967096-44967096 on Build GRCh38
rs750174593 Chr.21: 44967085-44967085 on Build GRCh38
rs780426500 Chr.21: 44967014-44967014 on Build GRCh38
rs2294168 Chr.21: 44967163-44967163 on Build GRCh38
rs61746213 Chr.21: 44967178-44967178 on Build GRCh38
rs753025285 Chr.21: 44967228-44967228 on Build GRCh38
rs375490605 Chr.21: 44967229-44967229 on Build GRCh38
rs763835312 Chr.21: 44967381-44967381 on Build GRCh38
rs866883521 Chr.21: 44966939-44966939 on Build GRCh38
rs748284255 Chr.21: 44967107-44967107 on Build GRCh38
rs529505700 Chr.21: 44967256-44967256 on Build GRCh38
rs1025907688 Chr.21: 44967263-44967263 on Build GRCh38
rs368642060 Chr.21: 44967209-44967209 on Build GRCh38
rs766317444 Chr.21: 44967165-44967165 on Build GRCh38
rs140621159 Chr.21: 44967363-44967363 on Build GRCh38
rs570636434 Chr.21: 44967153-44967153 on Build GRCh38
rs376710872 Chr.21: 44966959-44966959 on Build GRCh38
rs758520408 Chr.21: 44967180-44967180 on Build GRCh38
rs910292401 Chr.21: 44967295-44967295 on Build GRCh38
rs779228829 Chr.21: 44967177-44967177 on Build GRCh38
rs954989817 Chr.21: 44967149-44967149 on Build GRCh38
rs775140006 Chr.21: 44967045-44967045 on Build GRCh38
rs912696848 Chr.21: 44967152-44967152 on Build GRCh38
rs138283625 Chr.21: 44967094-44967094 on Build GRCh38
rs755107672 Chr.21: 44967169-44967169 on Build GRCh38
rs772419973 Chr.21: 44967111-44967111 on Build GRCh38
rs942000719 Chr.21: 44967234-44967234 on Build GRCh38
rs781482296 Chr.21: 44967189-44967189 on Build GRCh38
rs374516190 Chr.21: 44967010-44967010 on Build GRCh38
rs894537112 Chr.21: 44967358-44967358 on Build GRCh38
rs199645945 Chr.21: 44967135-44967135 on Build GRCh38
rs368719130 Chr.21: 44966990-44966990 on Build GRCh38
rs201400373 Chr.21: 44967211-44967211 on Build GRCh38
rs1011990091 Chr.21: 44966985-44966985 on Build GRCh38
rs767518048 Chr.21: 44967084-44967084 on Build GRCh38
rs376860499 Chr.21: 44967018-44967018 on Build GRCh38
rs991380126 Chr.21: 44967060-44967060 on Build GRCh38
rs778242001 Chr.21: 44967185-44967185 on Build GRCh38
rs751488287 Chr.21: 44967074-44967074 on Build GRCh38
rs947598057 Chr.21: 44967339-44967339 on Build GRCh38
rs367751057 Chr.21: 44967137-44967137 on Build GRCh38
rs140960368 Chr.21: 44967078-44967078 on Build GRCh38
rs914854517 Chr.21: 44967336-44967336 on Build GRCh38
rs774732123 Chr.21: 44967227-44967227 on Build GRCh38
rs755077892 Chr.21: 44967103-44967103 on Build GRCh38
rs968338542 Chr.21: 44967168-44967168 on Build GRCh38
rs761214738 Chr.21: 44967204-44967204 on Build GRCh38
rs764807371 Chr.21: 44967039-44967039 on Build GRCh38
rs755983842 Chr.21: 44967095-44967095 on Build GRCh38
rs775123537 Chr.21: 44967127-44967127 on Build GRCh38
rs567876115 Chr.21: 44967187-44967187 on Build GRCh38
rs533394907 Chr.21: 44967323-44967323 on Build GRCh38
rs762639735 Chr.21: 44967130-44967130 on Build GRCh38
rs369863667 Chr.21: 44967059-44967059 on Build GRCh38
rs780894832 Chr.21: 44966989-44966989 on Build GRCh38
rs773252546 Chr.21: 44967210-44967210 on Build GRCh38
rs1047736134 Chr.21: 44967093-44967093 on Build GRCh38
rs373928500 Chr.21: 44967132-44967132 on Build GRCh38
rs142454716 Chr.21: 44967192-44967192 on Build GRCh38
rs151316068 Chr.21: 44967193-44967193 on Build GRCh38
rs912867666 Chr.21: 44966975-44966975 on Build GRCh38
rs1037914579 Chr.21: 44967364-44967364 on Build GRCh38
rs113562569 Chr.21: 44967237-44967237 on Build GRCh38
rs754179261 Chr.21: 44967098-44967098 on Build GRCh38
rs746729409 Chr.21: 44967011-44967011 on Build GRCh38
rs2294167 Chr.21: 44967370-44967370 on Build GRCh38

View Full Product Details