Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23764552-23765061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf15
Entrez Gene ID
150248
Gene Name
chromosome 22 open reading frame 15
Gene Aliases
N27C7-3
UniGene
Hs.116254
Gene Chromosome Location
Chr.22: 23762522-23765863 on Build GRCh38
Transcripts
NM_182520.2 XM_011529907.2 XM_011529908.2 XM_011529912.2 XM_017028602.1 XM_017028603.1 XM_017028604.1 XM_017028605.1 XM_017028606.1 XM_017028607.1 XM_017028608.1 XM_017028609.1 XM_017028610.1 XM_017028611.1 XM_017028612.1 XM_017028613.1 XM_017028614.1 XM_017028615.1 XM_017028616.1 XM_017028617.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5522242 c.327G>A p.E109E Chr.22: 23764794-23764794 on Build GRCh38
5527710 c.251-1G>A p.? Chr.22: 23764638-23764638 on Build GRCh38
1327306 c.48C>T p.G16G Chr.22: 23764581-23764581 on Build GRCh38
1415152 c.326G>A p.R109Q Chr.22: 23764859-23764859 on Build GRCh38
159575 c.89C>T p.S30F Chr.22: 23764622-23764622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563890232 Chr.22: 23765000-23765000 on Build GRCh38
rs543669757 Chr.22: 23764685-23764685 on Build GRCh38
rs757246676 Chr.22: 23764844-23764844 on Build GRCh38
rs758186435 Chr.22: 23764713-23764713 on Build GRCh38
rs566868267 Chr.22: 23764640-23764640 on Build GRCh38
rs781298027 Chr.22: 23764613-23764615 on Build GRCh38
rs904590550 Chr.22: 23764848-23764848 on Build GRCh38
rs767404030 Chr.22: 23764937-23764937 on Build GRCh38
rs769306922 Chr.22: 23764895-23764895 on Build GRCh38
rs775155066 Chr.22: 23764813-23764813 on Build GRCh38
rs867510292 Chr.22: 23764829-23764829 on Build GRCh38
rs751455317 Chr.22: 23764726-23764726 on Build GRCh38
rs569317011 Chr.22: 23764587-23764587 on Build GRCh38
rs554034284 Chr.22: 23764683-23764683 on Build GRCh38
rs746415942 Chr.22: 23764641-23764642 on Build GRCh38
rs374945245 Chr.22: 23764858-23764858 on Build GRCh38
rs145039987 Chr.22: 23764619-23764619 on Build GRCh38
rs752475063 Chr.22: 23764710-23764710 on Build GRCh38
rs148288019 Chr.22: 23764577-23764577 on Build GRCh38
rs752522586 Chr.22: 23764630-23764630 on Build GRCh38
rs780338916 Chr.22: 23764763-23764778 on Build GRCh38
rs946576566 Chr.22: 23764801-23764801 on Build GRCh38
rs768691988 Chr.22: 23764827-23764828 on Build GRCh38
rs771550546 Chr.22: 23764792-23764792 on Build GRCh38
rs370216786 Chr.22: 23764868-23764868 on Build GRCh38
rs1027837257 Chr.22: 23764949-23764949 on Build GRCh38
rs756188575 Chr.22: 23764732-23764732 on Build GRCh38
rs756496280 Chr.22: 23764732-23764732 on Build GRCh38
rs376388637 Chr.22: 23764662-23764662 on Build GRCh38
rs779027015 Chr.22: 23764859-23764859 on Build GRCh38
rs374645764 Chr.22: 23764825-23764825 on Build GRCh38
rs748172568 Chr.22: 23764985-23764986 on Build GRCh38
rs541460829 Chr.22: 23765028-23765028 on Build GRCh38
rs772517921 Chr.22: 23764766-23764766 on Build GRCh38
rs145399522 Chr.22: 23764774-23764774 on Build GRCh38
rs756851140 Chr.22: 23764609-23764622 on Build GRCh38
rs145628774 Chr.22: 23764672-23764672 on Build GRCh38
rs140216229 Chr.22: 23764663-23764663 on Build GRCh38
rs546921891 Chr.22: 23764624-23764624 on Build GRCh38
rs774640235 Chr.22: 23764665-23764665 on Build GRCh38
rs773833552 Chr.22: 23764682-23764682 on Build GRCh38
rs1022550886 Chr.22: 23764875-23764875 on Build GRCh38
rs767441367 Chr.22: 23764846-23764846 on Build GRCh38
rs545965061 Chr.22: 23764822-23764822 on Build GRCh38
rs554903635 Chr.22: 23764653-23764653 on Build GRCh38
rs1055084864 Chr.22: 23764784-23764784 on Build GRCh38
rs149199198 Chr.22: 23764776-23764776 on Build GRCh38
rs749377402 Chr.22: 23764743-23764743 on Build GRCh38
rs774200237 Chr.22: 23764922-23764922 on Build GRCh38
rs376590073 Chr.22: 23764950-23764950 on Build GRCh38
rs774585856 Chr.22: 23764918-23764918 on Build GRCh38
rs763544705 Chr.22: 23764860-23764860 on Build GRCh38
rs759569791 Chr.22: 23764980-23764980 on Build GRCh38
rs776435213 Chr.22: 23764690-23764690 on Build GRCh38
rs140565519 Chr.22: 23764861-23764861 on Build GRCh38
rs964853683 Chr.22: 23764901-23764901 on Build GRCh38
rs775008335 Chr.22: 23764902-23764902 on Build GRCh38
rs759166916 Chr.22: 23764694-23764694 on Build GRCh38
rs764038416 Chr.22: 23764725-23764725 on Build GRCh38
rs747455082 Chr.22: 23764779-23764779 on Build GRCh38
rs368585570 Chr.22: 23764983-23764983 on Build GRCh38
rs765297949 Chr.22: 23764982-23764982 on Build GRCh38
rs747372262 Chr.22: 23764867-23764867 on Build GRCh38
rs141469034 Chr.22: 23764847-23764847 on Build GRCh38
rs758333362 Chr.22: 23764633-23764633 on Build GRCh38
rs762764707 Chr.22: 23764815-23764815 on Build GRCh38
rs748510417 Chr.22: 23764761-23764761 on Build GRCh38
rs964907974 Chr.22: 23764613-23764613 on Build GRCh38
rs779169693 Chr.22: 23764755-23764755 on Build GRCh38
rs147550502 Chr.22: 23764644-23764644 on Build GRCh38
rs745392134 Chr.22: 23764893-23764893 on Build GRCh38
rs371418996 Chr.22: 23764749-23764749 on Build GRCh38
rs927658711 Chr.22: 23764762-23764762 on Build GRCh38
rs764847300 Chr.22: 23764805-23764805 on Build GRCh38
rs764937850 Chr.22: 23764697-23764697 on Build GRCh38
rs1009156697 Chr.22: 23764585-23764585 on Build GRCh38
rs761565402 Chr.22: 23764935-23764935 on Build GRCh38
rs534046925 Chr.22: 23764676-23764676 on Build GRCh38
rs200704446 Chr.22: 23764802-23764802 on Build GRCh38
rs969215832 Chr.22: 23764911-23764911 on Build GRCh38
rs376881173 Chr.22: 23764862-23764862 on Build GRCh38
rs760231662 Chr.22: 23764600-23764600 on Build GRCh38
rs776068105 Chr.22: 23764736-23764736 on Build GRCh38
rs749675044 Chr.22: 23764820-23764820 on Build GRCh38
rs780053397 Chr.22: 23764738-23764738 on Build GRCh38
rs754835786 Chr.22: 23764620-23764620 on Build GRCh38

View Full Product Details