Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23767282-23767777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHCHD10
Entrez Gene ID
400916
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 10
Gene Aliases
C22orf16, FTDALS2, IMMD, N27C7-4, SMAJ
UniGene
Hs.66915
Gene Chromosome Location
Chr.22: 23765834-23767972 on Build GRCh38
Transcripts
NM_001301339.1 NM_213720.2 NR_125755.1 NR_125756.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4637059 c.235G>A p.E79K Chr.22: 23767400-23767400 on Build GRCh38
5960240 c.239C>T p.P80L Chr.22: 23767396-23767396 on Build GRCh38
340606 c.201C>T p.S67S Chr.22: 23767434-23767434 on Build GRCh38
4790457 c.234G>A p.S78S Chr.22: 23767401-23767401 on Build GRCh38
6627770 c.193A>G p.M65V Chr.22: 23767442-23767442 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111527940 Chr.22: 23767401-23767401 on Build GRCh38
rs13054979 Chr.22: 23767466-23767466 on Build GRCh38
rs747111150 Chr.22: 23767361-23767361 on Build GRCh38
rs958389013 Chr.22: 23767599-23767599 on Build GRCh38
rs756331072 Chr.22: 23767461-23767461 on Build GRCh38
rs917055742 Chr.22: 23767318-23767318 on Build GRCh38
rs369956455 Chr.22: 23767540-23767540 on Build GRCh38
rs770043821 Chr.22: 23767391-23767391 on Build GRCh38
rs730880032 Chr.22: 23767592-23767592 on Build GRCh38
rs551286243 Chr.22: 23767502-23767502 on Build GRCh38
rs761442567 Chr.22: 23767407-23767407 on Build GRCh38
rs1023954590 Chr.22: 23767614-23767614 on Build GRCh38
rs775332895 Chr.22: 23767396-23767396 on Build GRCh38
rs754027063 Chr.22: 23767597-23767597 on Build GRCh38
rs778882470 Chr.22: 23767360-23767360 on Build GRCh38
rs767069606 Chr.22: 23767411-23767411 on Build GRCh38
rs377627832 Chr.22: 23767320-23767320 on Build GRCh38
rs753658886 Chr.22: 23767343-23767343 on Build GRCh38
rs918789560 Chr.22: 23767651-23767651 on Build GRCh38
rs551521196 Chr.22: 23767535-23767535 on Build GRCh38
rs889991693 Chr.22: 23767701-23767701 on Build GRCh38
rs780298646 Chr.22: 23767464-23767464 on Build GRCh38
rs951675547 Chr.22: 23767570-23767570 on Build GRCh38
rs1020545985 Chr.22: 23767548-23767548 on Build GRCh38
rs567239313 Chr.22: 23767600-23767600 on Build GRCh38
rs984639260 Chr.22: 23767678-23767678 on Build GRCh38
rs748608906 Chr.22: 23767476-23767476 on Build GRCh38
rs730880031 Chr.22: 23767438-23767438 on Build GRCh38
rs982377475 Chr.22: 23767632-23767632 on Build GRCh38
rs1039679741 Chr.22: 23767705-23767705 on Build GRCh38
rs748522940 Chr.22: 23767618-23767618 on Build GRCh38
rs564997204 Chr.22: 23767522-23767522 on Build GRCh38
rs558021937 Chr.22: 23767718-23767718 on Build GRCh38
rs113097524 Chr.22: 23767711-23767711 on Build GRCh38
rs776304054 Chr.22: 23767503-23767503 on Build GRCh38
rs756168961 Chr.22: 23767563-23767563 on Build GRCh38
rs754940184 Chr.22: 23767357-23767357 on Build GRCh38
rs781304084 Chr.22: 23767445-23767445 on Build GRCh38
rs1042797673 Chr.22: 23767663-23767663 on Build GRCh38
rs972398258 Chr.22: 23767730-23767730 on Build GRCh38
rs965210367 Chr.22: 23767331-23767331 on Build GRCh38
rs13053635 Chr.22: 23767467-23767467 on Build GRCh38
rs998438660 Chr.22: 23767670-23767670 on Build GRCh38
rs1000021202 Chr.22: 23767721-23767721 on Build GRCh38
rs1044227714 Chr.22: 23767323-23767323 on Build GRCh38
rs578061070 Chr.22: 23767737-23767737 on Build GRCh38
rs549731350 Chr.22: 23767690-23767690 on Build GRCh38
rs950020917 Chr.22: 23767568-23767568 on Build GRCh38
rs997985425 Chr.22: 23767333-23767333 on Build GRCh38
rs749635935 Chr.22: 23767473-23767473 on Build GRCh38
rs762943183 Chr.22: 23767532-23767532 on Build GRCh38
rs131444 Chr.22: 23767363-23767363 on Build GRCh38
rs772696163 Chr.22: 23767479-23767479 on Build GRCh38
rs752472094 Chr.22: 23767526-23767526 on Build GRCh38
rs888466594 Chr.22: 23767489-23767489 on Build GRCh38
rs200831077 Chr.22: 23767499-23767499 on Build GRCh38
rs587777574 Chr.22: 23767459-23767459 on Build GRCh38
rs566366853 Chr.22: 23767697-23767697 on Build GRCh38
rs750549532 Chr.22: 23767539-23767539 on Build GRCh38
rs938100726 Chr.22: 23767341-23767341 on Build GRCh38
rs1006801085 Chr.22: 23767314-23767314 on Build GRCh38
rs1024398251 Chr.22: 23767743-23767743 on Build GRCh38
rs179468 Chr.22: 23767587-23767587 on Build GRCh38
rs772927642 Chr.22: 23767482-23767482 on Build GRCh38
rs868345557 Chr.22: 23767598-23767598 on Build GRCh38
rs779478955 Chr.22: 23767603-23767603 on Build GRCh38
rs746028073 Chr.22: 23767365-23767365 on Build GRCh38
rs917149626 Chr.22: 23767565-23767565 on Build GRCh38
rs542541060 Chr.22: 23767421-23767421 on Build GRCh38
rs536230019 Chr.22: 23767683-23767683 on Build GRCh38
rs760464861 Chr.22: 23767419-23767419 on Build GRCh38
rs770541978 Chr.22: 23767501-23767501 on Build GRCh38
rs781141000 Chr.22: 23767716-23767716 on Build GRCh38
rs202148090 Chr.22: 23767446-23767446 on Build GRCh38
rs958999990 Chr.22: 23767712-23767712 on Build GRCh38
rs1028506132 Chr.22: 23767412-23767412 on Build GRCh38
rs971285560 Chr.22: 23767745-23767745 on Build GRCh38
rs903041403 Chr.22: 23767720-23767720 on Build GRCh38
rs80196988 Chr.22: 23767453-23767453 on Build GRCh38
rs1020091431 Chr.22: 23767340-23767340 on Build GRCh38
rs766054125 Chr.22: 23767420-23767420 on Build GRCh38
rs970975150 Chr.22: 23767619-23767619 on Build GRCh38
rs765115592 Chr.22: 23767429-23767429 on Build GRCh38
rs967231769 Chr.22: 23767559-23767559 on Build GRCh38
rs927168789 Chr.22: 23767589-23767589 on Build GRCh38
rs766034799 Chr.22: 23767335-23767335 on Build GRCh38
rs901115236 Chr.22: 23767531-23767531 on Build GRCh38
rs370872556 Chr.22: 23767406-23767406 on Build GRCh38
rs933945111 Chr.22: 23767713-23767713 on Build GRCh38
rs730880030 Chr.22: 23767591-23767591 on Build GRCh38
rs1053828210 Chr.22: 23767638-23767638 on Build GRCh38
rs930141749 Chr.22: 23767659-23767659 on Build GRCh38
rs775004786 Chr.22: 23767474-23767474 on Build GRCh38
rs759251168 Chr.22: 23767516-23767516 on Build GRCh38
rs781393947 Chr.22: 23767364-23767364 on Build GRCh38
rs730880033 Chr.22: 23767463-23767463 on Build GRCh38
rs374211312 Chr.22: 23767439-23767439 on Build GRCh38
rs901394195 Chr.22: 23767669-23767669 on Build GRCh38
rs1033231706 Chr.22: 23767729-23767729 on Build GRCh38
rs754782836 Chr.22: 23767428-23767428 on Build GRCh38

View Full Product Details