Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23836419-23836916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DERL3
Entrez Gene ID
91319
Gene Name
derlin 3
Gene Aliases
C22orf14, IZP6, LLN2, derlin-3
UniGene
Hs.593679
Gene Chromosome Location
Chr.22: 23834503-23839344 on Build GRCh38
Transcripts
NM_001002862.2 NM_001135751.1 NM_198440.3 XM_011530505.2 XM_011530506.2 XM_017029078.1 XM_017029079.1 XM_017029080.1 XM_017029082.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6370536 c.682C>T p.P228S Chr.22: 23836895-23836895 on Build GRCh38
5484477 c.697C>A p.P233T Chr.22: 23836880-23836880 on Build GRCh38
4450582 c.698delC p.P233fs*>3 Chr.22: 23836879-23836879 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs903381090 Chr.22: 23836564-23836564 on Build GRCh38
rs1007433750 Chr.22: 23836615-23836615 on Build GRCh38
rs963308635 Chr.22: 23836624-23836624 on Build GRCh38
rs904815018 Chr.22: 23836471-23836471 on Build GRCh38
rs754291104 Chr.22: 23836877-23836878 on Build GRCh38
rs201545808 Chr.22: 23836851-23836851 on Build GRCh38
rs899062136 Chr.22: 23836693-23836693 on Build GRCh38
rs899047176 Chr.22: 23836876-23836876 on Build GRCh38
rs933056828 Chr.22: 23836575-23836575 on Build GRCh38
rs183685204 Chr.22: 23836865-23836865 on Build GRCh38
rs970185705 Chr.22: 23836768-23836768 on Build GRCh38
rs1026093304 Chr.22: 23836716-23836716 on Build GRCh38
rs879842005 Chr.22: 23836531-23836559 on Build GRCh38
rs991460563 Chr.22: 23836802-23836802 on Build GRCh38
rs577919346 Chr.22: 23836452-23836453 on Build GRCh38
rs1009327027 Chr.22: 23836520-23836520 on Build GRCh38
rs765949393 Chr.22: 23836867-23836868 on Build GRCh38
rs950819212 Chr.22: 23836671-23836671 on Build GRCh38
rs973782701 Chr.22: 23836653-23836653 on Build GRCh38
rs575259988 Chr.22: 23836509-23836509 on Build GRCh38
rs574670001 Chr.22: 23836625-23836625 on Build GRCh38
rs537816636 Chr.22: 23836518-23836518 on Build GRCh38
rs917353917 Chr.22: 23836764-23836764 on Build GRCh38
rs199845871 Chr.22: 23836823-23836823 on Build GRCh38
rs34590336 Chr.22: 23836708-23836709 on Build GRCh38
rs917424894 Chr.22: 23836440-23836440 on Build GRCh38
rs146532594 Chr.22: 23836661-23836661 on Build GRCh38
rs554443728 Chr.22: 23836595-23836595 on Build GRCh38
rs370457454 Chr.22: 23836446-23836446 on Build GRCh38
rs993370589 Chr.22: 23836878-23836878 on Build GRCh38
rs992921941 Chr.22: 23836710-23836710 on Build GRCh38
rs527714153 Chr.22: 23836445-23836445 on Build GRCh38
rs948927452 Chr.22: 23836442-23836442 on Build GRCh38
rs202246735 Chr.22: 23836850-23836850 on Build GRCh38
rs1053476888 Chr.22: 23836500-23836500 on Build GRCh38
rs113084135 Chr.22: 23836842-23836842 on Build GRCh38
rs368846689 Chr.22: 23836840-23836840 on Build GRCh38
rs888863989 Chr.22: 23836654-23836654 on Build GRCh38
rs746452240 Chr.22: 23836859-23836859 on Build GRCh38
rs769544563 Chr.22: 23836881-23836881 on Build GRCh38
rs763794922 Chr.22: 23836592-23836592 on Build GRCh38
rs200378544 Chr.22: 23836864-23836864 on Build GRCh38
rs775343325 Chr.22: 23836890-23836890 on Build GRCh38
rs766304663 Chr.22: 23836770-23836770 on Build GRCh38
rs562534869 Chr.22: 23836856-23836856 on Build GRCh38
rs762241597 Chr.22: 23836715-23836715 on Build GRCh38
rs980264904 Chr.22: 23836806-23836806 on Build GRCh38
rs116973561 Chr.22: 23836879-23836879 on Build GRCh38
rs936324475 Chr.22: 23836496-23836496 on Build GRCh38
rs977703392 Chr.22: 23836839-23836839 on Build GRCh38
rs1017514310 Chr.22: 23836618-23836618 on Build GRCh38
rs936272267 Chr.22: 23836834-23836834 on Build GRCh38
rs1030539461 Chr.22: 23836556-23836556 on Build GRCh38
rs777433321 Chr.22: 23836858-23836858 on Build GRCh38
rs892182925 Chr.22: 23836513-23836513 on Build GRCh38
rs959052565 Chr.22: 23836791-23836791 on Build GRCh38

View Full Product Details