Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24225155-24225655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT5
Entrez Gene ID
2687
Gene Name
gamma-glutamyltransferase 5
Gene Aliases
GGL, GGT 5, GGT-REL, GGTLA1
UniGene
Hs.437156
Gene Chromosome Location
Chr.22: 24219654-24245142 on Build GRCh38
Transcripts
NM_001099781.2 NM_001099782.2 NM_001302464.1 NM_001302465.1 NM_004121.3 XM_005261557.2 XM_005261558.2 XM_011530133.1 XM_011530134.1 XM_011530135.1 XM_011530136.1 XM_011530137.2 XM_017028768.1 XM_017028769.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4399676 c.1466G>A p.G489E Chr.22: 24225282-24225282 on Build GRCh38
5057727 c.1446_1447insG p.S483fs*30 Chr.22: 24225301-24225302 on Build GRCh38
4164902 c.1430T>A p.I477N Chr.22: 24225318-24225318 on Build GRCh38
3552628 c.1261G>A p.G421S Chr.22: 24225621-24225621 on Build GRCh38
6891914 c.1410C>T p.S470S Chr.22: 24225338-24225338 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777898121 Chr.22: 24225337-24225337 on Build GRCh38
rs1041111035 Chr.22: 24225304-24225304 on Build GRCh38
rs762309530 Chr.22: 24225448-24225448 on Build GRCh38
rs765588510 Chr.22: 24225263-24225263 on Build GRCh38
rs374329258 Chr.22: 24225433-24225433 on Build GRCh38
rs200937485 Chr.22: 24225548-24225548 on Build GRCh38
rs752530955 Chr.22: 24225583-24225583 on Build GRCh38
rs776163245 Chr.22: 24225579-24225579 on Build GRCh38
rs755737041 Chr.22: 24225388-24225388 on Build GRCh38
rs777095964 Chr.22: 24225223-24225223 on Build GRCh38
rs774748717 Chr.22: 24225446-24225446 on Build GRCh38
rs200684960 Chr.22: 24225222-24225222 on Build GRCh38
rs542440435 Chr.22: 24225396-24225396 on Build GRCh38
rs371281442 Chr.22: 24225565-24225565 on Build GRCh38
rs756289453 Chr.22: 24225588-24225588 on Build GRCh38
rs1011799844 Chr.22: 24225465-24225465 on Build GRCh38
rs766946185 Chr.22: 24225455-24225455 on Build GRCh38
rs779215682 Chr.22: 24225622-24225622 on Build GRCh38
rs745618291 Chr.22: 24225198-24225198 on Build GRCh38
rs781195762 Chr.22: 24225524-24225524 on Build GRCh38
rs145909552 Chr.22: 24225604-24225604 on Build GRCh38
rs759124656 Chr.22: 24225581-24225581 on Build GRCh38
rs761249346 Chr.22: 24225379-24225379 on Build GRCh38
rs760315913 Chr.22: 24225463-24225463 on Build GRCh38
rs747032399 Chr.22: 24225342-24225342 on Build GRCh38
rs999205973 Chr.22: 24225265-24225265 on Build GRCh38
rs75273222 Chr.22: 24225572-24225572 on Build GRCh38
rs200073930 Chr.22: 24225618-24225618 on Build GRCh38
rs928685613 Chr.22: 24225598-24225598 on Build GRCh38
rs192160185 Chr.22: 24225506-24225506 on Build GRCh38
rs148922656 Chr.22: 24225554-24225554 on Build GRCh38
rs758448257 Chr.22: 24225496-24225519 on Build GRCh38
rs759800101 Chr.22: 24225234-24225234 on Build GRCh38
rs55890517 Chr.22: 24225426-24225426 on Build GRCh38
rs749181027 Chr.22: 24225212-24225212 on Build GRCh38
rs367693056 Chr.22: 24225569-24225569 on Build GRCh38
rs748665403 Chr.22: 24225564-24225564 on Build GRCh38
rs371880180 Chr.22: 24225558-24225558 on Build GRCh38
rs183990532 Chr.22: 24225575-24225575 on Build GRCh38
rs756137997 Chr.22: 24225604-24225606 on Build GRCh38
rs774717334 Chr.22: 24225557-24225557 on Build GRCh38
rs764663086 Chr.22: 24225370-24225370 on Build GRCh38
rs112092021 Chr.22: 24225190-24225190 on Build GRCh38
rs767011018 Chr.22: 24225382-24225382 on Build GRCh38
rs916055461 Chr.22: 24225620-24225620 on Build GRCh38
rs769089462 Chr.22: 24225445-24225445 on Build GRCh38
rs747475402 Chr.22: 24225627-24225627 on Build GRCh38
rs564711431 Chr.22: 24225295-24225295 on Build GRCh38
rs768572532 Chr.22: 24225331-24225331 on Build GRCh38
rs756452604 Chr.22: 24225528-24225528 on Build GRCh38
rs925476156 Chr.22: 24225476-24225476 on Build GRCh38
rs750040366 Chr.22: 24225300-24225300 on Build GRCh38
rs778801768 Chr.22: 24225416-24225416 on Build GRCh38
rs780218835 Chr.22: 24225629-24225629 on Build GRCh38
rs143616956 Chr.22: 24225383-24225383 on Build GRCh38
rs113295783 Chr.22: 24225243-24225243 on Build GRCh38
rs374828667 Chr.22: 24225585-24225585 on Build GRCh38
rs780434605 Chr.22: 24225412-24225412 on Build GRCh38
rs772642258 Chr.22: 24225571-24225571 on Build GRCh38
rs150898918 Chr.22: 24225284-24225284 on Build GRCh38
rs755235008 Chr.22: 24225619-24225619 on Build GRCh38
rs753628065 Chr.22: 24225411-24225411 on Build GRCh38
rs779874745 Chr.22: 24225206-24225206 on Build GRCh38
rs1023091411 Chr.22: 24225576-24225576 on Build GRCh38
rs528779081 Chr.22: 24225303-24225303 on Build GRCh38
rs139383582 Chr.22: 24225299-24225299 on Build GRCh38
rs765031471 Chr.22: 24225582-24225582 on Build GRCh38
rs367843924 Chr.22: 24225617-24225617 on Build GRCh38
rs374350087 Chr.22: 24225497-24225497 on Build GRCh38
rs73167566 Chr.22: 24225553-24225553 on Build GRCh38
rs781271175 Chr.22: 24225503-24225504 on Build GRCh38
rs778369692 Chr.22: 24225626-24225626 on Build GRCh38
rs12627971 Chr.22: 24225523-24225523 on Build GRCh38
rs752800899 Chr.22: 24225323-24225324 on Build GRCh38
rs894850599 Chr.22: 24225188-24225188 on Build GRCh38
rs903249652 Chr.22: 24225405-24225405 on Build GRCh38
rs866363144 Chr.22: 24225270-24225270 on Build GRCh38
rs772769849 Chr.22: 24225458-24225458 on Build GRCh38
rs748071790 Chr.22: 24225421-24225421 on Build GRCh38
rs776035379 Chr.22: 24225431-24225431 on Build GRCh38
rs751403095 Chr.22: 24225587-24225587 on Build GRCh38
rs765401891 Chr.22: 24225257-24225258 on Build GRCh38
rs752581530 Chr.22: 24225510-24225510 on Build GRCh38
rs202092652 Chr.22: 24225570-24225570 on Build GRCh38
rs140980900 Chr.22: 24225573-24225573 on Build GRCh38
rs1030702214 Chr.22: 24225521-24225521 on Build GRCh38
rs140834056 Chr.22: 24225586-24225586 on Build GRCh38
rs1030793569 Chr.22: 24225274-24225274 on Build GRCh38
rs201616574 Chr.22: 24225219-24225219 on Build GRCh38
rs984702061 Chr.22: 24225555-24225555 on Build GRCh38
rs903488327 Chr.22: 24225504-24225504 on Build GRCh38
rs776743050 Chr.22: 24225350-24225350 on Build GRCh38
rs201765477 Chr.22: 24225631-24225631 on Build GRCh38
rs751162471 Chr.22: 24225377-24225377 on Build GRCh38
rs771235441 Chr.22: 24225346-24225346 on Build GRCh38
rs201667511 Chr.22: 24225525-24225525 on Build GRCh38
rs185866792 Chr.22: 24225485-24225485 on Build GRCh38
rs892435899 Chr.22: 24225247-24225247 on Build GRCh38
rs985183445 Chr.22: 24225375-24225375 on Build GRCh38
rs1026801373 Chr.22: 24225351-24225351 on Build GRCh38
rs529621965 Chr.22: 24225359-24225359 on Build GRCh38
rs746133651 Chr.22: 24225427-24225427 on Build GRCh38
rs200146637 Chr.22: 24225356-24225356 on Build GRCh38
rs369915142 Chr.22: 24225204-24225204 on Build GRCh38
rs558318043 Chr.22: 24225428-24225428 on Build GRCh38
rs199510644 Chr.22: 24225214-24225214 on Build GRCh38
rs748517292 Chr.22: 24225624-24225624 on Build GRCh38
rs899898018 Chr.22: 24225538-24225538 on Build GRCh38
rs376709868 Chr.22: 24225369-24225369 on Build GRCh38
rs142733225 Chr.22: 24225625-24225625 on Build GRCh38
rs370949394 Chr.22: 24225509-24225509 on Build GRCh38
rs763495821 Chr.22: 24225283-24225283 on Build GRCh38
rs899927211 Chr.22: 24225317-24225317 on Build GRCh38
rs777819007 Chr.22: 24225541-24225541 on Build GRCh38
rs150033594 Chr.22: 24225302-24225302 on Build GRCh38
rs756973423 Chr.22: 24225296-24225296 on Build GRCh38
rs7288201 Chr.22: 24225325-24225325 on Build GRCh38
rs370492248 Chr.22: 24225275-24225275 on Build GRCh38
rs745862633 Chr.22: 24225504-24225504 on Build GRCh38
rs140052476 Chr.22: 24225355-24225355 on Build GRCh38

View Full Product Details