Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24540357-24540867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCD1
Entrez Gene ID
83606
Gene Name
guanylyl cyclase domain containing 1
Gene Aliases
C22orf13, LLN4
UniGene
Hs.9850
Gene Chromosome Location
Chr.22: 24540423-24555935 on Build GRCh38
Transcripts
NM_001284251.1 NM_001284252.1 NM_001284253.1 NM_001284254.1 NM_001284255.1 NM_001284256.1 NM_001284257.1 NM_031444.3 NR_104286.1 XM_005261761.1

SNP Detail

SNP ID Chromosome Location
rs758761062 Chr.22: 24540447-24540447 on Build GRCh38
rs977845843 Chr.22: 24540487-24540487 on Build GRCh38
rs777159446 Chr.22: 24540827-24540827 on Build GRCh38
rs769393058 Chr.22: 24540832-24540832 on Build GRCh38
rs979257492 Chr.22: 24540379-24540379 on Build GRCh38
rs536085172 Chr.22: 24540429-24540429 on Build GRCh38
rs377318634 Chr.22: 24540656-24540656 on Build GRCh38
rs768112066 Chr.22: 24540385-24540385 on Build GRCh38
rs774012289 Chr.22: 24540401-24540401 on Build GRCh38
rs940749750 Chr.22: 24540708-24540708 on Build GRCh38
rs986559725 Chr.22: 24540803-24540803 on Build GRCh38
rs773510990 Chr.22: 24540565-24540565 on Build GRCh38
rs931826110 Chr.22: 24540608-24540608 on Build GRCh38
rs1003312923 Chr.22: 24540834-24540834 on Build GRCh38
rs568550174 Chr.22: 24540395-24540395 on Build GRCh38
rs747736741 Chr.22: 24540417-24540417 on Build GRCh38
rs886611891 Chr.22: 24540570-24540570 on Build GRCh38
rs766837867 Chr.22: 24540741-24540741 on Build GRCh38
rs115676339 Chr.22: 24540553-24540553 on Build GRCh38
rs921873522 Chr.22: 24540563-24540563 on Build GRCh38
rs11259 Chr.22: 24540694-24540694 on Build GRCh38
rs1004927139 Chr.22: 24540598-24540598 on Build GRCh38
rs909054024 Chr.22: 24540637-24540637 on Build GRCh38
rs1012690066 Chr.22: 24540687-24540687 on Build GRCh38
rs1051290459 Chr.22: 24540503-24540503 on Build GRCh38
rs576957886 Chr.22: 24540783-24540783 on Build GRCh38
rs960730650 Chr.22: 24540658-24540658 on Build GRCh38
rs552194300 Chr.22: 24540529-24540529 on Build GRCh38
rs1039291691 Chr.22: 24540527-24540527 on Build GRCh38
rs780458643 Chr.22: 24540511-24540511 on Build GRCh38
rs979330832 Chr.22: 24540725-24540725 on Build GRCh38
rs1035552361 Chr.22: 24540642-24540642 on Build GRCh38
rs71805047 Chr.22: 24540746-24540747 on Build GRCh38
rs763255706 Chr.22: 24540699-24540700 on Build GRCh38
rs146838757 Chr.22: 24540530-24540530 on Build GRCh38
rs1048922681 Chr.22: 24540617-24540617 on Build GRCh38
rs912228105 Chr.22: 24540506-24540506 on Build GRCh38
rs569537373 Chr.22: 24540599-24540599 on Build GRCh38
rs761083336 Chr.22: 24540650-24540650 on Build GRCh38
rs942395030 Chr.22: 24540514-24540514 on Build GRCh38
rs747383140 Chr.22: 24540534-24540534 on Build GRCh38
rs878861320 Chr.22: 24540567-24540567 on Build GRCh38
rs935061361 Chr.22: 24540453-24540453 on Build GRCh38
rs923574420 Chr.22: 24540519-24540519 on Build GRCh38
rs539710907 Chr.22: 24540649-24540649 on Build GRCh38
rs796394609 Chr.22: 24540705-24540706 on Build GRCh38
rs1022049104 Chr.22: 24540422-24540422 on Build GRCh38

View Full Product Details