Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24592772-24593270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGT1
Entrez Gene ID
2678
Gene Name
gamma-glutamyltransferase 1
Gene Aliases
CD224, D22S672, D22S732, GGT, GGT 1, GTG
UniGene
Hs.595809
Gene Chromosome Location
Chr.22: 24583750-24629005 on Build GRCh38
Transcripts
NM_013430.2 NM_207644.2 XM_005261600.2

Gene Symbol
LRRC75B
Entrez Gene ID
388886
Gene Name
leucine rich repeat containing 75B
Gene Aliases
C22orf36, FAM211B
UniGene
Hs.113314
Gene Chromosome Location
Chr.22: 24585620-24593104 on Build GRCh38
Transcripts
NM_013430.2 NM_207644.2 XM_005261600.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4156060 c.87G>A p.E29E Chr.22: 24592953-24592953 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs183293135 Chr.22: 24593226-24593226 on Build GRCh38
rs552282051 Chr.22: 24593053-24593053 on Build GRCh38
rs892170813 Chr.22: 24593146-24593146 on Build GRCh38
rs746252814 Chr.22: 24593191-24593191 on Build GRCh38
rs1000643265 Chr.22: 24593127-24593127 on Build GRCh38
rs751036249 Chr.22: 24593074-24593074 on Build GRCh38
rs921984341 Chr.22: 24592963-24592963 on Build GRCh38
rs927787314 Chr.22: 24592848-24592848 on Build GRCh38
rs1030700960 Chr.22: 24592845-24592845 on Build GRCh38
rs1019167095 Chr.22: 24593157-24593157 on Build GRCh38
rs559553917 Chr.22: 24592844-24592844 on Build GRCh38
rs990647719 Chr.22: 24593141-24593141 on Build GRCh38
rs930755180 Chr.22: 24592969-24592969 on Build GRCh38
rs780771421 Chr.22: 24593185-24593185 on Build GRCh38
rs57883872 Chr.22: 24593180-24593180 on Build GRCh38
rs759931550 Chr.22: 24592958-24592958 on Build GRCh38
rs947841961 Chr.22: 24593042-24593042 on Build GRCh38
rs747416487 Chr.22: 24592898-24592898 on Build GRCh38
rs996718356 Chr.22: 24593177-24593177 on Build GRCh38
rs563748821 Chr.22: 24592999-24592999 on Build GRCh38
rs1027762535 Chr.22: 24593217-24593217 on Build GRCh38
rs1023772084 Chr.22: 24592820-24592820 on Build GRCh38
rs530802654 Chr.22: 24593038-24593038 on Build GRCh38
rs745448300 Chr.22: 24592839-24592839 on Build GRCh38
rs961333770 Chr.22: 24592822-24592822 on Build GRCh38
rs1007904642 Chr.22: 24593153-24593153 on Build GRCh38
rs1056343972 Chr.22: 24593036-24593036 on Build GRCh38
rs546800774 Chr.22: 24593206-24593206 on Build GRCh38
rs1056210020 Chr.22: 24592805-24592805 on Build GRCh38
rs966165440 Chr.22: 24592939-24592939 on Build GRCh38
rs902489058 Chr.22: 24593181-24593181 on Build GRCh38
rs956257248 Chr.22: 24592858-24592858 on Build GRCh38
rs888232669 Chr.22: 24593219-24593219 on Build GRCh38
rs372869454 Chr.22: 24593092-24593092 on Build GRCh38
rs112108503 Chr.22: 24592838-24592838 on Build GRCh38
rs1011992746 Chr.22: 24592812-24592812 on Build GRCh38
rs961986244 Chr.22: 24593229-24593229 on Build GRCh38
rs944961930 Chr.22: 24593190-24593190 on Build GRCh38
rs940643824 Chr.22: 24592801-24592802 on Build GRCh38
rs737128 Chr.22: 24592953-24592953 on Build GRCh38
rs553659541 Chr.22: 24593174-24593174 on Build GRCh38
rs896365116 Chr.22: 24592809-24592809 on Build GRCh38
rs555026485 Chr.22: 24592877-24592877 on Build GRCh38
rs985185088 Chr.22: 24592988-24592988 on Build GRCh38
rs937795241 Chr.22: 24593132-24593132 on Build GRCh38
rs989100976 Chr.22: 24592865-24592865 on Build GRCh38
rs992123152 Chr.22: 24593236-24593236 on Build GRCh38
rs940611154 Chr.22: 24593017-24593017 on Build GRCh38
rs915350366 Chr.22: 24593160-24593160 on Build GRCh38
rs867922349 Chr.22: 24593208-24593208 on Build GRCh38
rs530009323 Chr.22: 24592908-24592908 on Build GRCh38
rs1019114133 Chr.22: 24592889-24592889 on Build GRCh38

View Full Product Details