Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 25203673-25204178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYBB3
Entrez Gene ID
1417
Gene Name
crystallin beta B3
Gene Aliases
CATCN2, CRYB3, CTRCT22
UniGene
Hs.533022
Gene Chromosome Location
Chr.22: 25199312-25207617 on Build GRCh38
Transcripts
NM_004076.4 XM_011529902.2 XM_017028599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4895268 c.223C>T p.R75C Chr.22: 25203791-25203791 on Build GRCh38
3759092 c.314G>A p.R105Q Chr.22: 25203882-25203882 on Build GRCh38
5634925 c.325A>G p.I109V Chr.22: 25203893-25203893 on Build GRCh38
280137 c.224G>A p.R75H Chr.22: 25203792-25203792 on Build GRCh38
6635858 c.265T>C p.W89R Chr.22: 25203833-25203833 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778806905 Chr.22: 25204013-25204014 on Build GRCh38
rs756452191 Chr.22: 25203917-25203917 on Build GRCh38
rs373102556 Chr.22: 25203809-25203809 on Build GRCh38
rs1032163713 Chr.22: 25204005-25204005 on Build GRCh38
rs775096922 Chr.22: 25203834-25203834 on Build GRCh38
rs138167578 Chr.22: 25203831-25203831 on Build GRCh38
rs953624839 Chr.22: 25203857-25203857 on Build GRCh38
rs746418605 Chr.22: 25203915-25203915 on Build GRCh38
rs751605828 Chr.22: 25203900-25203900 on Build GRCh38
rs768166313 Chr.22: 25203839-25203839 on Build GRCh38
rs747513628 Chr.22: 25203813-25203813 on Build GRCh38
rs1006768986 Chr.22: 25203872-25203872 on Build GRCh38
rs901613248 Chr.22: 25204099-25204099 on Build GRCh38
rs779077251 Chr.22: 25203739-25203739 on Build GRCh38
rs771347586 Chr.22: 25203818-25203818 on Build GRCh38
rs565359059 Chr.22: 25204056-25204056 on Build GRCh38
rs201863246 Chr.22: 25203781-25203781 on Build GRCh38
rs925135274 Chr.22: 25204047-25204047 on Build GRCh38
rs748932591 Chr.22: 25203947-25203947 on Build GRCh38
rs753007977 Chr.22: 25203806-25203806 on Build GRCh38
rs776894567 Chr.22: 25203771-25203771 on Build GRCh38
rs192527959 Chr.22: 25203756-25203756 on Build GRCh38
rs183587921 Chr.22: 25203792-25203792 on Build GRCh38
rs767272833 Chr.22: 25203794-25203794 on Build GRCh38
rs372687815 Chr.22: 25203728-25203728 on Build GRCh38
rs916467389 Chr.22: 25203994-25203994 on Build GRCh38
rs772668016 Chr.22: 25203786-25203786 on Build GRCh38
rs200104890 Chr.22: 25203849-25203849 on Build GRCh38
rs1006471324 Chr.22: 25204143-25204143 on Build GRCh38
rs201028562 Chr.22: 25203802-25203802 on Build GRCh38
rs999200147 Chr.22: 25203817-25203817 on Build GRCh38
rs972597491 Chr.22: 25203700-25203700 on Build GRCh38
rs575459371 Chr.22: 25204022-25204022 on Build GRCh38
rs773400804 Chr.22: 25203822-25203822 on Build GRCh38
rs1013210463 Chr.22: 25203908-25203908 on Build GRCh38
rs770214669 Chr.22: 25203832-25203832 on Build GRCh38
rs769821740 Chr.22: 25204051-25204051 on Build GRCh38
rs376115617 Chr.22: 25203881-25203881 on Build GRCh38
rs781479810 Chr.22: 25203830-25203830 on Build GRCh38
rs777736274 Chr.22: 25203811-25203811 on Build GRCh38
rs760515602 Chr.22: 25203875-25203875 on Build GRCh38
rs766325634 Chr.22: 25203878-25203878 on Build GRCh38
rs748224794 Chr.22: 25203747-25203747 on Build GRCh38
rs779269345 Chr.22: 25203805-25203805 on Build GRCh38
rs140588220 Chr.22: 25204082-25204082 on Build GRCh38
rs188334396 Chr.22: 25204155-25204155 on Build GRCh38
rs775185175 Chr.22: 25203788-25203788 on Build GRCh38
rs992305494 Chr.22: 25203736-25203736 on Build GRCh38
rs761680838 Chr.22: 25203793-25203793 on Build GRCh38
rs991466464 Chr.22: 25203984-25203984 on Build GRCh38
rs925090526 Chr.22: 25203727-25203727 on Build GRCh38
rs928738788 Chr.22: 25203702-25203702 on Build GRCh38
rs1048496531 Chr.22: 25204136-25204136 on Build GRCh38
rs781662479 Chr.22: 25203912-25203912 on Build GRCh38
rs1043908493 Chr.22: 25204044-25204044 on Build GRCh38
rs759124927 Chr.22: 25203779-25203779 on Build GRCh38
rs144697511 Chr.22: 25203770-25203770 on Build GRCh38
rs997156857 Chr.22: 25204107-25204107 on Build GRCh38
rs1052173980 Chr.22: 25204091-25204091 on Build GRCh38
rs912468962 Chr.22: 25204046-25204046 on Build GRCh38
rs376046044 Chr.22: 25203754-25203754 on Build GRCh38
rs141617094 Chr.22: 25203791-25203791 on Build GRCh38
rs935192722 Chr.22: 25204081-25204081 on Build GRCh38
rs938733733 Chr.22: 25203768-25203768 on Build GRCh38
rs16979730 Chr.22: 25204025-25204025 on Build GRCh38
rs778411611 Chr.22: 25203753-25203756 on Build GRCh38
rs529648288 Chr.22: 25204137-25204137 on Build GRCh38
rs755253697 Chr.22: 25203734-25203734 on Build GRCh38
rs943935869 Chr.22: 25204075-25204075 on Build GRCh38
rs985431612 Chr.22: 25204026-25204026 on Build GRCh38
rs559345693 Chr.22: 25204117-25204117 on Build GRCh38
rs113872601 Chr.22: 25203855-25203855 on Build GRCh38
rs747451205 Chr.22: 25203947-25203948 on Build GRCh38
rs17670506 Chr.22: 25203882-25203882 on Build GRCh38
rs534724280 Chr.22: 25203854-25203854 on Build GRCh38
rs757415735 Chr.22: 25203909-25203909 on Build GRCh38
rs746328073 Chr.22: 25203935-25203935 on Build GRCh38
rs143187630 Chr.22: 25203896-25203896 on Build GRCh38
rs577384441 Chr.22: 25203847-25203847 on Build GRCh38
rs762565779 Chr.22: 25203838-25203838 on Build GRCh38
rs748589671 Chr.22: 25203759-25203759 on Build GRCh38
rs564651735 Chr.22: 25204030-25204030 on Build GRCh38
rs200457939 Chr.22: 25203803-25203803 on Build GRCh38
rs937503457 Chr.22: 25204146-25204146 on Build GRCh38

View Full Product Details