Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89109908-89110313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP2
Entrez Gene ID
2634
Gene Name
guanylate binding protein 2
Gene Aliases
-
UniGene
Hs.386567
Gene Chromosome Location
Chr.1: 89106132-89126159 on Build GRCh38
Transcripts
NM_004120.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
330176 c.1383delA p.K461fs*32 Chr.1: 89110246-89110246 on Build GRCh38
7262620 c.1407G>T p.V469V Chr.1: 89110222-89110222 on Build GRCh38
6669784 c.1369G>T p.E457* Chr.1: 89110260-89110260 on Build GRCh38
1602759 c.1430A>C p.D477A Chr.1: 89110199-89110199 on Build GRCh38
4808081 c.1392G>A p.E464E Chr.1: 89110237-89110237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767224206 Chr.1: 89110208-89110208 on Build GRCh38
rs751782865 Chr.1: 89110259-89110259 on Build GRCh38
rs985067655 Chr.1: 89110272-89110272 on Build GRCh38
rs149073470 Chr.1: 89110065-89110065 on Build GRCh38
rs1009482104 Chr.1: 89109969-89109969 on Build GRCh38
rs144969028 Chr.1: 89110038-89110038 on Build GRCh38
rs746346593 Chr.1: 89110285-89110285 on Build GRCh38
rs377174334 Chr.1: 89110171-89110171 on Build GRCh38
rs190165784 Chr.1: 89110137-89110137 on Build GRCh38
rs2026043 Chr.1: 89109994-89109994 on Build GRCh38
rs535061352 Chr.1: 89110044-89110044 on Build GRCh38
rs760557109 Chr.1: 89110212-89110212 on Build GRCh38
rs896497703 Chr.1: 89109965-89109965 on Build GRCh38
rs757627951 Chr.1: 89110146-89110146 on Build GRCh38
rs374464113 Chr.1: 89109966-89109966 on Build GRCh38
rs1025530644 Chr.1: 89110138-89110138 on Build GRCh38
rs971729097 Chr.1: 89110225-89110225 on Build GRCh38
rs778818879 Chr.1: 89110019-89110019 on Build GRCh38
rs754026373 Chr.1: 89110243-89110243 on Build GRCh38
rs1001466583 Chr.1: 89110084-89110084 on Build GRCh38
rs1028490681 Chr.1: 89110105-89110105 on Build GRCh38
rs41288377 Chr.1: 89109940-89109940 on Build GRCh38
rs200764009 Chr.1: 89110251-89110251 on Build GRCh38
rs769298738 Chr.1: 89110166-89110166 on Build GRCh38
rs144421688 Chr.1: 89110133-89110133 on Build GRCh38
rs781431603 Chr.1: 89110276-89110276 on Build GRCh38
rs376348185 Chr.1: 89110109-89110109 on Build GRCh38
rs891206775 Chr.1: 89109975-89109975 on Build GRCh38
rs775172046 Chr.1: 89110168-89110168 on Build GRCh38
rs374892284 Chr.1: 89110132-89110132 on Build GRCh38
rs1035972357 Chr.1: 89110206-89110206 on Build GRCh38
rs1056423419 Chr.1: 89109962-89109962 on Build GRCh38
rs753544245 Chr.1: 89110118-89110118 on Build GRCh38
rs750741832 Chr.1: 89109939-89109939 on Build GRCh38
rs773147387 Chr.1: 89110211-89110211 on Build GRCh38
rs201969713 Chr.1: 89110172-89110172 on Build GRCh38
rs757583481 Chr.1: 89110264-89110264 on Build GRCh38
rs545998240 Chr.1: 89110153-89110153 on Build GRCh38
rs150718831 Chr.1: 89110242-89110242 on Build GRCh38
rs758844720 Chr.1: 89110061-89110061 on Build GRCh38
rs761667785 Chr.1: 89110174-89110174 on Build GRCh38
rs138667725 Chr.1: 89110235-89110235 on Build GRCh38
rs746374299 Chr.1: 89110164-89110164 on Build GRCh38

View Full Product Details