Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89117310-89117814 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP2
Entrez Gene ID
2634
Gene Name
guanylate binding protein 2
Gene Aliases
-
UniGene
Hs.386567
Gene Chromosome Location
Chr.1: 89106132-89126159 on Build GRCh38
Transcripts
NM_004120.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
682795 c.512G>C p.S171T Chr.1: 89117690-89117690 on Build GRCh38
4777019 c.520C>G p.P174A Chr.1: 89117682-89117682 on Build GRCh38
2258848 c.452G>A p.R151Q Chr.1: 89117750-89117750 on Build GRCh38
1688008 c.515T>A p.F172Y Chr.1: 89117687-89117687 on Build GRCh38
6669787 c.522A>G p.P174P Chr.1: 89117680-89117680 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764713117 Chr.1: 89117479-89117480 on Build GRCh38
rs140765409 Chr.1: 89117750-89117750 on Build GRCh38
rs374009578 Chr.1: 89117561-89117561 on Build GRCh38
rs762133629 Chr.1: 89117631-89117631 on Build GRCh38
rs199628302 Chr.1: 89117694-89117694 on Build GRCh38
rs1008359044 Chr.1: 89117583-89117583 on Build GRCh38
rs975801392 Chr.1: 89117596-89117596 on Build GRCh38
rs758001210 Chr.1: 89117718-89117718 on Build GRCh38
rs756647039 Chr.1: 89117550-89117550 on Build GRCh38
rs943121524 Chr.1: 89117345-89117345 on Build GRCh38
rs776295815 Chr.1: 89117662-89117663 on Build GRCh38
rs1001046025 Chr.1: 89117484-89117484 on Build GRCh38
rs894232187 Chr.1: 89117351-89117351 on Build GRCh38
rs915641793 Chr.1: 89117747-89117747 on Build GRCh38
rs374791564 Chr.1: 89117618-89117618 on Build GRCh38
rs761112927 Chr.1: 89117780-89117780 on Build GRCh38
rs781096238 Chr.1: 89117742-89117742 on Build GRCh38
rs750928912 Chr.1: 89117536-89117536 on Build GRCh38
rs774447945 Chr.1: 89117751-89117751 on Build GRCh38
rs768273867 Chr.1: 89117623-89117623 on Build GRCh38
rs1043213149 Chr.1: 89117766-89117766 on Build GRCh38
rs772362131 Chr.1: 89117756-89117756 on Build GRCh38
rs758100201 Chr.1: 89117581-89117581 on Build GRCh38
rs766827297 Chr.1: 89117731-89117731 on Build GRCh38
rs777420110 Chr.1: 89117726-89117726 on Build GRCh38
rs201208534 Chr.1: 89117663-89117663 on Build GRCh38
rs752182723 Chr.1: 89117714-89117714 on Build GRCh38
rs370782210 Chr.1: 89117617-89117617 on Build GRCh38
rs375803227 Chr.1: 89117542-89117542 on Build GRCh38
rs761131354 Chr.1: 89117689-89117689 on Build GRCh38
rs748199367 Chr.1: 89117752-89117752 on Build GRCh38
rs147240474 Chr.1: 89117650-89117650 on Build GRCh38
rs181512309 Chr.1: 89117371-89117371 on Build GRCh38
rs773661804 Chr.1: 89117672-89117672 on Build GRCh38
rs901534140 Chr.1: 89117412-89117412 on Build GRCh38
rs879687682 Chr.1: 89117577-89117577 on Build GRCh38
rs754433571 Chr.1: 89117702-89117702 on Build GRCh38
rs369852231 Chr.1: 89117552-89117552 on Build GRCh38
rs375563316 Chr.1: 89117337-89117337 on Build GRCh38
rs938458902 Chr.1: 89117730-89117730 on Build GRCh38
rs149996423 Chr.1: 89117593-89117593 on Build GRCh38
rs746739891 Chr.1: 89117735-89117735 on Build GRCh38
rs576676491 Chr.1: 89117707-89117707 on Build GRCh38
rs112937106 Chr.1: 89117655-89117655 on Build GRCh38
rs887675600 Chr.1: 89117507-89117507 on Build GRCh38
rs201386199 Chr.1: 89117781-89117781 on Build GRCh38
rs142567865 Chr.1: 89117746-89117746 on Build GRCh38
rs753338590 Chr.1: 89117556-89117556 on Build GRCh38
rs768043075 Chr.1: 89117528-89117528 on Build GRCh38
rs747889476 Chr.1: 89117562-89117562 on Build GRCh38
rs142187312 Chr.1: 89117710-89117710 on Build GRCh38
rs919967277 Chr.1: 89117603-89117603 on Build GRCh38
rs576540271 Chr.1: 89117404-89117404 on Build GRCh38
rs774412587 Chr.1: 89117628-89117628 on Build GRCh38
rs1040086434 Chr.1: 89117369-89117369 on Build GRCh38
rs761541886 Chr.1: 89117759-89117760 on Build GRCh38
rs754430142 Chr.1: 89117709-89117709 on Build GRCh38
rs770798916 Chr.1: 89117594-89117594 on Build GRCh38
rs904092787 Chr.1: 89117435-89117435 on Build GRCh38
rs377010688 Chr.1: 89117572-89117572 on Build GRCh38
rs773569604 Chr.1: 89117761-89117761 on Build GRCh38
rs202147173 Chr.1: 89117496-89117496 on Build GRCh38

View Full Product Details