Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89186919-89187419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP4
Entrez Gene ID
115361
Gene Name
guanylate binding protein 4
Gene Aliases
Mpa2
UniGene
Hs.409925
Gene Chromosome Location
Chr.1: 89181148-89198950 on Build GRCh38
Transcripts
NM_052941.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3493172 c.1485C>T p.A495A Chr.1: 89187028-89187028 on Build GRCh38
3493173 c.1434C>T p.F478F Chr.1: 89187079-89187079 on Build GRCh38
682788 c.1419G>T p.E473D Chr.1: 89187094-89187094 on Build GRCh38
7291693 c.1438C>T p.Q480* Chr.1: 89187075-89187075 on Build GRCh38
682789 c.1428G>A p.Q476Q Chr.1: 89187085-89187085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768688649 Chr.1: 89187046-89187046 on Build GRCh38
rs992168255 Chr.1: 89187044-89187044 on Build GRCh38
rs557167243 Chr.1: 89187245-89187245 on Build GRCh38
rs773378979 Chr.1: 89187033-89187036 on Build GRCh38
rs996413523 Chr.1: 89187258-89187258 on Build GRCh38
rs780956701 Chr.1: 89187230-89187230 on Build GRCh38
rs1007862537 Chr.1: 89187196-89187196 on Build GRCh38
rs777535950 Chr.1: 89187391-89187391 on Build GRCh38
rs937923171 Chr.1: 89187312-89187312 on Build GRCh38
rs142086287 Chr.1: 89187017-89187017 on Build GRCh38
rs149450766 Chr.1: 89187162-89187162 on Build GRCh38
rs763410577 Chr.1: 89187150-89187154 on Build GRCh38
rs1019256191 Chr.1: 89187200-89187200 on Build GRCh38
rs766436671 Chr.1: 89187064-89187064 on Build GRCh38
rs765445692 Chr.1: 89187107-89187107 on Build GRCh38
rs539707194 Chr.1: 89187323-89187323 on Build GRCh38
rs371055870 Chr.1: 89187133-89187133 on Build GRCh38
rs745465221 Chr.1: 89186956-89186956 on Build GRCh38
rs776589812 Chr.1: 89187065-89187065 on Build GRCh38
rs895359224 Chr.1: 89187284-89187284 on Build GRCh38
rs535659893 Chr.1: 89187190-89187190 on Build GRCh38
rs920012883 Chr.1: 89187248-89187248 on Build GRCh38
rs193063634 Chr.1: 89187198-89187198 on Build GRCh38
rs377546209 Chr.1: 89186952-89186952 on Build GRCh38
rs775349718 Chr.1: 89187006-89187006 on Build GRCh38
rs756834313 Chr.1: 89186951-89186951 on Build GRCh38
rs534736583 Chr.1: 89187295-89187314 on Build GRCh38
rs200881341 Chr.1: 89187314-89187314 on Build GRCh38
rs199938928 Chr.1: 89187111-89187111 on Build GRCh38
rs567744668 Chr.1: 89187175-89187175 on Build GRCh38
rs984578078 Chr.1: 89187278-89187278 on Build GRCh38
rs951845469 Chr.1: 89187263-89187263 on Build GRCh38
rs755773876 Chr.1: 89187144-89187144 on Build GRCh38
rs148621560 Chr.1: 89187361-89187361 on Build GRCh38
rs202178008 Chr.1: 89187128-89187128 on Build GRCh38
rs947941358 Chr.1: 89187389-89187389 on Build GRCh38
rs769985149 Chr.1: 89186971-89186972 on Build GRCh38
rs1044904349 Chr.1: 89187131-89187131 on Build GRCh38
rs749058427 Chr.1: 89187146-89187146 on Build GRCh38
rs533399156 Chr.1: 89187097-89187097 on Build GRCh38
rs954317844 Chr.1: 89187157-89187157 on Build GRCh38
rs753046594 Chr.1: 89187112-89187112 on Build GRCh38
rs917948068 Chr.1: 89187388-89187388 on Build GRCh38
rs138486401 Chr.1: 89187047-89187047 on Build GRCh38
rs374218635 Chr.1: 89187068-89187068 on Build GRCh38
rs780407654 Chr.1: 89187145-89187145 on Build GRCh38
rs758813899 Chr.1: 89187119-89187119 on Build GRCh38
rs572798577 Chr.1: 89187375-89187375 on Build GRCh38
rs368263528 Chr.1: 89187121-89187121 on Build GRCh38
rs12129014 Chr.1: 89187163-89187163 on Build GRCh38
rs917812273 Chr.1: 89187096-89187096 on Build GRCh38
rs752012458 Chr.1: 89187126-89187126 on Build GRCh38
rs779830769 Chr.1: 89186964-89186964 on Build GRCh38
rs952284981 Chr.1: 89187333-89187333 on Build GRCh38
rs940664022 Chr.1: 89186989-89186989 on Build GRCh38
rs780686772 Chr.1: 89187129-89187129 on Build GRCh38
rs367579847 Chr.1: 89186948-89186948 on Build GRCh38
rs761869608 Chr.1: 89187063-89187063 on Build GRCh38
rs764266996 Chr.1: 89187058-89187058 on Build GRCh38
rs143862427 Chr.1: 89187274-89187274 on Build GRCh38
rs549235095 Chr.1: 89187172-89187172 on Build GRCh38
rs985365049 Chr.1: 89187353-89187353 on Build GRCh38
rs769336075 Chr.1: 89186983-89186983 on Build GRCh38
rs962452096 Chr.1: 89187374-89187374 on Build GRCh38

View Full Product Details