Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89189920-89190466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP4
Entrez Gene ID
115361
Gene Name
guanylate binding protein 4
Gene Aliases
Mpa2
UniGene
Hs.409925
Gene Chromosome Location
Chr.1: 89181148-89198950 on Build GRCh38
Transcripts
NM_052941.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6669800 c.1107C>T p.D369D Chr.1: 89190128-89190128 on Build GRCh38
3706082 c.1089G>T p.T363T Chr.1: 89190146-89190146 on Build GRCh38
6669801 c.1029G>T p.Q343H Chr.1: 89190206-89190206 on Build GRCh38
2153090 c.1089G>A p.T363T Chr.1: 89190146-89190146 on Build GRCh38
6127921 c.986C>T p.A329V Chr.1: 89190249-89190249 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757568251 Chr.1: 89190185-89190185 on Build GRCh38
rs868770550 Chr.1: 89190260-89190260 on Build GRCh38
rs755443191 Chr.1: 89190330-89190330 on Build GRCh38
rs111594101 Chr.1: 89190278-89190278 on Build GRCh38
rs75815237 Chr.1: 89189998-89189998 on Build GRCh38
rs955447052 Chr.1: 89190030-89190030 on Build GRCh38
rs541559940 Chr.1: 89190000-89190000 on Build GRCh38
rs570368419 Chr.1: 89190193-89190193 on Build GRCh38
rs542414272 Chr.1: 89190363-89190363 on Build GRCh38
rs918619706 Chr.1: 89190237-89190237 on Build GRCh38
rs769828427 Chr.1: 89190278-89190278 on Build GRCh38
rs766196513 Chr.1: 89190243-89190243 on Build GRCh38
rs751784119 Chr.1: 89190183-89190183 on Build GRCh38
rs41302774 Chr.1: 89190112-89190112 on Build GRCh38
rs775921871 Chr.1: 89190149-89190149 on Build GRCh38
rs778789961 Chr.1: 89190120-89190120 on Build GRCh38
rs986038242 Chr.1: 89190216-89190216 on Build GRCh38
rs948392838 Chr.1: 89190388-89190388 on Build GRCh38
rs558982077 Chr.1: 89190197-89190197 on Build GRCh38
rs766400073 Chr.1: 89190164-89190164 on Build GRCh38
rs773447623 Chr.1: 89190052-89190052 on Build GRCh38
rs557520852 Chr.1: 89190242-89190243 on Build GRCh38
rs560868087 Chr.1: 89190089-89190089 on Build GRCh38
rs41296177 Chr.1: 89190037-89190037 on Build GRCh38
rs200314067 Chr.1: 89190145-89190145 on Build GRCh38
rs763863492 Chr.1: 89190099-89190099 on Build GRCh38
rs41305862 Chr.1: 89190104-89190104 on Build GRCh38
rs752254198 Chr.1: 89189999-89189999 on Build GRCh38
rs757537588 Chr.1: 89190284-89190284 on Build GRCh38
rs574103174 Chr.1: 89190196-89190196 on Build GRCh38
rs1831240 Chr.1: 89190100-89190100 on Build GRCh38
rs767327035 Chr.1: 89190105-89190105 on Build GRCh38
rs747439654 Chr.1: 89190062-89190062 on Build GRCh38
rs757173083 Chr.1: 89190015-89190015 on Build GRCh38
rs778388918 Chr.1: 89190338-89190338 on Build GRCh38
rs753573560 Chr.1: 89190226-89190226 on Build GRCh38
rs768162538 Chr.1: 89190224-89190224 on Build GRCh38
rs764604789 Chr.1: 89190081-89190081 on Build GRCh38
rs748258713 Chr.1: 89190038-89190038 on Build GRCh38
rs761473019 Chr.1: 89190155-89190155 on Build GRCh38
rs781155764 Chr.1: 89190026-89190026 on Build GRCh38
rs756055755 Chr.1: 89190119-89190119 on Build GRCh38
rs749975942 Chr.1: 89190009-89190010 on Build GRCh38
rs369474687 Chr.1: 89190156-89190156 on Build GRCh38
rs763766416 Chr.1: 89190219-89190219 on Build GRCh38
rs770714474 Chr.1: 89190360-89190360 on Build GRCh38
rs760568418 Chr.1: 89190238-89190238 on Build GRCh38
rs560707763 Chr.1: 89190373-89190373 on Build GRCh38
rs145816730 Chr.1: 89190228-89190228 on Build GRCh38
rs374274451 Chr.1: 89190272-89190272 on Build GRCh38
rs12038448 Chr.1: 89190319-89190319 on Build GRCh38
rs750221101 Chr.1: 89190158-89190158 on Build GRCh38
rs777318349 Chr.1: 89190073-89190073 on Build GRCh38
rs781301967 Chr.1: 89190069-89190069 on Build GRCh38
rs760151993 Chr.1: 89190080-89190080 on Build GRCh38
rs778109948 Chr.1: 89190127-89190127 on Build GRCh38
rs369843962 Chr.1: 89190014-89190014 on Build GRCh38
rs974081550 Chr.1: 89190230-89190230 on Build GRCh38
rs747032080 Chr.1: 89190061-89190062 on Build GRCh38
rs371314987 Chr.1: 89190293-89190293 on Build GRCh38
rs749336341 Chr.1: 89190035-89190035 on Build GRCh38
rs1045473554 Chr.1: 89190392-89190392 on Build GRCh38
rs374179728 Chr.1: 89190167-89190167 on Build GRCh38
rs934422941 Chr.1: 89190417-89190417 on Build GRCh38
rs116011862 Chr.1: 89190123-89190123 on Build GRCh38
rs759454847 Chr.1: 89190364-89190364 on Build GRCh38
rs200329667 Chr.1: 89190232-89190232 on Build GRCh38
rs948466953 Chr.1: 89190101-89190101 on Build GRCh38
rs374581949 Chr.1: 89190051-89190051 on Build GRCh38
rs747716386 Chr.1: 89190344-89190344 on Build GRCh38
rs771776242 Chr.1: 89190229-89190229 on Build GRCh38
rs906950530 Chr.1: 89190404-89190405 on Build GRCh38
rs749330638 Chr.1: 89190028-89190028 on Build GRCh38
rs928540528 Chr.1: 89190281-89190281 on Build GRCh38
rs771383080 Chr.1: 89190132-89190132 on Build GRCh38
rs565623766 Chr.1: 89190150-89190150 on Build GRCh38
rs758549125 Chr.1: 89190169-89190169 on Build GRCh38
rs964814647 Chr.1: 89189962-89189962 on Build GRCh38
rs377276928 Chr.1: 89190059-89190059 on Build GRCh38
rs144326439 Chr.1: 89190188-89190188 on Build GRCh38
rs1038021556 Chr.1: 89190088-89190088 on Build GRCh38
rs746844590 Chr.1: 89189986-89189986 on Build GRCh38
rs371383412 Chr.1: 89189988-89189988 on Build GRCh38
rs1410871 Chr.1: 89190311-89190311 on Build GRCh38
rs138472408 Chr.1: 89190147-89190147 on Build GRCh38
rs201574307 Chr.1: 89190299-89190299 on Build GRCh38
rs777746594 Chr.1: 89190172-89190172 on Build GRCh38
rs759418502 Chr.1: 89190256-89190256 on Build GRCh38
rs747351685 Chr.1: 89190128-89190128 on Build GRCh38
rs770397320 Chr.1: 89190141-89190141 on Build GRCh38
rs779697015 Chr.1: 89190040-89190041 on Build GRCh38
rs150829936 Chr.1: 89190276-89190276 on Build GRCh38
rs771692300 Chr.1: 89190347-89190347 on Build GRCh38
rs746672685 Chr.1: 89190354-89190354 on Build GRCh38
rs200509294 Chr.1: 89190349-89190349 on Build GRCh38
rs749744566 Chr.1: 89190326-89190326 on Build GRCh38
rs536853908 Chr.1: 89190153-89190153 on Build GRCh38
rs780647917 Chr.1: 89190215-89190215 on Build GRCh38
rs374533033 Chr.1: 89190034-89190034 on Build GRCh38
rs775081314 Chr.1: 89190083-89190083 on Build GRCh38
rs776592075 Chr.1: 89190246-89190246 on Build GRCh38
rs780304326 Chr.1: 89190317-89190317 on Build GRCh38
rs559571426 Chr.1: 89190001-89190001 on Build GRCh38
rs760528342 Chr.1: 89190161-89190161 on Build GRCh38
rs201246795 Chr.1: 89190036-89190036 on Build GRCh38
rs10922590 Chr.1: 89190440-89190440 on Build GRCh38
rs754875448 Chr.1: 89190062-89190063 on Build GRCh38
rs781708260 Chr.1: 89190134-89190134 on Build GRCh38

View Full Product Details