Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89192818-89193314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP4
Entrez Gene ID
115361
Gene Name
guanylate binding protein 4
Gene Aliases
Mpa2
UniGene
Hs.409925
Gene Chromosome Location
Chr.1: 89181148-89198950 on Build GRCh38
Transcripts
NM_052941.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
394724 c.629C>A p.T210K Chr.1: 89192945-89192945 on Build GRCh38
4829015 c.484G>C p.E162Q Chr.1: 89193090-89193090 on Build GRCh38
5601541 c.561C>T p.F187F Chr.1: 89193013-89193013 on Build GRCh38
5954181 c.473+1G>T p.? Chr.1: 89193302-89193302 on Build GRCh38
6895348 c.667C>G p.P223A Chr.1: 89192907-89192907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758362147 Chr.1: 89192917-89192917 on Build GRCh38
rs749544876 Chr.1: 89192965-89192965 on Build GRCh38
rs753652717 Chr.1: 89192887-89192887 on Build GRCh38
rs766020046 Chr.1: 89192994-89192994 on Build GRCh38
rs745920473 Chr.1: 89193019-89193019 on Build GRCh38
rs769301570 Chr.1: 89193287-89193287 on Build GRCh38
rs544537323 Chr.1: 89193177-89193177 on Build GRCh38
rs773813164 Chr.1: 89193068-89193068 on Build GRCh38
rs1056261263 Chr.1: 89192869-89192869 on Build GRCh38
rs543635650 Chr.1: 89192899-89192899 on Build GRCh38
rs563179550 Chr.1: 89193229-89193229 on Build GRCh38
rs763360832 Chr.1: 89192853-89192853 on Build GRCh38
rs760300985 Chr.1: 89192993-89192993 on Build GRCh38
rs186821951 Chr.1: 89193028-89193028 on Build GRCh38
rs762222254 Chr.1: 89192989-89192989 on Build GRCh38
rs771384856 Chr.1: 89193101-89193102 on Build GRCh38
rs757189387 Chr.1: 89193126-89193126 on Build GRCh38
rs374871857 Chr.1: 89192901-89192901 on Build GRCh38
rs139853417 Chr.1: 89193041-89193041 on Build GRCh38
rs775733093 Chr.1: 89192961-89192961 on Build GRCh38
rs752608414 Chr.1: 89193005-89193005 on Build GRCh38
rs778298335 Chr.1: 89193256-89193256 on Build GRCh38
rs780099646 Chr.1: 89193127-89193127 on Build GRCh38
rs747978614 Chr.1: 89192916-89192916 on Build GRCh38
rs561908001 Chr.1: 89193192-89193192 on Build GRCh38
rs372214198 Chr.1: 89192882-89192882 on Build GRCh38
rs974785870 Chr.1: 89193051-89193051 on Build GRCh38
rs764875892 Chr.1: 89193004-89193004 on Build GRCh38
rs754006692 Chr.1: 89193128-89193128 on Build GRCh38
rs768985990 Chr.1: 89193035-89193035 on Build GRCh38
rs779132251 Chr.1: 89193038-89193038 on Build GRCh38
rs775171223 Chr.1: 89193290-89193290 on Build GRCh38
rs985910768 Chr.1: 89193243-89193243 on Build GRCh38
rs535788014 Chr.1: 89193000-89193000 on Build GRCh38
rs544553218 Chr.1: 89193198-89193198 on Build GRCh38
rs759400322 Chr.1: 89192988-89192988 on Build GRCh38
rs944652452 Chr.1: 89193025-89193025 on Build GRCh38
rs369929194 Chr.1: 89193097-89193097 on Build GRCh38
rs762849400 Chr.1: 89193102-89193102 on Build GRCh38
rs754689977 Chr.1: 89192900-89192900 on Build GRCh38
rs371766927 Chr.1: 89193153-89193153 on Build GRCh38
rs763911627 Chr.1: 89193105-89193105 on Build GRCh38
rs140595004 Chr.1: 89193250-89193250 on Build GRCh38
rs772517963 Chr.1: 89193252-89193252 on Build GRCh38
rs371947273 Chr.1: 89192862-89192862 on Build GRCh38
rs776209967 Chr.1: 89193085-89193085 on Build GRCh38
rs529438588 Chr.1: 89193194-89193194 on Build GRCh38
rs140648377 Chr.1: 89192935-89192935 on Build GRCh38
rs917692700 Chr.1: 89193210-89193210 on Build GRCh38
rs779065326 Chr.1: 89193138-89193138 on Build GRCh38
rs747453596 Chr.1: 89193269-89193269 on Build GRCh38
rs1009851498 Chr.1: 89193163-89193163 on Build GRCh38
rs374804401 Chr.1: 89193275-89193275 on Build GRCh38
rs759139414 Chr.1: 89193282-89193282 on Build GRCh38
rs367618325 Chr.1: 89192987-89192987 on Build GRCh38
rs759107357 Chr.1: 89193093-89193093 on Build GRCh38
rs752416116 Chr.1: 89193101-89193101 on Build GRCh38
rs369250923 Chr.1: 89193270-89193270 on Build GRCh38
rs770510201 Chr.1: 89193084-89193084 on Build GRCh38
rs372380560 Chr.1: 89193129-89193129 on Build GRCh38
rs750019411 Chr.1: 89192878-89192878 on Build GRCh38
rs766868354 Chr.1: 89192874-89192874 on Build GRCh38
rs149907641 Chr.1: 89193006-89193006 on Build GRCh38
rs762391173 Chr.1: 89192866-89192866 on Build GRCh38
rs368080989 Chr.1: 89193079-89193079 on Build GRCh38
rs201239901 Chr.1: 89192966-89192966 on Build GRCh38
rs769983270 Chr.1: 89192953-89192953 on Build GRCh38
rs766180922 Chr.1: 89192885-89192885 on Build GRCh38
rs371719087 Chr.1: 89193003-89193003 on Build GRCh38
rs146807380 Chr.1: 89193147-89193147 on Build GRCh38
rs762604969 Chr.1: 89193292-89193292 on Build GRCh38
rs906127699 Chr.1: 89192968-89192968 on Build GRCh38
rs200066635 Chr.1: 89193288-89193288 on Build GRCh38
rs775994157 Chr.1: 89193276-89193276 on Build GRCh38
rs968456484 Chr.1: 89193202-89193202 on Build GRCh38
rs777739942 Chr.1: 89193012-89193012 on Build GRCh38
rs377180415 Chr.1: 89193061-89193061 on Build GRCh38
rs780118405 Chr.1: 89193027-89193027 on Build GRCh38
rs1056682676 Chr.1: 89193181-89193181 on Build GRCh38
rs746009436 Chr.1: 89192940-89192940 on Build GRCh38
rs143885653 Chr.1: 89192907-89192907 on Build GRCh38
rs774669086 Chr.1: 89192865-89192865 on Build GRCh38
rs748348712 Chr.1: 89193148-89193148 on Build GRCh38
rs954167687 Chr.1: 89193238-89193239 on Build GRCh38
rs150777601 Chr.1: 89192991-89192991 on Build GRCh38
rs1028818688 Chr.1: 89193226-89193226 on Build GRCh38
rs756135431 Chr.1: 89193020-89193020 on Build GRCh38

View Full Product Details