Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 91511503-91511938 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC7
Entrez Gene ID
8317
Gene Name
cell division cycle 7
Gene Aliases
CDC7L1, HsCDC7, Hsk1, huCDC7
UniGene
Hs.533573
Gene Chromosome Location
Chr.1: 91500814-91525764 on Build GRCh38
Transcripts
NM_001134419.1 NM_001134420.1 NM_003503.3 XM_005271241.2 XM_005271244.2 XM_017002425.1 XM_017002426.1 XM_017002427.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6449161 c.337G>T p.G113W Chr.1: 91511598-91511598 on Build GRCh38
5773236 c.537G>A p.K179K Chr.1: 91511888-91511888 on Build GRCh38
1561035 c.491C>T p.A164V Chr.1: 91511842-91511842 on Build GRCh38
6364406 c.537G>C p.K179N Chr.1: 91511888-91511888 on Build GRCh38
1344866 c.546delT p.L184fs*6 Chr.1: 91511897-91511897 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755854755 Chr.1: 91511876-91511876 on Build GRCh38
rs141839416 Chr.1: 91511684-91511684 on Build GRCh38
rs369009955 Chr.1: 91511751-91511751 on Build GRCh38
rs764839778 Chr.1: 91511734-91511734 on Build GRCh38
rs1002965604 Chr.1: 91511771-91511771 on Build GRCh38
rs372624837 Chr.1: 91511753-91511753 on Build GRCh38
rs192393377 Chr.1: 91511737-91511737 on Build GRCh38
rs752362072 Chr.1: 91511796-91511800 on Build GRCh38
rs545630891 Chr.1: 91511654-91511654 on Build GRCh38
rs748433817 Chr.1: 91511664-91511664 on Build GRCh38
rs539320135 Chr.1: 91511913-91511913 on Build GRCh38
rs570397909 Chr.1: 91511644-91511644 on Build GRCh38
rs13447503 Chr.1: 91511837-91511837 on Build GRCh38
rs549548896 Chr.1: 91511685-91511685 on Build GRCh38
rs763590123 Chr.1: 91511740-91511740 on Build GRCh38
rs923049737 Chr.1: 91511587-91511587 on Build GRCh38
rs974367033 Chr.1: 91511580-91511580 on Build GRCh38
rs758464467 Chr.1: 91511907-91511907 on Build GRCh38
rs916468328 Chr.1: 91511723-91511723 on Build GRCh38
rs369955837 Chr.1: 91511593-91511593 on Build GRCh38
rs747270363 Chr.1: 91511911-91511911 on Build GRCh38
rs556566766 Chr.1: 91511538-91511538 on Build GRCh38
rs745789199 Chr.1: 91511639-91511639 on Build GRCh38
rs746791289 Chr.1: 91511547-91511547 on Build GRCh38
rs766161953 Chr.1: 91511898-91511898 on Build GRCh38
rs779072497 Chr.1: 91511791-91511791 on Build GRCh38
rs777426874 Chr.1: 91511638-91511650 on Build GRCh38
rs770145582 Chr.1: 91511832-91511832 on Build GRCh38
rs772136364 Chr.1: 91511666-91511666 on Build GRCh38
rs751307469 Chr.1: 91511862-91511862 on Build GRCh38
rs941854527 Chr.1: 91511673-91511673 on Build GRCh38
rs374676600 Chr.1: 91511747-91511747 on Build GRCh38
rs750195064 Chr.1: 91511889-91511889 on Build GRCh38
rs763649740 Chr.1: 91511851-91511851 on Build GRCh38
rs369750539 Chr.1: 91511839-91511839 on Build GRCh38
rs60472082 Chr.1: 91511861-91511861 on Build GRCh38
rs767452060 Chr.1: 91511885-91511885 on Build GRCh38
rs768799588 Chr.1: 91511585-91511585 on Build GRCh38
rs1046494803 Chr.1: 91511607-91511607 on Build GRCh38
rs375886193 Chr.1: 91511814-91511814 on Build GRCh38
rs573122638 Chr.1: 91511805-91511805 on Build GRCh38
rs777805381 Chr.1: 91511910-91511910 on Build GRCh38
rs773638369 Chr.1: 91511680-91511680 on Build GRCh38
rs776945490 Chr.1: 91511819-91511819 on Build GRCh38
rs772989323 Chr.1: 91511750-91511750 on Build GRCh38
rs983789539 Chr.1: 91511807-91511807 on Build GRCh38
rs183169466 Chr.1: 91511738-91511738 on Build GRCh38
rs772341414 Chr.1: 91511797-91511797 on Build GRCh38
rs751256308 Chr.1: 91511743-91511743 on Build GRCh38
rs951126126 Chr.1: 91511776-91511776 on Build GRCh38
rs753349287 Chr.1: 91511553-91511553 on Build GRCh38
rs375702482 Chr.1: 91511794-91511794 on Build GRCh38
rs747885913 Chr.1: 91511683-91511683 on Build GRCh38
rs754546437 Chr.1: 91511562-91511562 on Build GRCh38
rs373272987 Chr.1: 91511868-91511868 on Build GRCh38
rs760011700 Chr.1: 91511731-91511731 on Build GRCh38
rs756090077 Chr.1: 91511891-91511891 on Build GRCh38
rs148946131 Chr.1: 91511630-91511630 on Build GRCh38
rs770345842 Chr.1: 91511815-91511815 on Build GRCh38
rs781348040 Chr.1: 91511632-91511632 on Build GRCh38
rs373489388 Chr.1: 91511694-91511694 on Build GRCh38
rs567242685 Chr.1: 91511540-91511540 on Build GRCh38
rs907173187 Chr.1: 91511754-91511754 on Build GRCh38
rs752439011 Chr.1: 91511581-91511581 on Build GRCh38
rs768907349 Chr.1: 91511641-91511641 on Build GRCh38
rs753807413 Chr.1: 91511899-91511899 on Build GRCh38
rs762439340 Chr.1: 91511846-91511846 on Build GRCh38
rs200604322 Chr.1: 91511789-91511789 on Build GRCh38
rs750427977 Chr.1: 91511772-91511772 on Build GRCh38
rs753858262 Chr.1: 91511755-91511759 on Build GRCh38
rs201651444 Chr.1: 91511609-91511609 on Build GRCh38
rs1053781837 Chr.1: 91511634-91511634 on Build GRCh38
rs764637291 Chr.1: 91511579-91511579 on Build GRCh38
rs766811931 Chr.1: 91511547-91511547 on Build GRCh38

View Full Product Details