Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 91513810-91514327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC7
Entrez Gene ID
8317
Gene Name
cell division cycle 7
Gene Aliases
CDC7L1, HsCDC7, Hsk1, huCDC7
UniGene
Hs.533573
Gene Chromosome Location
Chr.1: 91500814-91525764 on Build GRCh38
Transcripts
NM_001134419.1 NM_001134420.1 NM_003503.3 XM_005271241.2 XM_005271244.2 XM_017002425.1 XM_017002426.1 XM_017002427.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5735529 c.823-1G>T p.? Chr.1: 91513947-91513947 on Build GRCh38
6127874 c.903G>T p.E301D Chr.1: 91514028-91514028 on Build GRCh38
6855362 c.882A>G p.I294M Chr.1: 91514007-91514007 on Build GRCh38
1344867 c.869G>T p.R290I Chr.1: 91513994-91513994 on Build GRCh38
6540623 c.827G>T p.G276V Chr.1: 91513952-91513952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs981313771 Chr.1: 91513912-91513912 on Build GRCh38
rs758985281 Chr.1: 91514059-91514059 on Build GRCh38
rs559506095 Chr.1: 91513957-91513957 on Build GRCh38
rs865935428 Chr.1: 91513951-91513951 on Build GRCh38
rs1027831683 Chr.1: 91514207-91514207 on Build GRCh38
rs147452385 Chr.1: 91514225-91514225 on Build GRCh38
rs377195713 Chr.1: 91514082-91514082 on Build GRCh38
rs1016110161 Chr.1: 91514236-91514236 on Build GRCh38
rs777168500 Chr.1: 91513922-91513922 on Build GRCh38
rs775228172 Chr.1: 91513976-91513976 on Build GRCh38
rs773556661 Chr.1: 91514156-91514156 on Build GRCh38
rs768583317 Chr.1: 91513988-91513988 on Build GRCh38
rs1008886372 Chr.1: 91514107-91514107 on Build GRCh38
rs764592045 Chr.1: 91514063-91514063 on Build GRCh38
rs770490952 Chr.1: 91513941-91513941 on Build GRCh38
rs746465373 Chr.1: 91513929-91513929 on Build GRCh38
rs773701370 Chr.1: 91513926-91513927 on Build GRCh38
rs759798939 Chr.1: 91514038-91514038 on Build GRCh38
rs765725228 Chr.1: 91514052-91514052 on Build GRCh38
rs546865871 Chr.1: 91514130-91514131 on Build GRCh38
rs756842846 Chr.1: 91514091-91514091 on Build GRCh38
rs758930308 Chr.1: 91513920-91513920 on Build GRCh38
rs1038663632 Chr.1: 91514141-91514141 on Build GRCh38
rs780959002 Chr.1: 91513944-91513944 on Build GRCh38
rs969926206 Chr.1: 91513879-91513879 on Build GRCh38
rs189644752 Chr.1: 91514167-91514167 on Build GRCh38
rs773171042 Chr.1: 91514020-91514020 on Build GRCh38
rs767088019 Chr.1: 91514304-91514304 on Build GRCh38
rs1043578920 Chr.1: 91513911-91513911 on Build GRCh38
rs374379764 Chr.1: 91513983-91513983 on Build GRCh38
rs370367545 Chr.1: 91514028-91514028 on Build GRCh38
rs777040391 Chr.1: 91514037-91514037 on Build GRCh38
rs753296151 Chr.1: 91513903-91513903 on Build GRCh38
rs185516031 Chr.1: 91514125-91514125 on Build GRCh38
rs575596960 Chr.1: 91513955-91513955 on Build GRCh38
rs368391670 Chr.1: 91514024-91514024 on Build GRCh38
rs889149891 Chr.1: 91514080-91514080 on Build GRCh38
rs914303765 Chr.1: 91514066-91514066 on Build GRCh38
rs753059019 Chr.1: 91514057-91514057 on Build GRCh38
rs952153979 Chr.1: 91514209-91514209 on Build GRCh38
rs765513529 Chr.1: 91514142-91514142 on Build GRCh38
rs186600897 Chr.1: 91514217-91514217 on Build GRCh38
rs180687089 Chr.1: 91514205-91514205 on Build GRCh38
rs371099029 Chr.1: 91513972-91513972 on Build GRCh38
rs769620136 Chr.1: 91513975-91513975 on Build GRCh38
rs763589588 Chr.1: 91513958-91513958 on Build GRCh38
rs554253311 Chr.1: 91513896-91513896 on Build GRCh38
rs934427983 Chr.1: 91513939-91513939 on Build GRCh38

View Full Product Details