Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92244093-92244603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf146
Entrez Gene ID
388649
Gene Name
chromosome 1 open reading frame 146
Gene Aliases
-
UniGene
Hs.558823
Gene Chromosome Location
Chr.1: 92217920-92245812 on Build GRCh38
Transcripts
NM_001012425.1 XM_011541447.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
274242 c.211G>T p.E71* Chr.1: 92244267-92244267 on Build GRCh38
2260008 c.305A>G p.K102R Chr.1: 92244361-92244361 on Build GRCh38
4667306 c.260_261insT p.L89fs*11 Chr.1: 92244316-92244317 on Build GRCh38
1688041 c.304A>G p.K102E Chr.1: 92244360-92244360 on Build GRCh38
6460780 c.211G>A p.E71K Chr.1: 92244267-92244267 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021448307 Chr.1: 92244562-92244562 on Build GRCh38
rs745428835 Chr.1: 92244288-92244288 on Build GRCh38
rs201472965 Chr.1: 92244384-92244384 on Build GRCh38
rs553649684 Chr.1: 92244542-92244542 on Build GRCh38
rs748677441 Chr.1: 92244249-92244249 on Build GRCh38
rs200889079 Chr.1: 92244190-92244190 on Build GRCh38
rs140165531 Chr.1: 92244357-92244357 on Build GRCh38
rs143946132 Chr.1: 92244244-92244244 on Build GRCh38
rs1032207891 Chr.1: 92244316-92244316 on Build GRCh38
rs764198776 Chr.1: 92244375-92244375 on Build GRCh38
rs943764375 Chr.1: 92244409-92244409 on Build GRCh38
rs533666303 Chr.1: 92244559-92244559 on Build GRCh38
rs112867262 Chr.1: 92244456-92244456 on Build GRCh38
rs746586562 Chr.1: 92244295-92244295 on Build GRCh38
rs925434887 Chr.1: 92244455-92244455 on Build GRCh38
rs776356714 Chr.1: 92244313-92244313 on Build GRCh38
rs760154979 Chr.1: 92244167-92244167 on Build GRCh38
rs190954769 Chr.1: 92244556-92244556 on Build GRCh38
rs953736162 Chr.1: 92244333-92244333 on Build GRCh38
rs10676893 Chr.1: 92244132-92244133 on Build GRCh38
rs568323805 Chr.1: 92244543-92244543 on Build GRCh38
rs749050927 Chr.1: 92244393-92244394 on Build GRCh38
rs371721685 Chr.1: 92244175-92244175 on Build GRCh38
rs564970621 Chr.1: 92244250-92244250 on Build GRCh38
rs760766972 Chr.1: 92244311-92244314 on Build GRCh38
rs369724113 Chr.1: 92244433-92244433 on Build GRCh38
rs775722574 Chr.1: 92244293-92244294 on Build GRCh38
rs146856959 Chr.1: 92244380-92244380 on Build GRCh38
rs368712519 Chr.1: 92244338-92244338 on Build GRCh38
rs771623972 Chr.1: 92244301-92244301 on Build GRCh38
rs966941726 Chr.1: 92244276-92244276 on Build GRCh38
rs142040705 Chr.1: 92244579-92244579 on Build GRCh38
rs780384280 Chr.1: 92244232-92244232 on Build GRCh38
rs376479300 Chr.1: 92244399-92244399 on Build GRCh38
rs993804815 Chr.1: 92244124-92244124 on Build GRCh38
rs765519238 Chr.1: 92244426-92244427 on Build GRCh38
rs772333623 Chr.1: 92244207-92244207 on Build GRCh38
rs773612324 Chr.1: 92244545-92244545 on Build GRCh38
rs186098177 Chr.1: 92244389-92244389 on Build GRCh38
rs756262609 Chr.1: 92244229-92244229 on Build GRCh38
rs767723876 Chr.1: 92244216-92244216 on Build GRCh38
rs749414740 Chr.1: 92244233-92244233 on Build GRCh38
rs149037808 Chr.1: 92244531-92244531 on Build GRCh38
rs1045400612 Chr.1: 92244525-92244525 on Build GRCh38
rs369209440 Chr.1: 92244218-92244218 on Build GRCh38
rs35478871 Chr.1: 92244133-92244134 on Build GRCh38
rs1045705544 Chr.1: 92244122-92244122 on Build GRCh38
rs762282552 Chr.1: 92244419-92244423 on Build GRCh38
rs756245235 Chr.1: 92244447-92244448 on Build GRCh38
rs762814187 Chr.1: 92244374-92244374 on Build GRCh38
rs949721348 Chr.1: 92244520-92244520 on Build GRCh38
rs200386451 Chr.1: 92244198-92244198 on Build GRCh38
rs755107636 Chr.1: 92244437-92244437 on Build GRCh38
rs929453113 Chr.1: 92244478-92244478 on Build GRCh38
rs912957804 Chr.1: 92244509-92244509 on Build GRCh38
rs766060337 Chr.1: 92244170-92244170 on Build GRCh38
rs779107058 Chr.1: 92244245-92244245 on Build GRCh38
rs61732740 Chr.1: 92244267-92244267 on Build GRCh38
rs769658360 Chr.1: 92244353-92244353 on Build GRCh38
rs764438167 Chr.1: 92244316-92244317 on Build GRCh38
rs200991118 Chr.1: 92244129-92244129 on Build GRCh38
rs745786290 Chr.1: 92244182-92244183 on Build GRCh38
rs576505202 Chr.1: 92244173-92244173 on Build GRCh38
rs201931838 Chr.1: 92244177-92244177 on Build GRCh38
rs17131614 Chr.1: 92244482-92244482 on Build GRCh38
rs550217292 Chr.1: 92244343-92244343 on Build GRCh38
rs767533577 Chr.1: 92244459-92244463 on Build GRCh38

View Full Product Details