Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92833253-92833775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM69A
Entrez Gene ID
388650
Gene Name
family with sequence similarity 69 member A
Gene Aliases
-
UniGene
Hs.180946
Gene Chromosome Location
Chr.1: 92832729-92961522 on Build GRCh38
Transcripts
NM_001252273.1 NM_000969.3

Gene Symbol
RPL5
Entrez Gene ID
6125
Gene Name
ribosomal protein L5
Gene Aliases
DBA6, L5, MSTP030, PPP1R135
UniGene
Hs.532359
Gene Chromosome Location
Chr.1: 92832037-92841924 on Build GRCh38
Transcripts
NM_001252273.1 NM_000969.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3736985 c.188A>G p.Q63R Chr.1: 92833659-92833659 on Build GRCh38
6848210 c.112_113insT p.Q39fs*3 Chr.1: 92833583-92833584 on Build GRCh38
3665020 c.173G>C p.R58T Chr.1: 92833644-92833644 on Build GRCh38
5573979 c.171_172insA p.R58fs*55 Chr.1: 92833642-92833643 on Build GRCh38
5501414 c.74-7C>T p.? Chr.1: 92833538-92833538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767035217 Chr.1: 92833605-92833605 on Build GRCh38
rs757146931 Chr.1: 92833526-92833526 on Build GRCh38
rs372954310 Chr.1: 92833659-92833659 on Build GRCh38
rs746944177 Chr.1: 92833385-92833385 on Build GRCh38
rs777744297 Chr.1: 92833383-92833383 on Build GRCh38
rs142156224 Chr.1: 92833460-92833460 on Build GRCh38
rs114860423 Chr.1: 92833319-92833319 on Build GRCh38
rs569054211 Chr.1: 92833506-92833506 on Build GRCh38
rs571720015 Chr.1: 92833364-92833364 on Build GRCh38
rs777880100 Chr.1: 92833486-92833486 on Build GRCh38
rs14685 Chr.1: 92833448-92833448 on Build GRCh38
rs186600450 Chr.1: 92833360-92833360 on Build GRCh38
rs756108510 Chr.1: 92833483-92833483 on Build GRCh38
rs754413727 Chr.1: 92833490-92833490 on Build GRCh38
rs769877057 Chr.1: 92833391-92833391 on Build GRCh38
rs376462650 Chr.1: 92833359-92833359 on Build GRCh38
rs369728038 Chr.1: 92833342-92833342 on Build GRCh38
rs762299279 Chr.1: 92833418-92833418 on Build GRCh38
rs121434405 Chr.1: 92833452-92833452 on Build GRCh38
rs748318627 Chr.1: 92833559-92833559 on Build GRCh38
rs746972158 Chr.1: 92833513-92833513 on Build GRCh38
rs769428365 Chr.1: 92833685-92833688 on Build GRCh38
rs778601996 Chr.1: 92833361-92833361 on Build GRCh38
rs779636137 Chr.1: 92833354-92833354 on Build GRCh38
rs372392388 Chr.1: 92833297-92833297 on Build GRCh38
rs764649729 Chr.1: 92833631-92833631 on Build GRCh38
rs762712969 Chr.1: 92833707-92833708 on Build GRCh38
rs755086283 Chr.1: 92833683-92833683 on Build GRCh38
rs772388093 Chr.1: 92833706-92833706 on Build GRCh38
rs771831213 Chr.1: 92833372-92833372 on Build GRCh38
rs768608404 Chr.1: 92833549-92833549 on Build GRCh38
rs933158035 Chr.1: 92833527-92833527 on Build GRCh38
rs749504844 Chr.1: 92833401-92833401 on Build GRCh38
rs774705183 Chr.1: 92833554-92833554 on Build GRCh38
rs768965548 Chr.1: 92833403-92833403 on Build GRCh38
rs772575122 Chr.1: 92833574-92833574 on Build GRCh38
rs764987679 Chr.1: 92833491-92833491 on Build GRCh38
rs757021956 Chr.1: 92833671-92833671 on Build GRCh38
rs373050134 Chr.1: 92833346-92833346 on Build GRCh38
rs11540838 Chr.1: 92833573-92833573 on Build GRCh38
rs750956864 Chr.1: 92833343-92833343 on Build GRCh38
rs943313650 Chr.1: 92833380-92833380 on Build GRCh38
rs752988812 Chr.1: 92833333-92833333 on Build GRCh38
rs773060381 Chr.1: 92833685-92833686 on Build GRCh38
rs774416689 Chr.1: 92833417-92833417 on Build GRCh38
rs772661926 Chr.1: 92833358-92833358 on Build GRCh38
rs780962420 Chr.1: 92833538-92833538 on Build GRCh38
rs912779029 Chr.1: 92833632-92833632 on Build GRCh38
rs947480137 Chr.1: 92833730-92833731 on Build GRCh38
rs147872195 Chr.1: 92833577-92833577 on Build GRCh38
rs763905952 Chr.1: 92833649-92833649 on Build GRCh38
rs565570301 Chr.1: 92833722-92833722 on Build GRCh38
rs758256418 Chr.1: 92833495-92833495 on Build GRCh38
rs974273887 Chr.1: 92833377-92833377 on Build GRCh38
rs778112670 Chr.1: 92833710-92833710 on Build GRCh38
rs781245464 Chr.1: 92833672-92833672 on Build GRCh38
rs1032177577 Chr.1: 92833294-92833294 on Build GRCh38
rs761298943 Chr.1: 92833590-92833590 on Build GRCh38
rs555012303 Chr.1: 92833307-92833308 on Build GRCh38
rs761202801 Chr.1: 92833462-92833462 on Build GRCh38
rs745785928 Chr.1: 92833676-92833676 on Build GRCh38
rs1013212765 Chr.1: 92833744-92833744 on Build GRCh38
rs748406812 Chr.1: 92833697-92833697 on Build GRCh38
rs1013654956 Chr.1: 92833293-92833293 on Build GRCh38
rs780657111 Chr.1: 92833376-92833376 on Build GRCh38
rs150375325 Chr.1: 92833612-92833612 on Build GRCh38
rs766006888 Chr.1: 92833475-92833475 on Build GRCh38
rs549727048 Chr.1: 92833276-92833276 on Build GRCh38
rs376226148 Chr.1: 92833400-92833400 on Build GRCh38
rs778887807 Chr.1: 92833748-92833748 on Build GRCh38
rs974629918 Chr.1: 92833470-92833470 on Build GRCh38
rs773855879 Chr.1: 92833454-92833454 on Build GRCh38
rs767579888 Chr.1: 92833318-92833318 on Build GRCh38
rs777486641 Chr.1: 92833510-92833510 on Build GRCh38
rs148673599 Chr.1: 92833433-92833433 on Build GRCh38
rs370327480 Chr.1: 92833492-92833492 on Build GRCh38
rs751394001 Chr.1: 92833665-92833665 on Build GRCh38
rs58263806 Chr.1: 92833636-92833636 on Build GRCh38
rs767994083 Chr.1: 92833435-92833435 on Build GRCh38
rs753968704 Chr.1: 92833627-92833627 on Build GRCh38
rs757996884 Chr.1: 92833637-92833637 on Build GRCh38
rs778804578 Chr.1: 92833686-92833686 on Build GRCh38
rs747703119 Chr.1: 92833531-92833531 on Build GRCh38
rs894793689 Chr.1: 92833734-92833734 on Build GRCh38

View Full Product Details