Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 93532802-93533346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNBP1L
Entrez Gene ID
54874
Gene Name
formin binding protein 1 like
Gene Aliases
C1orf39, TOCA1
UniGene
Hs.134060
Gene Chromosome Location
Chr.1: 93448067-93569671 on Build GRCh38
Transcripts
NM_001024948.2 NM_001164473.2 NM_017737.4 XM_011541625.2 XM_017001533.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
350138 c.778G>A p.E260K Chr.1: 93533060-93533060 on Build GRCh38
3806131 c.753G>C p.M251I Chr.1: 93533035-93533035 on Build GRCh38
3386396 c.772G>T p.V258F Chr.1: 93533054-93533054 on Build GRCh38
6665839 c.736A>G p.K246E Chr.1: 93533018-93533018 on Build GRCh38
7368584 c.744G>C p.L248F Chr.1: 93533026-93533026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550084998 Chr.1: 93532977-93532977 on Build GRCh38
rs749702432 Chr.1: 93533002-93533002 on Build GRCh38
rs774882822 Chr.1: 93532927-93532927 on Build GRCh38
rs377284497 Chr.1: 93532984-93532984 on Build GRCh38
rs765071378 Chr.1: 93533088-93533088 on Build GRCh38
rs200815089 Chr.1: 93533083-93533083 on Build GRCh38
rs769066231 Chr.1: 93532924-93532924 on Build GRCh38
rs766994624 Chr.1: 93532939-93532939 on Build GRCh38
rs185227108 Chr.1: 93532943-93532943 on Build GRCh38
rs34814765 Chr.1: 93533084-93533084 on Build GRCh38
rs746077375 Chr.1: 93532885-93532885 on Build GRCh38
rs189310017 Chr.1: 93533098-93533098 on Build GRCh38
rs904459550 Chr.1: 93533269-93533269 on Build GRCh38
rs778742407 Chr.1: 93533302-93533302 on Build GRCh38
rs760367327 Chr.1: 93533058-93533058 on Build GRCh38
rs374859446 Chr.1: 93533309-93533309 on Build GRCh38
rs891418574 Chr.1: 93533244-93533244 on Build GRCh38
rs938465059 Chr.1: 93533230-93533230 on Build GRCh38
rs556789113 Chr.1: 93533298-93533298 on Build GRCh38
rs934163580 Chr.1: 93533310-93533310 on Build GRCh38
rs182219744 Chr.1: 93533295-93533295 on Build GRCh38
rs1051234132 Chr.1: 93533316-93533316 on Build GRCh38
rs1034249064 Chr.1: 93533256-93533256 on Build GRCh38
rs948224636 Chr.1: 93532942-93532942 on Build GRCh38
rs1010026415 Chr.1: 93533206-93533206 on Build GRCh38
rs771547672 Chr.1: 93533039-93533039 on Build GRCh38
rs370837605 Chr.1: 93532994-93532994 on Build GRCh38
rs141240755 Chr.1: 93532884-93532884 on Build GRCh38
rs909847078 Chr.1: 93533271-93533271 on Build GRCh38
rs761233097 Chr.1: 93532930-93532930 on Build GRCh38
rs763990566 Chr.1: 93532873-93532876 on Build GRCh38
rs927199666 Chr.1: 93532914-93532914 on Build GRCh38
rs957112860 Chr.1: 93532866-93532866 on Build GRCh38
rs920851196 Chr.1: 93533161-93533161 on Build GRCh38
rs938038511 Chr.1: 93532933-93532933 on Build GRCh38
rs532285266 Chr.1: 93533032-93533032 on Build GRCh38
rs981223023 Chr.1: 93533232-93533232 on Build GRCh38
rs753616935 Chr.1: 93532990-93532991 on Build GRCh38
rs954981830 Chr.1: 93533091-93533091 on Build GRCh38
rs531482489 Chr.1: 93532963-93532963 on Build GRCh38
rs752646786 Chr.1: 93533089-93533089 on Build GRCh38
rs371195306 Chr.1: 93532908-93532908 on Build GRCh38
rs143226457 Chr.1: 93532949-93532949 on Build GRCh38
rs1037611001 Chr.1: 93533082-93533082 on Build GRCh38
rs756193665 Chr.1: 93532940-93532940 on Build GRCh38
rs766200009 Chr.1: 93532950-93532950 on Build GRCh38
rs988000337 Chr.1: 93533321-93533321 on Build GRCh38
rs767756517 Chr.1: 93533115-93533115 on Build GRCh38
rs1056799658 Chr.1: 93533231-93533231 on Build GRCh38
rs914445033 Chr.1: 93532862-93532862 on Build GRCh38
rs10541830 Chr.1: 93533263-93533263 on Build GRCh38
rs397802228 Chr.1: 93533085-93533085 on Build GRCh38
rs759705056 Chr.1: 93532972-93532972 on Build GRCh38
rs768998180 Chr.1: 93533006-93533006 on Build GRCh38
rs879006169 Chr.1: 93532840-93532840 on Build GRCh38
rs200518030 Chr.1: 93533060-93533060 on Build GRCh38
rs759267130 Chr.1: 93533087-93533087 on Build GRCh38
rs760314460 Chr.1: 93532944-93532944 on Build GRCh38
rs1021522704 Chr.1: 93533263-93533263 on Build GRCh38
rs948193590 Chr.1: 93533166-93533166 on Build GRCh38
rs237437 Chr.1: 93533123-93533123 on Build GRCh38
rs560168338 Chr.1: 93532876-93532876 on Build GRCh38
rs777591453 Chr.1: 93532968-93532968 on Build GRCh38
rs545294939 Chr.1: 93532874-93532874 on Build GRCh38
rs569701255 Chr.1: 93533259-93533259 on Build GRCh38
rs528683853 Chr.1: 93533263-93533265 on Build GRCh38
rs779301575 Chr.1: 93533014-93533014 on Build GRCh38
rs749206535 Chr.1: 93533137-93533137 on Build GRCh38
rs569650501 Chr.1: 93533293-93533293 on Build GRCh38
rs1045226347 Chr.1: 93533188-93533188 on Build GRCh38
rs752863040 Chr.1: 93532868-93532868 on Build GRCh38
rs772674673 Chr.1: 93533048-93533048 on Build GRCh38
rs1052218744 Chr.1: 93532879-93532879 on Build GRCh38
rs867057992 Chr.1: 93533178-93533178 on Build GRCh38
rs867743845 Chr.1: 93533084-93533085 on Build GRCh38
rs1002517607 Chr.1: 93532966-93532966 on Build GRCh38
rs1006573196 Chr.1: 93533148-93533148 on Build GRCh38
rs750564377 Chr.1: 93533101-93533104 on Build GRCh38
rs374671121 Chr.1: 93532952-93532952 on Build GRCh38
rs780375202 Chr.1: 93532998-93532998 on Build GRCh38
rs756385190 Chr.1: 93532995-93532995 on Build GRCh38
rs974785994 Chr.1: 93532869-93532869 on Build GRCh38
rs529714856 Chr.1: 93532831-93532831 on Build GRCh38
rs775967671 Chr.1: 93532901-93532901 on Build GRCh38
rs554583502 Chr.1: 93533183-93533183 on Build GRCh38
rs548846333 Chr.1: 93533286-93533287 on Build GRCh38
rs757003263 Chr.1: 93533233-93533233 on Build GRCh38
rs973583752 Chr.1: 93533144-93533144 on Build GRCh38
rs201658281 Chr.1: 93533084-93533084 on Build GRCh38
rs905601933 Chr.1: 93533196-93533196 on Build GRCh38
rs774705770 Chr.1: 93533263-93533266 on Build GRCh38

View Full Product Details