Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29232215-29232723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMID1
Entrez Gene ID
129080
Gene Name
EMI domain containing 1
Gene Aliases
EMI5, EMU1
UniGene
Hs.289106
Gene Chromosome Location
Chr.22: 29205912-29259597 on Build GRCh38
Transcripts
NM_001267895.1 NM_133455.3 XM_005261328.2 XM_005261329.2 XM_011529868.2 XM_011529869.2 XM_011529870.2 XM_011529871.2 XM_011529872.2 XM_011529873.2 XM_011529874.2 XM_011529875.2 XM_011529876.1 XM_017028589.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6389689 c.722G>A p.G241D Chr.22: 29232301-29232301 on Build GRCh38
3553100 c.690C>T p.P230P Chr.22: 29232269-29232269 on Build GRCh38
7091044 c.780C>A p.P260P Chr.22: 29232359-29232359 on Build GRCh38
3553102 c.772C>T p.P258S Chr.22: 29232351-29232351 on Build GRCh38
6433092 c.731G>T p.G244V Chr.22: 29232310-29232310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772441794 Chr.22: 29232342-29232350 on Build GRCh38
rs559904498 Chr.22: 29232440-29232440 on Build GRCh38
rs567572427 Chr.22: 29232685-29232685 on Build GRCh38
rs1025238493 Chr.22: 29232444-29232444 on Build GRCh38
rs747662850 Chr.22: 29232356-29232356 on Build GRCh38
rs757667430 Chr.22: 29232259-29232259 on Build GRCh38
rs1056317078 Chr.22: 29232266-29232266 on Build GRCh38
rs369552985 Chr.22: 29232288-29232288 on Build GRCh38
rs1003703 Chr.22: 29232626-29232626 on Build GRCh38
rs959484019 Chr.22: 29232400-29232400 on Build GRCh38
rs751941516 Chr.22: 29232253-29232253 on Build GRCh38
rs182938652 Chr.22: 29232637-29232637 on Build GRCh38
rs915063079 Chr.22: 29232434-29232434 on Build GRCh38
rs984363795 Chr.22: 29232501-29232501 on Build GRCh38
rs775716975 Chr.22: 29232369-29232369 on Build GRCh38
rs754539460 Chr.22: 29232350-29232350 on Build GRCh38
rs774531707 Chr.22: 29232635-29232635 on Build GRCh38
rs115104475 Chr.22: 29232625-29232625 on Build GRCh38
rs531311071 Chr.22: 29232560-29232560 on Build GRCh38
rs1037210887 Chr.22: 29232565-29232565 on Build GRCh38
rs910125893 Chr.22: 29232514-29232514 on Build GRCh38
rs759206152 Chr.22: 29232650-29232650 on Build GRCh38
rs778622895 Chr.22: 29232436-29232436 on Build GRCh38
rs1033395799 Chr.22: 29232378-29232378 on Build GRCh38
rs748444532 Chr.22: 29232339-29232365 on Build GRCh38
rs537967185 Chr.22: 29232398-29232398 on Build GRCh38
rs772024266 Chr.22: 29232287-29232287 on Build GRCh38
rs754626449 Chr.22: 29232430-29232430 on Build GRCh38
rs377342880 Chr.22: 29232321-29232321 on Build GRCh38
rs555505087 Chr.22: 29232285-29232285 on Build GRCh38
rs199875803 Chr.22: 29232332-29232332 on Build GRCh38
rs773375108 Chr.22: 29232363-29232383 on Build GRCh38
rs773118816 Chr.22: 29232357-29232357 on Build GRCh38
rs944016171 Chr.22: 29232508-29232508 on Build GRCh38
rs577417674 Chr.22: 29232423-29232423 on Build GRCh38
rs201706040 Chr.22: 29232329-29232329 on Build GRCh38
rs566383118 Chr.22: 29232484-29232484 on Build GRCh38
rs201785515 Chr.22: 29232294-29232294 on Build GRCh38
rs746207260 Chr.22: 29232268-29232268 on Build GRCh38
rs552051943 Chr.22: 29232633-29232633 on Build GRCh38
rs528009329 Chr.22: 29232680-29232680 on Build GRCh38
rs749765391 Chr.22: 29232465-29232465 on Build GRCh38
rs767985172 Chr.22: 29232278-29232278 on Build GRCh38
rs749783442 Chr.22: 29232265-29232266 on Build GRCh38
rs775629159 Chr.22: 29232297-29232297 on Build GRCh38
rs774668612 Chr.22: 29232639-29232639 on Build GRCh38
rs543157675 Chr.22: 29232589-29232589 on Build GRCh38
rs1018850528 Chr.22: 29232693-29232693 on Build GRCh38
rs373030911 Chr.22: 29232318-29232318 on Build GRCh38
rs781725508 Chr.22: 29232262-29232262 on Build GRCh38
rs757748555 Chr.22: 29232306-29232306 on Build GRCh38
rs756180356 Chr.22: 29232307-29232307 on Build GRCh38
rs779841392 Chr.22: 29232272-29232272 on Build GRCh38
rs950140689 Chr.22: 29232546-29232546 on Build GRCh38
rs1048569581 Chr.22: 29232557-29232557 on Build GRCh38
rs201201625 Chr.22: 29232335-29232335 on Build GRCh38
rs924616184 Chr.22: 29232495-29232495 on Build GRCh38
rs16987293 Chr.22: 29232681-29232681 on Build GRCh38
rs766801220 Chr.22: 29232426-29232426 on Build GRCh38
rs900004071 Chr.22: 29232264-29232264 on Build GRCh38
rs544760924 Chr.22: 29232439-29232439 on Build GRCh38
rs768896217 Chr.22: 29232387-29232387 on Build GRCh38
rs752839840 Chr.22: 29232238-29232238 on Build GRCh38
rs545100700 Chr.22: 29232481-29232481 on Build GRCh38
rs965774709 Chr.22: 29232701-29232701 on Build GRCh38
rs779331447 Chr.22: 29232336-29232353 on Build GRCh38
rs906210726 Chr.22: 29232616-29232616 on Build GRCh38
rs573961947 Chr.22: 29232304-29232304 on Build GRCh38
rs748853539 Chr.22: 29232337-29232337 on Build GRCh38
rs534503563 Chr.22: 29232275-29232275 on Build GRCh38
rs764672609 Chr.22: 29232247-29232247 on Build GRCh38
rs76367973 Chr.22: 29232271-29232271 on Build GRCh38
rs774365327 Chr.22: 29232388-29232388 on Build GRCh38
rs867549274 Chr.22: 29232545-29232545 on Build GRCh38
rs893665283 Chr.22: 29232347-29232347 on Build GRCh38
rs924635699 Chr.22: 29232587-29232587 on Build GRCh38
rs151003653 Chr.22: 29232239-29232239 on Build GRCh38
rs750896070 Chr.22: 29232419-29232419 on Build GRCh38
rs935979049 Chr.22: 29232613-29232613 on Build GRCh38
rs1005173391 Chr.22: 29232697-29232697 on Build GRCh38
rs747987026 Chr.22: 29232286-29232286 on Build GRCh38
rs755498438 Chr.22: 29232321-29232338 on Build GRCh38
rs756491503 Chr.22: 29232320-29232320 on Build GRCh38
rs888612809 Chr.22: 29232559-29232559 on Build GRCh38
rs546220988 Chr.22: 29232542-29232542 on Build GRCh38
rs1041546047 Chr.22: 29232537-29232537 on Build GRCh38
rs560738360 Chr.22: 29232543-29232543 on Build GRCh38
rs959167745 Chr.22: 29232526-29232526 on Build GRCh38
rs763032422 Chr.22: 29232383-29232383 on Build GRCh38
rs778336091 Chr.22: 29232352-29232352 on Build GRCh38
rs780589393 Chr.22: 29232322-29232322 on Build GRCh38
rs746817352 Chr.22: 29232364-29232364 on Build GRCh38
rs371020814 Chr.22: 29232405-29232405 on Build GRCh38
rs770643528 Chr.22: 29232365-29232365 on Build GRCh38

View Full Product Details