Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30285924-30286416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATSL3
Entrez Gene ID
652968
Gene Name
GATS protein like 3
Gene Aliases
CASTOR1
UniGene
Hs.444950
Gene Chromosome Location
Chr.22: 30285118-30289627 on Build GRCh38
Transcripts
NM_001037666.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7425750 c.751A>T p.S251C Chr.22: 30286091-30286091 on Build GRCh38
3553262 c.665C>T p.P222L Chr.22: 30286341-30286341 on Build GRCh38
4829834 c.800G>A p.R267H Chr.22: 30286042-30286042 on Build GRCh38
4801333 c.777G>T p.S259S Chr.22: 30286065-30286065 on Build GRCh38
1033247 c.699C>T p.I233I Chr.22: 30286307-30286307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776262884 Chr.22: 30285989-30285989 on Build GRCh38
rs1000404870 Chr.22: 30286064-30286064 on Build GRCh38
rs778164967 Chr.22: 30286108-30286108 on Build GRCh38
rs772758847 Chr.22: 30285977-30285977 on Build GRCh38
rs758898589 Chr.22: 30286374-30286374 on Build GRCh38
rs891910469 Chr.22: 30286101-30286101 on Build GRCh38
rs769461138 Chr.22: 30286310-30286310 on Build GRCh38
rs202028147 Chr.22: 30286000-30286000 on Build GRCh38
rs767247380 Chr.22: 30286333-30286333 on Build GRCh38
rs779487292 Chr.22: 30286015-30286015 on Build GRCh38
rs760639054 Chr.22: 30286393-30286393 on Build GRCh38
rs772134409 Chr.22: 30286269-30286270 on Build GRCh38
rs758913505 Chr.22: 30286253-30286253 on Build GRCh38
rs750642300 Chr.22: 30286037-30286037 on Build GRCh38
rs745481482 Chr.22: 30286146-30286146 on Build GRCh38
rs1021389156 Chr.22: 30286027-30286027 on Build GRCh38
rs367889379 Chr.22: 30286335-30286335 on Build GRCh38
rs765264145 Chr.22: 30285993-30285993 on Build GRCh38
rs772350946 Chr.22: 30286106-30286106 on Build GRCh38
rs759719627 Chr.22: 30286341-30286341 on Build GRCh38
rs769373208 Chr.22: 30286145-30286145 on Build GRCh38
rs755070256 Chr.22: 30286245-30286245 on Build GRCh38
rs906024231 Chr.22: 30286038-30286038 on Build GRCh38
rs565934560 Chr.22: 30286172-30286172 on Build GRCh38
rs773513712 Chr.22: 30286312-30286313 on Build GRCh38
rs772421093 Chr.22: 30285973-30285973 on Build GRCh38
rs201713389 Chr.22: 30286128-30286128 on Build GRCh38
rs753908989 Chr.22: 30286065-30286065 on Build GRCh38
rs572104208 Chr.22: 30286036-30286036 on Build GRCh38
rs940163335 Chr.22: 30285960-30285960 on Build GRCh38
rs955277495 Chr.22: 30286195-30286195 on Build GRCh38
rs749592222 Chr.22: 30286376-30286376 on Build GRCh38
rs907545940 Chr.22: 30286302-30286302 on Build GRCh38
rs769138635 Chr.22: 30285961-30285961 on Build GRCh38
rs962847535 Chr.22: 30286306-30286306 on Build GRCh38
rs763835841 Chr.22: 30286236-30286236 on Build GRCh38
rs931497511 Chr.22: 30286331-30286331 on Build GRCh38
rs376300447 Chr.22: 30285983-30285983 on Build GRCh38
rs941241835 Chr.22: 30285981-30285981 on Build GRCh38
rs756306909 Chr.22: 30286043-30286043 on Build GRCh38
rs373022461 Chr.22: 30285965-30285965 on Build GRCh38
rs771714895 Chr.22: 30286239-30286239 on Build GRCh38
rs753180298 Chr.22: 30286246-30286246 on Build GRCh38
rs374308991 Chr.22: 30286329-30286329 on Build GRCh38
rs759609513 Chr.22: 30285990-30285990 on Build GRCh38
rs778060456 Chr.22: 30286254-30286254 on Build GRCh38
rs531606907 Chr.22: 30286328-30286328 on Build GRCh38
rs1035987181 Chr.22: 30286255-30286255 on Build GRCh38
rs748636183 Chr.22: 30286072-30286072 on Build GRCh38
rs561008977 Chr.22: 30286200-30286200 on Build GRCh38
rs754000045 Chr.22: 30286243-30286243 on Build GRCh38
rs745556150 Chr.22: 30286390-30286390 on Build GRCh38
rs774763697 Chr.22: 30285964-30285964 on Build GRCh38
rs200317782 Chr.22: 30285988-30285988 on Build GRCh38
rs776349713 Chr.22: 30286141-30286141 on Build GRCh38
rs370723725 Chr.22: 30285984-30285984 on Build GRCh38
rs765442258 Chr.22: 30285989-30285991 on Build GRCh38
rs762856019 Chr.22: 30286319-30286319 on Build GRCh38
rs745470736 Chr.22: 30286304-30286304 on Build GRCh38
rs547927578 Chr.22: 30286098-30286098 on Build GRCh38
rs188064023 Chr.22: 30286017-30286017 on Build GRCh38
rs765279131 Chr.22: 30286371-30286371 on Build GRCh38
rs770698252 Chr.22: 30286133-30286133 on Build GRCh38
rs891849043 Chr.22: 30286005-30286005 on Build GRCh38
rs748558260 Chr.22: 30286117-30286123 on Build GRCh38
rs775034458 Chr.22: 30286313-30286313 on Build GRCh38
rs376001918 Chr.22: 30286001-30286001 on Build GRCh38
rs766590935 Chr.22: 30286240-30286240 on Build GRCh38
rs1044650086 Chr.22: 30286364-30286364 on Build GRCh38
rs757719774 Chr.22: 30286384-30286384 on Build GRCh38
rs751355467 Chr.22: 30286237-30286237 on Build GRCh38
rs1029346314 Chr.22: 30286153-30286153 on Build GRCh38
rs754982494 Chr.22: 30286066-30286066 on Build GRCh38
rs771356842 Chr.22: 30286285-30286285 on Build GRCh38
rs780165118 Chr.22: 30286046-30286046 on Build GRCh38
rs370649027 Chr.22: 30286016-30286016 on Build GRCh38
rs371123714 Chr.22: 30286142-30286142 on Build GRCh38
rs781657051 Chr.22: 30286386-30286386 on Build GRCh38
rs1029674366 Chr.22: 30286152-30286152 on Build GRCh38
rs769966102 Chr.22: 30286085-30286085 on Build GRCh38
rs754087927 Chr.22: 30286334-30286334 on Build GRCh38
rs775313464 Chr.22: 30286149-30286149 on Build GRCh38
rs747378262 Chr.22: 30286269-30286269 on Build GRCh38
rs987576084 Chr.22: 30286244-30286244 on Build GRCh38
rs781008682 Chr.22: 30286303-30286303 on Build GRCh38
rs1050352549 Chr.22: 30286171-30286200 on Build GRCh38
rs762908621 Chr.22: 30286230-30286230 on Build GRCh38
rs760954094 Chr.22: 30286238-30286238 on Build GRCh38

View Full Product Details