Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30287243-30287735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATSL3
Entrez Gene ID
652968
Gene Name
GATS protein like 3
Gene Aliases
CASTOR1
UniGene
Hs.444950
Gene Chromosome Location
Chr.22: 30285118-30289627 on Build GRCh38
Transcripts
NM_001037666.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7369260 c.407C>A p.T136K Chr.22: 30287253-30287253 on Build GRCh38
4842023 c.261G>A p.A87A Chr.22: 30287484-30287484 on Build GRCh38
4428444 c.246_248delGTC p.S83delS Chr.22: 30287497-30287499 on Build GRCh38
4420541 c.257G>A p.G86D Chr.22: 30287488-30287488 on Build GRCh38
6453835 c.319G>A p.E107K Chr.22: 30287426-30287426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759489489 Chr.22: 30287437-30287437 on Build GRCh38
rs760863995 Chr.22: 30287512-30287512 on Build GRCh38
rs747812777 Chr.22: 30287417-30287417 on Build GRCh38
rs532340147 Chr.22: 30287308-30287308 on Build GRCh38
rs975515066 Chr.22: 30287478-30287478 on Build GRCh38
rs774190144 Chr.22: 30287624-30287625 on Build GRCh38
rs893574631 Chr.22: 30287616-30287616 on Build GRCh38
rs746711882 Chr.22: 30287598-30287598 on Build GRCh38
rs756121324 Chr.22: 30287355-30287355 on Build GRCh38
rs776262740 Chr.22: 30287490-30287490 on Build GRCh38
rs550057547 Chr.22: 30287571-30287571 on Build GRCh38
rs1020524089 Chr.22: 30287384-30287384 on Build GRCh38
rs547890105 Chr.22: 30287464-30287464 on Build GRCh38
rs967530986 Chr.22: 30287665-30287665 on Build GRCh38
rs766503111 Chr.22: 30287520-30287520 on Build GRCh38
rs759275416 Chr.22: 30287313-30287313 on Build GRCh38
rs905015882 Chr.22: 30287605-30287605 on Build GRCh38
rs1049003084 Chr.22: 30287480-30287480 on Build GRCh38
rs1033306736 Chr.22: 30287694-30287694 on Build GRCh38
rs1009214816 Chr.22: 30287647-30287647 on Build GRCh38
rs189381704 Chr.22: 30287485-30287485 on Build GRCh38
rs773905426 Chr.22: 30287266-30287266 on Build GRCh38
rs368955430 Chr.22: 30287449-30287449 on Build GRCh38
rs780554358 Chr.22: 30287363-30287363 on Build GRCh38
rs773460979 Chr.22: 30287510-30287510 on Build GRCh38
rs752526947 Chr.22: 30287445-30287445 on Build GRCh38
rs768939653 Chr.22: 30287386-30287386 on Build GRCh38
rs2240421 Chr.22: 30287455-30287455 on Build GRCh38
rs375767108 Chr.22: 30287499-30287499 on Build GRCh38
rs368151687 Chr.22: 30287484-30287484 on Build GRCh38
rs1002474096 Chr.22: 30287610-30287610 on Build GRCh38
rs529764080 Chr.22: 30287454-30287454 on Build GRCh38
rs766421774 Chr.22: 30287303-30287303 on Build GRCh38
rs780075887 Chr.22: 30287465-30287465 on Build GRCh38
rs781308218 Chr.22: 30287339-30287339 on Build GRCh38
rs765044634 Chr.22: 30287325-30287325 on Build GRCh38
rs377272415 Chr.22: 30287267-30287267 on Build GRCh38
rs569590873 Chr.22: 30287482-30287482 on Build GRCh38
rs868349810 Chr.22: 30287660-30287660 on Build GRCh38
rs772929352 Chr.22: 30287426-30287426 on Build GRCh38
rs770514255 Chr.22: 30287430-30287430 on Build GRCh38
rs748546063 Chr.22: 30287479-30287479 on Build GRCh38
rs374442282 Chr.22: 30287336-30287336 on Build GRCh38
rs370157427 Chr.22: 30287504-30287504 on Build GRCh38
rs753947047 Chr.22: 30287522-30287522 on Build GRCh38
rs779207814 Chr.22: 30287477-30287477 on Build GRCh38
rs755496950 Chr.22: 30287474-30287474 on Build GRCh38
rs772831027 Chr.22: 30287285-30287285 on Build GRCh38
rs760272584 Chr.22: 30287427-30287427 on Build GRCh38
rs957677237 Chr.22: 30287309-30287309 on Build GRCh38
rs770608537 Chr.22: 30287489-30287489 on Build GRCh38
rs769766060 Chr.22: 30287500-30287500 on Build GRCh38
rs147532076 Chr.22: 30287678-30287678 on Build GRCh38
rs5753058 Chr.22: 30287296-30287296 on Build GRCh38
rs868129388 Chr.22: 30287615-30287615 on Build GRCh38
rs758863424 Chr.22: 30287578-30287578 on Build GRCh38
rs769129448 Chr.22: 30287612-30287612 on Build GRCh38
rs754942237 Chr.22: 30287544-30287544 on Build GRCh38
rs772989090 Chr.22: 30287661-30287661 on Build GRCh38
rs745388479 Chr.22: 30287609-30287609 on Build GRCh38
rs750588499 Chr.22: 30287453-30287453 on Build GRCh38
rs1000389680 Chr.22: 30287670-30287670 on Build GRCh38
rs749410812 Chr.22: 30287420-30287420 on Build GRCh38
rs753767134 Chr.22: 30287306-30287306 on Build GRCh38
rs374275854 Chr.22: 30287439-30287439 on Build GRCh38
rs767731570 Chr.22: 30287452-30287452 on Build GRCh38
rs780637456 Chr.22: 30287608-30287608 on Build GRCh38
rs199977995 Chr.22: 30287354-30287354 on Build GRCh38
rs757482864 Chr.22: 30287337-30287337 on Build GRCh38
rs751300008 Chr.22: 30287593-30287594 on Build GRCh38
rs771531140 Chr.22: 30287283-30287283 on Build GRCh38
rs370113637 Chr.22: 30287385-30287385 on Build GRCh38
rs200867191 Chr.22: 30287287-30287287 on Build GRCh38
rs763829995 Chr.22: 30287505-30287505 on Build GRCh38
rs536912538 Chr.22: 30287583-30287583 on Build GRCh38
rs557296879 Chr.22: 30287471-30287471 on Build GRCh38
rs371777166 Chr.22: 30287436-30287436 on Build GRCh38
rs201164151 Chr.22: 30287450-30287450 on Build GRCh38
rs779314689 Chr.22: 30287406-30287406 on Build GRCh38

View Full Product Details