Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31399339-31399846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRG1
Entrez Gene ID
4733
Gene Name
developmentally regulated GTP binding protein 1
Gene Aliases
NEDD3
UniGene
Hs.115242
Gene Chromosome Location
Chr.22: 31399553-31434186 on Build GRCh38
Transcripts
NM_004147.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160579 c.24C>T p.I8I Chr.22: 31399707-31399707 on Build GRCh38
239647 c.29A>T p.E10V Chr.22: 31399712-31399712 on Build GRCh38
5663770 c.27G>T p.A9A Chr.22: 31399710-31399710 on Build GRCh38
5690598 c.30G>T p.E10D Chr.22: 31399713-31399713 on Build GRCh38
3785658 c.30G>A p.E10E Chr.22: 31399713-31399713 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372906460 Chr.22: 31399638-31399638 on Build GRCh38
rs1038202467 Chr.22: 31399794-31399794 on Build GRCh38
rs1046397375 Chr.22: 31399395-31399395 on Build GRCh38
rs898615697 Chr.22: 31399627-31399627 on Build GRCh38
rs772096001 Chr.22: 31399661-31399661 on Build GRCh38
rs1041446339 Chr.22: 31399383-31399383 on Build GRCh38
rs908480648 Chr.22: 31399781-31399781 on Build GRCh38
rs760246786 Chr.22: 31399675-31399675 on Build GRCh38
rs760008728 Chr.22: 31399631-31399631 on Build GRCh38
rs910378823 Chr.22: 31399369-31399369 on Build GRCh38
rs530513423 Chr.22: 31399744-31399744 on Build GRCh38
rs776210701 Chr.22: 31399679-31399679 on Build GRCh38
rs113639921 Chr.22: 31399434-31399434 on Build GRCh38
rs1017491797 Chr.22: 31399760-31399760 on Build GRCh38
rs992422701 Chr.22: 31399530-31399530 on Build GRCh38
rs778798878 Chr.22: 31399642-31399642 on Build GRCh38
rs765631156 Chr.22: 31399632-31399632 on Build GRCh38
rs1056273871 Chr.22: 31399517-31399517 on Build GRCh38
rs374352220 Chr.22: 31399771-31399771 on Build GRCh38
rs957141612 Chr.22: 31399667-31399667 on Build GRCh38
rs879755552 Chr.22: 31399621-31399621 on Build GRCh38
rs748033226 Chr.22: 31399743-31399743 on Build GRCh38
rs544641928 Chr.22: 31399362-31399362 on Build GRCh38
rs1004262970 Chr.22: 31399451-31399451 on Build GRCh38
rs772697154 Chr.22: 31399662-31399662 on Build GRCh38
rs1039659900 Chr.22: 31399396-31399396 on Build GRCh38
rs966867395 Chr.22: 31399551-31399551 on Build GRCh38
rs937925727 Chr.22: 31399509-31399509 on Build GRCh38
rs966601672 Chr.22: 31399807-31399807 on Build GRCh38
rs768285736 Chr.22: 31399641-31399641 on Build GRCh38
rs929027611 Chr.22: 31399714-31399714 on Build GRCh38
rs564602848 Chr.22: 31399422-31399422 on Build GRCh38
rs368813775 Chr.22: 31399635-31399635 on Build GRCh38
rs764536786 Chr.22: 31399633-31399633 on Build GRCh38
rs560757286 Chr.22: 31399527-31399527 on Build GRCh38
rs73160605 Chr.22: 31399783-31399783 on Build GRCh38
rs766501589 Chr.22: 31399625-31399625 on Build GRCh38
rs989541057 Chr.22: 31399388-31399388 on Build GRCh38
rs770435340 Chr.22: 31399677-31399677 on Build GRCh38
rs945823919 Chr.22: 31399380-31399380 on Build GRCh38
rs746965969 Chr.22: 31399768-31399768 on Build GRCh38
rs935622354 Chr.22: 31399393-31399393 on Build GRCh38
rs764329018 Chr.22: 31399699-31399699 on Build GRCh38
rs1033629091 Chr.22: 31399523-31399523 on Build GRCh38
rs762210000 Chr.22: 31399705-31399705 on Build GRCh38
rs540437070 Chr.22: 31399513-31399513 on Build GRCh38
rs751625606 Chr.22: 31399690-31399690 on Build GRCh38
rs571360551 Chr.22: 31399720-31399720 on Build GRCh38
rs181315466 Chr.22: 31399659-31399659 on Build GRCh38
rs935717204 Chr.22: 31399573-31399573 on Build GRCh38
rs920931131 Chr.22: 31399594-31399594 on Build GRCh38
rs778551099 Chr.22: 31399742-31399742 on Build GRCh38
rs773908059 Chr.22: 31399391-31399391 on Build GRCh38
rs914068680 Chr.22: 31399364-31399364 on Build GRCh38
rs1021192221 Chr.22: 31399788-31399788 on Build GRCh38
rs35624041 Chr.22: 31399586-31399587 on Build GRCh38
rs756397608 Chr.22: 31399637-31399637 on Build GRCh38
rs1051472129 Chr.22: 31399767-31399767 on Build GRCh38
rs372320146 Chr.22: 31399624-31399624 on Build GRCh38
rs975157808 Chr.22: 31399670-31399670 on Build GRCh38
rs756203478 Chr.22: 31399721-31399721 on Build GRCh38
rs886955517 Chr.22: 31399619-31399619 on Build GRCh38
rs749532013 Chr.22: 31399639-31399639 on Build GRCh38
rs990995173 Chr.22: 31399653-31399653 on Build GRCh38
rs771734400 Chr.22: 31399567-31399567 on Build GRCh38
rs758494034 Chr.22: 31399649-31399649 on Build GRCh38
rs998099563 Chr.22: 31399435-31399435 on Build GRCh38
rs559711206 Chr.22: 31399614-31399614 on Build GRCh38
rs771972575 Chr.22: 31399659-31399660 on Build GRCh38
rs768133885 Chr.22: 31399707-31399707 on Build GRCh38
rs372584389 Chr.22: 31399361-31399361 on Build GRCh38
rs750836357 Chr.22: 31399626-31399626 on Build GRCh38
rs933905216 Chr.22: 31399605-31399605 on Build GRCh38
rs773029291 Chr.22: 31399753-31399754 on Build GRCh38
rs1039416567 Chr.22: 31399590-31399590 on Build GRCh38
rs1055339252 Chr.22: 31399413-31399413 on Build GRCh38
rs1002185763 Chr.22: 31399520-31399520 on Build GRCh38
rs550587370 Chr.22: 31399745-31399745 on Build GRCh38
rs941530972 Chr.22: 31399433-31399433 on Build GRCh38
rs993141452 Chr.22: 31399439-31399439 on Build GRCh38
rs780301658 Chr.22: 31399722-31399722 on Build GRCh38
rs368688522 Chr.22: 31399618-31399618 on Build GRCh38
rs375750965 Chr.22: 31399698-31399698 on Build GRCh38
rs199760860 Chr.22: 31399634-31399634 on Build GRCh38
rs759258892 Chr.22: 31399695-31399695 on Build GRCh38
rs979619111 Chr.22: 31399655-31399655 on Build GRCh38
rs958148829 Chr.22: 31399387-31399387 on Build GRCh38
rs925667676 Chr.22: 31399571-31399571 on Build GRCh38
rs933642930 Chr.22: 31399750-31399750 on Build GRCh38
rs764142771 Chr.22: 31399666-31399666 on Build GRCh38
rs752000126 Chr.22: 31399701-31399701 on Build GRCh38
rs755238238 Chr.22: 31399808-31399808 on Build GRCh38
rs755268077 Chr.22: 31399735-31399735 on Build GRCh38
rs754073105 Chr.22: 31399732-31399732 on Build GRCh38
rs914057626 Chr.22: 31399544-31399544 on Build GRCh38
rs189087736 Chr.22: 31399437-31399437 on Build GRCh38
rs890085172 Chr.22: 31399739-31399739 on Build GRCh38
rs886907548 Chr.22: 31399440-31399440 on Build GRCh38
rs374527751 Chr.22: 31399470-31399470 on Build GRCh38
rs961285637 Chr.22: 31399529-31399529 on Build GRCh38
rs957861046 Chr.22: 31399557-31399557 on Build GRCh38
rs926573603 Chr.22: 31399504-31399504 on Build GRCh38
rs1016946023 Chr.22: 31399462-31399462 on Build GRCh38
rs898334036 Chr.22: 31399824-31399824 on Build GRCh38

View Full Product Details