Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 31420177-31420676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRG1
Entrez Gene ID
4733
Gene Name
developmentally regulated GTP binding protein 1
Gene Aliases
NEDD3
UniGene
Hs.115242
Gene Chromosome Location
Chr.22: 31399553-31434186 on Build GRCh38
Transcripts
NM_004147.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2938169 c.541T>A p.F181I Chr.22: 31420384-31420384 on Build GRCh38
419593 c.456G>A p.L152L Chr.22: 31420299-31420299 on Build GRCh38
1033456 c.477G>A p.K159K Chr.22: 31420320-31420320 on Build GRCh38
5885416 c.560G>C p.G187A Chr.22: 31420403-31420403 on Build GRCh38
4882050 c.534C>A p.N178K Chr.22: 31420377-31420377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758080107 Chr.22: 31420318-31420318 on Build GRCh38
rs764659097 Chr.22: 31420232-31420235 on Build GRCh38
rs1015888357 Chr.22: 31420408-31420408 on Build GRCh38
rs201965741 Chr.22: 31420235-31420235 on Build GRCh38
rs200429738 Chr.22: 31420231-31420231 on Build GRCh38
rs779871347 Chr.22: 31420565-31420565 on Build GRCh38
rs780368583 Chr.22: 31420351-31420351 on Build GRCh38
rs368152350 Chr.22: 31420225-31420225 on Build GRCh38
rs751214634 Chr.22: 31420336-31420336 on Build GRCh38
rs765204779 Chr.22: 31420416-31420416 on Build GRCh38
rs372624238 Chr.22: 31420232-31420232 on Build GRCh38
rs200607015 Chr.22: 31420435-31420435 on Build GRCh38
rs758030539 Chr.22: 31420212-31420212 on Build GRCh38
rs781481591 Chr.22: 31420315-31420317 on Build GRCh38
rs148069806 Chr.22: 31420373-31420373 on Build GRCh38
rs766054397 Chr.22: 31420281-31420281 on Build GRCh38
rs772188007 Chr.22: 31420379-31420379 on Build GRCh38
rs191733493 Chr.22: 31420418-31420418 on Build GRCh38
rs757508879 Chr.22: 31420244-31420244 on Build GRCh38
rs532120254 Chr.22: 31420488-31420488 on Build GRCh38
rs199705711 Chr.22: 31420270-31420270 on Build GRCh38
rs974209870 Chr.22: 31420238-31420238 on Build GRCh38
rs763597562 Chr.22: 31420232-31420232 on Build GRCh38
rs755430359 Chr.22: 31420369-31420369 on Build GRCh38
rs756502164 Chr.22: 31420219-31420219 on Build GRCh38
rs1045368747 Chr.22: 31420554-31420554 on Build GRCh38
rs2273248 Chr.22: 31420453-31420453 on Build GRCh38
rs752068607 Chr.22: 31420601-31420601 on Build GRCh38
rs879078533 Chr.22: 31420471-31420471 on Build GRCh38
rs369426199 Chr.22: 31420266-31420266 on Build GRCh38
rs556285945 Chr.22: 31420536-31420536 on Build GRCh38
rs530662815 Chr.22: 31420646-31420646 on Build GRCh38
rs145716937 Chr.22: 31420417-31420418 on Build GRCh38
rs999597564 Chr.22: 31420514-31420514 on Build GRCh38
rs938170954 Chr.22: 31420504-31420504 on Build GRCh38
rs961921597 Chr.22: 31420411-31420411 on Build GRCh38
rs374540101 Chr.22: 31420382-31420382 on Build GRCh38
rs200991435 Chr.22: 31420213-31420213 on Build GRCh38
rs199993249 Chr.22: 31420444-31420444 on Build GRCh38
rs931923666 Chr.22: 31420497-31420497 on Build GRCh38
rs969615786 Chr.22: 31420233-31420233 on Build GRCh38
rs924582340 Chr.22: 31420611-31420611 on Build GRCh38
rs879810421 Chr.22: 31420493-31420493 on Build GRCh38
rs770670606 Chr.22: 31420391-31420391 on Build GRCh38
rs762432870 Chr.22: 31420419-31420419 on Build GRCh38
rs75732306 Chr.22: 31420505-31420505 on Build GRCh38
rs746280319 Chr.22: 31420217-31420217 on Build GRCh38
rs370820729 Chr.22: 31420374-31420374 on Build GRCh38
rs764869196 Chr.22: 31420290-31420290 on Build GRCh38
rs776525243 Chr.22: 31420413-31420413 on Build GRCh38
rs1052924619 Chr.22: 31420548-31420548 on Build GRCh38
rs751213154 Chr.22: 31420426-31420426 on Build GRCh38
rs536347629 Chr.22: 31420456-31420456 on Build GRCh38
rs756488418 Chr.22: 31420343-31420343 on Build GRCh38
rs903716150 Chr.22: 31420511-31420511 on Build GRCh38
rs750679045 Chr.22: 31420438-31420438 on Build GRCh38
rs763641476 Chr.22: 31420423-31420423 on Build GRCh38
rs201871268 Chr.22: 31420417-31420417 on Build GRCh38
rs567903114 Chr.22: 31420613-31420613 on Build GRCh38
rs376758660 Chr.22: 31420296-31420296 on Build GRCh38
rs375337814 Chr.22: 31420226-31420226 on Build GRCh38
rs763724899 Chr.22: 31420325-31420325 on Build GRCh38
rs1013965786 Chr.22: 31420614-31420614 on Build GRCh38
rs749827762 Chr.22: 31420368-31420368 on Build GRCh38
rs201304954 Chr.22: 31420347-31420347 on Build GRCh38
rs753418924 Chr.22: 31420312-31420312 on Build GRCh38
rs759564970 Chr.22: 31420415-31420415 on Build GRCh38
rs760320234 Chr.22: 31420256-31420256 on Build GRCh38
rs548322843 Chr.22: 31420557-31420557 on Build GRCh38
rs1023631106 Chr.22: 31420631-31420631 on Build GRCh38
rs528515838 Chr.22: 31420398-31420398 on Build GRCh38
rs528377038 Chr.22: 31420542-31420542 on Build GRCh38
rs915626333 Chr.22: 31420519-31420519 on Build GRCh38
rs756895891 Chr.22: 31420432-31420432 on Build GRCh38
rs907638353 Chr.22: 31420393-31420393 on Build GRCh38
rs768680765 Chr.22: 31420227-31420227 on Build GRCh38
rs751996339 Chr.22: 31420232-31420233 on Build GRCh38

View Full Product Details