Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32397604-32398123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTCB
Entrez Gene ID
51493
Gene Name
RNA 2',3'-cyclic phosphate and 5'-OH ligase
Gene Aliases
C22orf28, DJ149A16.6, FAAP, HSPC117
UniGene
Hs.474643
Gene Chromosome Location
Chr.22: 32387575-32412287 on Build GRCh38
Transcripts
NM_014306.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3553751 c.694C>T p.Q232* Chr.22: 32398061-32398061 on Build GRCh38
7238017 c.759G>A p.Q253Q Chr.22: 32397996-32397996 on Build GRCh38
4188447 c.771G>A p.M257I Chr.22: 32397984-32397984 on Build GRCh38
7238134 c.760G>T p.V254L Chr.22: 32397995-32397995 on Build GRCh38
1130446 c.726T>C p.A242A Chr.22: 32398029-32398029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs959024723 Chr.22: 32397989-32397989 on Build GRCh38
rs1057211840 Chr.22: 32397948-32397948 on Build GRCh38
rs769004092 Chr.22: 32398001-32398001 on Build GRCh38
rs772523212 Chr.22: 32398011-32398011 on Build GRCh38
rs748636265 Chr.22: 32398007-32398007 on Build GRCh38
rs1010765504 Chr.22: 32397837-32397837 on Build GRCh38
rs759312250 Chr.22: 32398039-32398039 on Build GRCh38
rs75225516 Chr.22: 32397665-32397665 on Build GRCh38
rs889771477 Chr.22: 32397719-32397719 on Build GRCh38
rs1020333555 Chr.22: 32398031-32398031 on Build GRCh38
rs369895519 Chr.22: 32398058-32398058 on Build GRCh38
rs551423764 Chr.22: 32397930-32397930 on Build GRCh38
rs1037566968 Chr.22: 32397652-32397652 on Build GRCh38
rs907542432 Chr.22: 32397770-32397770 on Build GRCh38
rs184237886 Chr.22: 32397662-32397662 on Build GRCh38
rs1021180557 Chr.22: 32397862-32397862 on Build GRCh38
rs906086748 Chr.22: 32397872-32397872 on Build GRCh38
rs763851320 Chr.22: 32397821-32397821 on Build GRCh38
rs187267463 Chr.22: 32398056-32398056 on Build GRCh38
rs73168721 Chr.22: 32397654-32397654 on Build GRCh38
rs974543777 Chr.22: 32397718-32397718 on Build GRCh38
rs371918269 Chr.22: 32398087-32398087 on Build GRCh38
rs762762096 Chr.22: 32397788-32397788 on Build GRCh38
rs780776411 Chr.22: 32398098-32398098 on Build GRCh38
rs567490670 Chr.22: 32397887-32397887 on Build GRCh38
rs754741994 Chr.22: 32397910-32397910 on Build GRCh38
rs760745019 Chr.22: 32398025-32398025 on Build GRCh38
rs11545748 Chr.22: 32397993-32397993 on Build GRCh38
rs779583062 Chr.22: 32397658-32397658 on Build GRCh38
rs371949637 Chr.22: 32397974-32397974 on Build GRCh38
rs756888848 Chr.22: 32397960-32397960 on Build GRCh38
rs531232451 Chr.22: 32397830-32397830 on Build GRCh38
rs58917959 Chr.22: 32397828-32397828 on Build GRCh38
rs925185244 Chr.22: 32397675-32397675 on Build GRCh38
rs529189308 Chr.22: 32397721-32397721 on Build GRCh38
rs1034731690 Chr.22: 32397966-32397966 on Build GRCh38
rs11545750 Chr.22: 32398038-32398038 on Build GRCh38
rs964917526 Chr.22: 32397696-32397696 on Build GRCh38
rs766520125 Chr.22: 32398030-32398030 on Build GRCh38
rs548682786 Chr.22: 32397731-32397731 on Build GRCh38
rs765172190 Chr.22: 32397741-32397741 on Build GRCh38
rs758021911 Chr.22: 32397937-32397937 on Build GRCh38
rs756779393 Chr.22: 32398092-32398092 on Build GRCh38
rs1007518988 Chr.22: 32397742-32397742 on Build GRCh38
rs765319217 Chr.22: 32397924-32397924 on Build GRCh38
rs895945236 Chr.22: 32398010-32398010 on Build GRCh38
rs771992011 Chr.22: 32398018-32398019 on Build GRCh38
rs777398875 Chr.22: 32397949-32397949 on Build GRCh38
rs1047464697 Chr.22: 32397744-32397744 on Build GRCh38
rs776896020 Chr.22: 32398033-32398033 on Build GRCh38
rs756749502 Chr.22: 32397834-32397834 on Build GRCh38
rs1001686917 Chr.22: 32397877-32397877 on Build GRCh38
rs766585986 Chr.22: 32397900-32397900 on Build GRCh38
rs775270680 Chr.22: 32397773-32397773 on Build GRCh38
rs565053068 Chr.22: 32397649-32397649 on Build GRCh38
rs760832161 Chr.22: 32397892-32397892 on Build GRCh38
rs534345258 Chr.22: 32398072-32398072 on Build GRCh38
rs201313702 Chr.22: 32397908-32397908 on Build GRCh38
rs746477420 Chr.22: 32397950-32397950 on Build GRCh38
rs773197974 Chr.22: 32398014-32398014 on Build GRCh38

View Full Product Details