Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 32401645-32402126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTCB
Entrez Gene ID
51493
Gene Name
RNA 2',3'-cyclic phosphate and 5'-OH ligase
Gene Aliases
C22orf28, DJ149A16.6, FAAP, HSPC117
UniGene
Hs.474643
Gene Chromosome Location
Chr.22: 32387575-32412287 on Build GRCh38
Transcripts
NM_014306.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4103705 c.349G>A p.G117R Chr.22: 32401895-32401895 on Build GRCh38
6035934 c.349G>T p.G117W Chr.22: 32401895-32401895 on Build GRCh38
3553752 c.347T>C p.V116A Chr.22: 32401897-32401897 on Build GRCh38
4879898 c.371T>G p.V124G Chr.22: 32401873-32401873 on Build GRCh38
6035933 c.349G>T p.G117W Chr.22: 32401895-32401895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111814063 Chr.22: 32401949-32401949 on Build GRCh38
rs758258203 Chr.22: 32401884-32401884 on Build GRCh38
rs771756217 Chr.22: 32401782-32401782 on Build GRCh38
rs776148354 Chr.22: 32401814-32401814 on Build GRCh38
rs1035305946 Chr.22: 32402076-32402076 on Build GRCh38
rs373314931 Chr.22: 32401676-32401676 on Build GRCh38
rs764755630 Chr.22: 32401720-32401720 on Build GRCh38
rs753967453 Chr.22: 32401866-32401866 on Build GRCh38
rs956740566 Chr.22: 32401840-32401840 on Build GRCh38
rs957158983 Chr.22: 32402081-32402081 on Build GRCh38
rs759252324 Chr.22: 32401717-32401717 on Build GRCh38
rs774747345 Chr.22: 32401734-32401734 on Build GRCh38
rs781047399 Chr.22: 32401896-32401896 on Build GRCh38
rs1022480247 Chr.22: 32402089-32402089 on Build GRCh38
rs927108323 Chr.22: 32402002-32402002 on Build GRCh38
rs988063998 Chr.22: 32401954-32401954 on Build GRCh38
rs370704850 Chr.22: 32401823-32401823 on Build GRCh38
rs984547974 Chr.22: 32402058-32402058 on Build GRCh38
rs539176815 Chr.22: 32401907-32401907 on Build GRCh38
rs374886725 Chr.22: 32401732-32401732 on Build GRCh38
rs986180825 Chr.22: 32401819-32401819 on Build GRCh38
rs768127346 Chr.22: 32401731-32401731 on Build GRCh38
rs754299636 Chr.22: 32401940-32401940 on Build GRCh38
rs767053488 Chr.22: 32401937-32401937 on Build GRCh38
rs929852182 Chr.22: 32401957-32401957 on Build GRCh38
rs746763996 Chr.22: 32401887-32401887 on Build GRCh38
rs200793446 Chr.22: 32401757-32401757 on Build GRCh38
rs758902352 Chr.22: 32401818-32401818 on Build GRCh38
rs773717146 Chr.22: 32401934-32401934 on Build GRCh38
rs370826873 Chr.22: 32402065-32402065 on Build GRCh38
rs752005092 Chr.22: 32401845-32401867 on Build GRCh38
rs576516923 Chr.22: 32401882-32401882 on Build GRCh38
rs552673577 Chr.22: 32401955-32401955 on Build GRCh38
rs761066121 Chr.22: 32401935-32401935 on Build GRCh38
rs777723724 Chr.22: 32401797-32401797 on Build GRCh38
rs562240206 Chr.22: 32402013-32402013 on Build GRCh38
rs751957205 Chr.22: 32401723-32401723 on Build GRCh38
rs774991787 Chr.22: 32401828-32401828 on Build GRCh38
rs768384779 Chr.22: 32401932-32401932 on Build GRCh38
rs199904567 Chr.22: 32401820-32401820 on Build GRCh38
rs769182638 Chr.22: 32401909-32401909 on Build GRCh38
rs770858359 Chr.22: 32401895-32401895 on Build GRCh38
rs988364985 Chr.22: 32402086-32402086 on Build GRCh38
rs956906672 Chr.22: 32401711-32401711 on Build GRCh38
rs1003590814 Chr.22: 32402053-32402053 on Build GRCh38
rs746925438 Chr.22: 32401805-32401805 on Build GRCh38
rs775258897 Chr.22: 32401914-32401914 on Build GRCh38
rs764594076 Chr.22: 32401843-32401843 on Build GRCh38
rs761297061 Chr.22: 32401860-32401860 on Build GRCh38
rs780303764 Chr.22: 32401740-32401740 on Build GRCh38
rs762535322 Chr.22: 32401931-32401931 on Build GRCh38
rs748037176 Chr.22: 32401767-32401767 on Build GRCh38
rs777564270 Chr.22: 32401886-32401886 on Build GRCh38
rs762278833 Chr.22: 32401830-32401830 on Build GRCh38
rs757504822 Chr.22: 32401724-32401724 on Build GRCh38
rs750769652 Chr.22: 32401846-32401846 on Build GRCh38
rs148510767 Chr.22: 32401875-32401875 on Build GRCh38
rs183969634 Chr.22: 32402092-32402092 on Build GRCh38
rs536642868 Chr.22: 32401838-32401838 on Build GRCh38
rs904884116 Chr.22: 32402014-32402014 on Build GRCh38
rs766928941 Chr.22: 32401862-32401862 on Build GRCh38
rs770803205 Chr.22: 32401813-32401813 on Build GRCh38
rs776310691 Chr.22: 32401698-32401698 on Build GRCh38
rs11545747 Chr.22: 32401786-32401786 on Build GRCh38
rs1005265034 Chr.22: 32401979-32401979 on Build GRCh38
rs7289526 Chr.22: 32401972-32401972 on Build GRCh38
rs749340829 Chr.22: 32401749-32401749 on Build GRCh38
rs910559308 Chr.22: 32401843-32401843 on Build GRCh38
rs201561593 Chr.22: 32401821-32401821 on Build GRCh38
rs377323703 Chr.22: 32401735-32401735 on Build GRCh38
rs573066733 Chr.22: 32401964-32401964 on Build GRCh38
rs908919275 Chr.22: 32402072-32402072 on Build GRCh38

View Full Product Details