Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35656298-35656802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL6
Entrez Gene ID
80830
Gene Name
apolipoprotein L6
Gene Aliases
APOL-VI, APOLVI
UniGene
Hs.257352
Gene Chromosome Location
Chr.22: 35648317-35668409 on Build GRCh38
Transcripts
NM_030641.3 XM_011530392.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5057789 c.15G>A p.A5A Chr.22: 35656440-35656440 on Build GRCh38
1714325 c.34G>T p.G12C Chr.22: 35656459-35656459 on Build GRCh38
4794929 c.34G>A p.G12S Chr.22: 35656459-35656459 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768067400 Chr.22: 35656459-35656459 on Build GRCh38
rs989482120 Chr.22: 35656331-35656331 on Build GRCh38
rs967097145 Chr.22: 35656606-35656606 on Build GRCh38
rs753303937 Chr.22: 35656421-35656421 on Build GRCh38
rs751559917 Chr.22: 35656440-35656440 on Build GRCh38
rs760504680 Chr.22: 35656498-35656498 on Build GRCh38
rs767308597 Chr.22: 35656441-35656441 on Build GRCh38
rs750300064 Chr.22: 35656540-35656540 on Build GRCh38
rs9622242 Chr.22: 35656363-35656363 on Build GRCh38
rs188375278 Chr.22: 35656679-35656679 on Build GRCh38
rs371730868 Chr.22: 35656384-35656384 on Build GRCh38
rs750217115 Chr.22: 35656496-35656496 on Build GRCh38
rs181211939 Chr.22: 35656718-35656718 on Build GRCh38
rs547448969 Chr.22: 35656357-35656357 on Build GRCh38
rs753552603 Chr.22: 35656506-35656506 on Build GRCh38
rs566908522 Chr.22: 35656654-35656654 on Build GRCh38
rs758120602 Chr.22: 35656539-35656539 on Build GRCh38
rs916718732 Chr.22: 35656701-35656701 on Build GRCh38
rs537387869 Chr.22: 35656475-35656475 on Build GRCh38
rs765995194 Chr.22: 35656501-35656501 on Build GRCh38
rs200976901 Chr.22: 35656434-35656434 on Build GRCh38
rs998172207 Chr.22: 35656662-35656662 on Build GRCh38
rs896846900 Chr.22: 35656723-35656723 on Build GRCh38
rs5750145 Chr.22: 35656433-35656433 on Build GRCh38
rs369709207 Chr.22: 35656700-35656700 on Build GRCh38
rs1004658136 Chr.22: 35656711-35656711 on Build GRCh38
rs746303294 Chr.22: 35656472-35656472 on Build GRCh38
rs776944436 Chr.22: 35656414-35656414 on Build GRCh38
rs759851636 Chr.22: 35656419-35656419 on Build GRCh38
rs1001001663 Chr.22: 35656643-35656643 on Build GRCh38
rs750904747 Chr.22: 35656526-35656526 on Build GRCh38
rs755214415 Chr.22: 35656727-35656727 on Build GRCh38
rs576795263 Chr.22: 35656488-35656488 on Build GRCh38
rs1039143359 Chr.22: 35656652-35656652 on Build GRCh38
rs778101319 Chr.22: 35656459-35656459 on Build GRCh38
rs1055339007 Chr.22: 35656423-35656423 on Build GRCh38
rs374929762 Chr.22: 35656418-35656418 on Build GRCh38
rs375243166 Chr.22: 35656353-35656353 on Build GRCh38
rs761755664 Chr.22: 35656487-35656487 on Build GRCh38
rs994228928 Chr.22: 35656779-35656779 on Build GRCh38
rs754555812 Chr.22: 35656456-35656456 on Build GRCh38
rs978230832 Chr.22: 35656640-35656640 on Build GRCh38
rs541942069 Chr.22: 35656699-35656699 on Build GRCh38
rs762915615 Chr.22: 35656480-35656480 on Build GRCh38
rs771364780 Chr.22: 35656461-35656461 on Build GRCh38
rs780376255 Chr.22: 35656565-35656565 on Build GRCh38
rs373262943 Chr.22: 35656597-35656597 on Build GRCh38
rs747304252 Chr.22: 35656393-35656393 on Build GRCh38
rs139103148 Chr.22: 35656541-35656541 on Build GRCh38
rs748723037 Chr.22: 35656447-35656447 on Build GRCh38
rs915264937 Chr.22: 35656598-35656598 on Build GRCh38
rs369673517 Chr.22: 35656432-35656432 on Build GRCh38
rs979942998 Chr.22: 35656744-35656744 on Build GRCh38
rs559499581 Chr.22: 35656624-35656624 on Build GRCh38
rs781429777 Chr.22: 35656753-35656753 on Build GRCh38
rs1042852728 Chr.22: 35656573-35656573 on Build GRCh38
rs763615209 Chr.22: 35656354-35656354 on Build GRCh38
rs146182015 Chr.22: 35656486-35656486 on Build GRCh38
rs945515619 Chr.22: 35656556-35656556 on Build GRCh38
rs1016041191 Chr.22: 35656716-35656716 on Build GRCh38
rs373407904 Chr.22: 35656533-35656533 on Build GRCh38
rs558682248 Chr.22: 35656722-35656722 on Build GRCh38
rs763919417 Chr.22: 35656439-35656439 on Build GRCh38
rs747573264 Chr.22: 35656460-35656460 on Build GRCh38
rs755756726 Chr.22: 35656442-35656442 on Build GRCh38
rs765098918 Chr.22: 35656470-35656470 on Build GRCh38
rs771452833 Chr.22: 35656401-35656401 on Build GRCh38
rs779736731 Chr.22: 35656446-35656446 on Build GRCh38
rs900765616 Chr.22: 35656657-35656657 on Build GRCh38

View Full Product Details