Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35720560-35721107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL5
Entrez Gene ID
80831
Gene Name
apolipoprotein L5
Gene Aliases
APOL-V, APOLV
UniGene
Hs.326561
Gene Chromosome Location
Chr.22: 35708424-35730279 on Build GRCh38
Transcripts
NM_030642.1 XM_006724321.3 XM_017028945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5926137 c.81G>T p.M27I Chr.22: 35720593-35720593 on Build GRCh38
3149323 c.88C>T p.R30* Chr.22: 35720600-35720600 on Build GRCh38
444927 c.76G>A p.E26K Chr.22: 35720588-35720588 on Build GRCh38
1327249 c.112G>A p.V38I Chr.22: 35720624-35720624 on Build GRCh38
1033774 c.68G>T p.G23V Chr.22: 35720580-35720580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1025103766 Chr.22: 35720659-35720659 on Build GRCh38
rs748021263 Chr.22: 35720957-35720957 on Build GRCh38
rs929485124 Chr.22: 35721047-35721047 on Build GRCh38
rs567801962 Chr.22: 35720688-35720688 on Build GRCh38
rs538957260 Chr.22: 35720945-35720945 on Build GRCh38
rs1028641423 Chr.22: 35720942-35720942 on Build GRCh38
rs779810376 Chr.22: 35720597-35720597 on Build GRCh38
rs928451090 Chr.22: 35720773-35720773 on Build GRCh38
rs757956930 Chr.22: 35720653-35720653 on Build GRCh38
rs370168171 Chr.22: 35720673-35720673 on Build GRCh38
rs568136674 Chr.22: 35720600-35720600 on Build GRCh38
rs35709397 Chr.22: 35720876-35720876 on Build GRCh38
rs562567171 Chr.22: 35720935-35720935 on Build GRCh38
rs1002593969 Chr.22: 35720907-35720907 on Build GRCh38
rs149294583 Chr.22: 35720647-35720647 on Build GRCh38
rs1035948562 Chr.22: 35720924-35720924 on Build GRCh38
rs1026676800 Chr.22: 35720724-35720724 on Build GRCh38
rs921081646 Chr.22: 35720809-35720809 on Build GRCh38
rs982408926 Chr.22: 35720736-35720741 on Build GRCh38
rs910523782 Chr.22: 35721007-35721007 on Build GRCh38
rs368877696 Chr.22: 35720666-35720666 on Build GRCh38
rs952486842 Chr.22: 35720734-35720734 on Build GRCh38
rs749176300 Chr.22: 35720672-35720672 on Build GRCh38
rs182057020 Chr.22: 35720655-35720655 on Build GRCh38
rs1005890192 Chr.22: 35720895-35720895 on Build GRCh38
rs763071253 Chr.22: 35720634-35720634 on Build GRCh38
rs757403130 Chr.22: 35720644-35720644 on Build GRCh38
rs191392031 Chr.22: 35721036-35721036 on Build GRCh38
rs543577613 Chr.22: 35720902-35720902 on Build GRCh38
rs868121277 Chr.22: 35720871-35720871 on Build GRCh38
rs965290559 Chr.22: 35720798-35720798 on Build GRCh38
rs545087435 Chr.22: 35720816-35720816 on Build GRCh38
rs979410881 Chr.22: 35720998-35720998 on Build GRCh38
rs529854100 Chr.22: 35720894-35720894 on Build GRCh38
rs779661935 Chr.22: 35720725-35720725 on Build GRCh38
rs565380661 Chr.22: 35720993-35720993 on Build GRCh38
rs889311918 Chr.22: 35720612-35720612 on Build GRCh38
rs375308767 Chr.22: 35720903-35720903 on Build GRCh38
rs1022716295 Chr.22: 35720721-35720721 on Build GRCh38
rs888617393 Chr.22: 35721069-35721069 on Build GRCh38
rs570364677 Chr.22: 35720906-35720906 on Build GRCh38
rs977076528 Chr.22: 35720807-35720807 on Build GRCh38
rs552749917 Chr.22: 35720815-35720815 on Build GRCh38
rs954027605 Chr.22: 35720943-35720943 on Build GRCh38
rs988244906 Chr.22: 35720996-35720996 on Build GRCh38
rs968060109 Chr.22: 35720997-35720997 on Build GRCh38
rs59436568 Chr.22: 35720763-35720764 on Build GRCh38
rs1042934997 Chr.22: 35720819-35720819 on Build GRCh38
rs754559059 Chr.22: 35720679-35720679 on Build GRCh38
rs777145664 Chr.22: 35720628-35720628 on Build GRCh38
rs965434282 Chr.22: 35720920-35720921 on Build GRCh38
rs564828835 Chr.22: 35720892-35720892 on Build GRCh38
rs771644523 Chr.22: 35720687-35720687 on Build GRCh38
rs771320900 Chr.22: 35720626-35720626 on Build GRCh38
rs559025443 Chr.22: 35720824-35720824 on Build GRCh38
rs759856437 Chr.22: 35720629-35720629 on Build GRCh38
rs374963485 Chr.22: 35720685-35720685 on Build GRCh38
rs547887765 Chr.22: 35721035-35721035 on Build GRCh38
rs910640011 Chr.22: 35720870-35720870 on Build GRCh38
rs936702748 Chr.22: 35720965-35720965 on Build GRCh38
rs187778230 Chr.22: 35720745-35720745 on Build GRCh38
rs938297657 Chr.22: 35721051-35721051 on Build GRCh38
rs957611244 Chr.22: 35720989-35720989 on Build GRCh38
rs768415716 Chr.22: 35720601-35720601 on Build GRCh38
rs925357067 Chr.22: 35720826-35720826 on Build GRCh38
rs565323452 Chr.22: 35720630-35720630 on Build GRCh38
rs763585623 Chr.22: 35720768-35720769 on Build GRCh38
rs369749894 Chr.22: 35720632-35720632 on Build GRCh38
rs147667623 Chr.22: 35720890-35720890 on Build GRCh38
rs779984317 Chr.22: 35720657-35720657 on Build GRCh38
rs892467204 Chr.22: 35720863-35720863 on Build GRCh38
rs139593449 Chr.22: 35720615-35720615 on Build GRCh38
rs769730762 Chr.22: 35720588-35720588 on Build GRCh38
rs1052403440 Chr.22: 35720859-35720859 on Build GRCh38
rs367616849 Chr.22: 35721014-35721014 on Build GRCh38
rs556682754 Chr.22: 35721059-35721059 on Build GRCh38
rs573641837 Chr.22: 35720808-35720808 on Build GRCh38
rs147206206 Chr.22: 35720614-35720614 on Build GRCh38
rs140020632 Chr.22: 35720844-35720844 on Build GRCh38
rs913876253 Chr.22: 35720999-35720999 on Build GRCh38
rs777057908 Chr.22: 35720693-35720693 on Build GRCh38
rs988720197 Chr.22: 35720814-35720814 on Build GRCh38
rs913913588 Chr.22: 35720817-35720817 on Build GRCh38
rs183149604 Chr.22: 35721054-35721054 on Build GRCh38
rs1015174262 Chr.22: 35720918-35720918 on Build GRCh38
rs1047101647 Chr.22: 35720878-35720878 on Build GRCh38
rs993736490 Chr.22: 35720645-35720645 on Build GRCh38
rs773858698 Chr.22: 35720684-35720684 on Build GRCh38
rs750413929 Chr.22: 35720654-35720654 on Build GRCh38
rs994181596 Chr.22: 35720891-35720891 on Build GRCh38
rs137974900 Chr.22: 35720747-35720748 on Build GRCh38

View Full Product Details