Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 35728643-35729146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOL5
Entrez Gene ID
80831
Gene Name
apolipoprotein L5
Gene Aliases
APOL-V, APOLV
UniGene
Hs.326561
Gene Chromosome Location
Chr.22: 35708424-35730279 on Build GRCh38
Transcripts
NM_030642.1 XM_006724321.3 XM_017028945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1033782 c.1167G>T p.Q389H Chr.22: 35728763-35728763 on Build GRCh38
5900838 c.1249C>T p.P417S Chr.22: 35728845-35728845 on Build GRCh38
1161424 c.1144C>T p.P382S Chr.22: 35728740-35728740 on Build GRCh38
7428081 c.1217C>G p.S406C Chr.22: 35728813-35728813 on Build GRCh38
5526814 c.1272G>A p.G424G Chr.22: 35728868-35728868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750219467 Chr.22: 35728677-35728677 on Build GRCh38
rs779506754 Chr.22: 35728688-35728688 on Build GRCh38
rs774540854 Chr.22: 35728747-35728747 on Build GRCh38
rs752353710 Chr.22: 35728880-35728880 on Build GRCh38
rs761942950 Chr.22: 35728834-35728834 on Build GRCh38
rs768827112 Chr.22: 35728733-35728733 on Build GRCh38
rs752157146 Chr.22: 35728796-35728796 on Build GRCh38
rs867564567 Chr.22: 35728823-35728823 on Build GRCh38
rs974139899 Chr.22: 35729007-35729007 on Build GRCh38
rs767603160 Chr.22: 35728675-35728675 on Build GRCh38
rs766268094 Chr.22: 35728868-35728868 on Build GRCh38
rs753644173 Chr.22: 35728781-35728781 on Build GRCh38
rs778576283 Chr.22: 35728746-35728746 on Build GRCh38
rs759275735 Chr.22: 35728878-35728878 on Build GRCh38
rs918700063 Chr.22: 35729007-35729007 on Build GRCh38
rs180727140 Chr.22: 35728941-35728941 on Build GRCh38
rs760656500 Chr.22: 35728858-35728858 on Build GRCh38
rs779220439 Chr.22: 35728911-35728911 on Build GRCh38
rs75305023 Chr.22: 35728843-35728843 on Build GRCh38
rs757370353 Chr.22: 35728740-35728740 on Build GRCh38
rs890841017 Chr.22: 35728793-35728793 on Build GRCh38
rs781681932 Chr.22: 35728717-35728717 on Build GRCh38
rs557900035 Chr.22: 35729050-35729050 on Build GRCh38
rs1055111263 Chr.22: 35728956-35728956 on Build GRCh38
rs7511207 Chr.22: 35728992-35728992 on Build GRCh38
rs114129965 Chr.22: 35729068-35729068 on Build GRCh38
rs778391053 Chr.22: 35728711-35728711 on Build GRCh38
rs779255435 Chr.22: 35728831-35728832 on Build GRCh38
rs755423266 Chr.22: 35728904-35728904 on Build GRCh38
rs13053994 Chr.22: 35728982-35728982 on Build GRCh38
rs776467421 Chr.22: 35728938-35728938 on Build GRCh38
rs148906900 Chr.22: 35728849-35728849 on Build GRCh38
rs746397810 Chr.22: 35728806-35728806 on Build GRCh38
rs932309789 Chr.22: 35729001-35729001 on Build GRCh38
rs145495755 Chr.22: 35728728-35728728 on Build GRCh38
rs917453500 Chr.22: 35728687-35728687 on Build GRCh38
rs767511524 Chr.22: 35728757-35728757 on Build GRCh38
rs577843601 Chr.22: 35729051-35729051 on Build GRCh38
rs747699892 Chr.22: 35728712-35728712 on Build GRCh38
rs148034663 Chr.22: 35728891-35728891 on Build GRCh38
rs769800211 Chr.22: 35728934-35728944 on Build GRCh38
rs758801359 Chr.22: 35728893-35728893 on Build GRCh38
rs753544492 Chr.22: 35728706-35728706 on Build GRCh38
rs753648908 Chr.22: 35728869-35728869 on Build GRCh38
rs778591084 Chr.22: 35728791-35728791 on Build GRCh38
rs13054309 Chr.22: 35728996-35728996 on Build GRCh38
rs147690523 Chr.22: 35728749-35728749 on Build GRCh38
rs201193967 Chr.22: 35728832-35728832 on Build GRCh38
rs765027182 Chr.22: 35728944-35728944 on Build GRCh38
rs3885449 Chr.22: 35729094-35729094 on Build GRCh38
rs757843541 Chr.22: 35728889-35728889 on Build GRCh38
rs772489546 Chr.22: 35728915-35728915 on Build GRCh38
rs950220870 Chr.22: 35728709-35728709 on Build GRCh38
rs757937833 Chr.22: 35728799-35728799 on Build GRCh38
rs977636919 Chr.22: 35728921-35728921 on Build GRCh38
rs142361344 Chr.22: 35728763-35728763 on Build GRCh38
rs201846595 Chr.22: 35728786-35728786 on Build GRCh38
rs370966999 Chr.22: 35728800-35728800 on Build GRCh38
rs929960725 Chr.22: 35729008-35729008 on Build GRCh38
rs2076673 Chr.22: 35728813-35728813 on Build GRCh38
rs893319266 Chr.22: 35729077-35729077 on Build GRCh38
rs752841163 Chr.22: 35728840-35728841 on Build GRCh38
rs1051132417 Chr.22: 35729044-35729044 on Build GRCh38
rs111838714 Chr.22: 35728935-35728935 on Build GRCh38
rs79062836 Chr.22: 35728669-35728669 on Build GRCh38
rs537813444 Chr.22: 35729043-35729043 on Build GRCh38
rs920972656 Chr.22: 35728980-35728980 on Build GRCh38
rs898315221 Chr.22: 35729083-35729083 on Build GRCh38
rs766183368 Chr.22: 35728771-35728771 on Build GRCh38
rs770196932 Chr.22: 35728726-35728726 on Build GRCh38
rs903449763 Chr.22: 35728745-35728745 on Build GRCh38
rs780458091 Chr.22: 35728821-35728821 on Build GRCh38
rs200999318 Chr.22: 35728769-35728769 on Build GRCh38
rs777960363 Chr.22: 35728868-35728869 on Build GRCh38
rs201960531 Chr.22: 35728916-35728916 on Build GRCh38
rs1004349438 Chr.22: 35729061-35729061 on Build GRCh38
rs772292828 Chr.22: 35728848-35728848 on Build GRCh38
rs755958134 Chr.22: 35728685-35728685 on Build GRCh38
rs200371039 Chr.22: 35728831-35728831 on Build GRCh38
rs560362940 Chr.22: 35728669-35728669 on Build GRCh38
rs764832192 Chr.22: 35728879-35728879 on Build GRCh38
rs748369776 Chr.22: 35728913-35728913 on Build GRCh38
rs371512315 Chr.22: 35728673-35728673 on Build GRCh38
rs771410087 Chr.22: 35728713-35728713 on Build GRCh38
rs745827837 Chr.22: 35728928-35728931 on Build GRCh38
rs775320169 Chr.22: 35728949-35728949 on Build GRCh38
rs936296192 Chr.22: 35728953-35728953 on Build GRCh38
rs143581523 Chr.22: 35728890-35728890 on Build GRCh38
rs13054304 Chr.22: 35728986-35728986 on Build GRCh38
rs754644924 Chr.22: 35728707-35728707 on Build GRCh38
rs749613751 Chr.22: 35728899-35728899 on Build GRCh38
rs763219644 Chr.22: 35728729-35728729 on Build GRCh38
rs398102316 Chr.22: 35728846-35728846 on Build GRCh38
rs746193988 Chr.22: 35728723-35728723 on Build GRCh38
rs749653865 Chr.22: 35728826-35728826 on Build GRCh38

View Full Product Details