Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36520509-36520951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3D
Entrez Gene ID
8664
Gene Name
eukaryotic translation initiation factor 3 subunit D
Gene Aliases
EIF3S7, eIF3-p66, eIF3-zeta
UniGene
Hs.55682
Gene Chromosome Location
Chr.22: 36510850-36529431 on Build GRCh38
Transcripts
NM_003753.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1033868 c.575A>G p.D192G Chr.22: 36520579-36520579 on Build GRCh38
4945237 c.499G>A p.D167N Chr.22: 36520655-36520655 on Build GRCh38
280755 c.493C>T p.R165C Chr.22: 36520661-36520661 on Build GRCh38
149287 c.548G>A p.R183H Chr.22: 36520606-36520606 on Build GRCh38
5057801 c.515A>C p.E172A Chr.22: 36520639-36520639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920114849 Chr.22: 36520800-36520800 on Build GRCh38
rs949354482 Chr.22: 36520839-36520839 on Build GRCh38
rs528288783 Chr.22: 36520745-36520745 on Build GRCh38
rs761592759 Chr.22: 36520534-36520534 on Build GRCh38
rs538981018 Chr.22: 36520873-36520873 on Build GRCh38
rs954374862 Chr.22: 36520678-36520678 on Build GRCh38
rs189105765 Chr.22: 36520837-36520837 on Build GRCh38
rs966877858 Chr.22: 36520548-36520548 on Build GRCh38
rs749458696 Chr.22: 36520606-36520606 on Build GRCh38
rs570032520 Chr.22: 36520848-36520848 on Build GRCh38
rs778011146 Chr.22: 36520712-36520712 on Build GRCh38
rs1028732484 Chr.22: 36520660-36520660 on Build GRCh38
rs568025233 Chr.22: 36520796-36520796 on Build GRCh38
rs774413017 Chr.22: 36520623-36520623 on Build GRCh38
rs757689482 Chr.22: 36520568-36520568 on Build GRCh38
rs530534397 Chr.22: 36520797-36520797 on Build GRCh38
rs765988002 Chr.22: 36520792-36520792 on Build GRCh38
rs765039092 Chr.22: 36520723-36520723 on Build GRCh38
rs762380098 Chr.22: 36520788-36520788 on Build GRCh38
rs957064298 Chr.22: 36520903-36520903 on Build GRCh38
rs764589961 Chr.22: 36520708-36520708 on Build GRCh38
rs890892419 Chr.22: 36520830-36520830 on Build GRCh38
rs41283217 Chr.22: 36520706-36520706 on Build GRCh38
rs771870770 Chr.22: 36520661-36520661 on Build GRCh38
rs762013498 Chr.22: 36520642-36520642 on Build GRCh38
rs184984194 Chr.22: 36520793-36520793 on Build GRCh38
rs377313757 Chr.22: 36520732-36520732 on Build GRCh38
rs768874987 Chr.22: 36520619-36520619 on Build GRCh38
rs116625095 Chr.22: 36520915-36520915 on Build GRCh38
rs747192958 Chr.22: 36520718-36520719 on Build GRCh38
rs773157607 Chr.22: 36520664-36520664 on Build GRCh38
rs760548858 Chr.22: 36520695-36520695 on Build GRCh38
rs755974436 Chr.22: 36520537-36520537 on Build GRCh38
rs865898513 Chr.22: 36520625-36520625 on Build GRCh38
rs778498772 Chr.22: 36520545-36520545 on Build GRCh38
rs201648054 Chr.22: 36520601-36520601 on Build GRCh38
rs751200789 Chr.22: 36520881-36520881 on Build GRCh38
rs780419646 Chr.22: 36520603-36520603 on Build GRCh38
rs781748190 Chr.22: 36520582-36520582 on Build GRCh38
rs747725480 Chr.22: 36520560-36520560 on Build GRCh38
rs753487950 Chr.22: 36520538-36520538 on Build GRCh38
rs138824472 Chr.22: 36520833-36520833 on Build GRCh38
rs752231322 Chr.22: 36520726-36520726 on Build GRCh38
rs546102849 Chr.22: 36520704-36520704 on Build GRCh38
rs990902590 Chr.22: 36520822-36520822 on Build GRCh38
rs759256340 Chr.22: 36520719-36520719 on Build GRCh38
rs993993898 Chr.22: 36520811-36520811 on Build GRCh38
rs931504757 Chr.22: 36520807-36520807 on Build GRCh38
rs941251182 Chr.22: 36520730-36520730 on Build GRCh38
rs754690445 Chr.22: 36520542-36520542 on Build GRCh38
rs201042002 Chr.22: 36520585-36520585 on Build GRCh38
rs373238372 Chr.22: 36520536-36520536 on Build GRCh38
rs1028680120 Chr.22: 36520924-36520924 on Build GRCh38
rs1055564891 Chr.22: 36520701-36520701 on Build GRCh38
rs371225142 Chr.22: 36520714-36520714 on Build GRCh38
rs569491972 Chr.22: 36520646-36520646 on Build GRCh38
rs1018335378 Chr.22: 36520862-36520862 on Build GRCh38
rs750008894 Chr.22: 36520784-36520784 on Build GRCh38
rs987120499 Chr.22: 36520686-36520686 on Build GRCh38
rs995889095 Chr.22: 36520555-36520555 on Build GRCh38
rs1053995074 Chr.22: 36520904-36520904 on Build GRCh38
rs893631458 Chr.22: 36520914-36520914 on Build GRCh38

View Full Product Details