Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36523817-36524322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3D
Entrez Gene ID
8664
Gene Name
eukaryotic translation initiation factor 3 subunit D
Gene Aliases
EIF3S7, eIF3-p66, eIF3-zeta
UniGene
Hs.55682
Gene Chromosome Location
Chr.22: 36510850-36529431 on Build GRCh38
Transcripts
NM_003753.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5909877 c.331C>T p.R111C Chr.22: 36523956-36523956 on Build GRCh38
3149811 c.316C>T p.R106C Chr.22: 36523971-36523971 on Build GRCh38
3149807 c.355C>A p.L119M Chr.22: 36523932-36523932 on Build GRCh38
5909876 c.331C>T p.R111C Chr.22: 36523956-36523956 on Build GRCh38
3149805 c.391A>G p.R131G Chr.22: 36523896-36523896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183157472 Chr.22: 36524144-36524144 on Build GRCh38
rs12157889 Chr.22: 36524015-36524015 on Build GRCh38
rs769859450 Chr.22: 36523878-36523878 on Build GRCh38
rs372458185 Chr.22: 36524016-36524016 on Build GRCh38
rs940569024 Chr.22: 36524079-36524079 on Build GRCh38
rs942577636 Chr.22: 36524032-36524032 on Build GRCh38
rs370073245 Chr.22: 36524021-36524021 on Build GRCh38
rs749190705 Chr.22: 36523847-36523847 on Build GRCh38
rs146392043 Chr.22: 36523967-36523967 on Build GRCh38
rs975912965 Chr.22: 36524240-36524240 on Build GRCh38
rs931738172 Chr.22: 36524101-36524101 on Build GRCh38
rs952052411 Chr.22: 36524169-36524169 on Build GRCh38
rs373842275 Chr.22: 36523847-36523847 on Build GRCh38
rs138662362 Chr.22: 36524143-36524143 on Build GRCh38
rs749099879 Chr.22: 36523939-36523939 on Build GRCh38
rs766952896 Chr.22: 36523970-36523970 on Build GRCh38
rs750601737 Chr.22: 36524023-36524023 on Build GRCh38
rs761474679 Chr.22: 36523955-36523955 on Build GRCh38
rs149044977 Chr.22: 36524172-36524172 on Build GRCh38
rs749082358 Chr.22: 36524031-36524031 on Build GRCh38
rs55889583 Chr.22: 36524017-36524017 on Build GRCh38
rs1034412949 Chr.22: 36524171-36524171 on Build GRCh38
rs1000416121 Chr.22: 36524120-36524120 on Build GRCh38
rs73411765 Chr.22: 36523848-36523848 on Build GRCh38
rs538598235 Chr.22: 36524147-36524147 on Build GRCh38
rs540553746 Chr.22: 36524197-36524197 on Build GRCh38
rs907836313 Chr.22: 36524206-36524206 on Build GRCh38
rs879697684 Chr.22: 36524141-36524141 on Build GRCh38
rs376193132 Chr.22: 36523992-36523992 on Build GRCh38
rs753082851 Chr.22: 36524005-36524005 on Build GRCh38
rs1024433978 Chr.22: 36523889-36523889 on Build GRCh38
rs757373637 Chr.22: 36523866-36523866 on Build GRCh38
rs777904466 Chr.22: 36523861-36523861 on Build GRCh38
rs776354644 Chr.22: 36524097-36524097 on Build GRCh38
rs747053431 Chr.22: 36523865-36523865 on Build GRCh38
rs750092095 Chr.22: 36524147-36524147 on Build GRCh38
rs770219542 Chr.22: 36524128-36524128 on Build GRCh38
rs1001730197 Chr.22: 36524163-36524163 on Build GRCh38
rs779907626 Chr.22: 36524026-36524026 on Build GRCh38
rs1005128577 Chr.22: 36524134-36524134 on Build GRCh38
rs534377146 Chr.22: 36524050-36524050 on Build GRCh38
rs143180024 Chr.22: 36524272-36524272 on Build GRCh38
rs765720474 Chr.22: 36524001-36524001 on Build GRCh38
rs201208549 Chr.22: 36523881-36523881 on Build GRCh38
rs12166028 Chr.22: 36524153-36524153 on Build GRCh38
rs903523350 Chr.22: 36524119-36524119 on Build GRCh38
rs967386995 Chr.22: 36524289-36524289 on Build GRCh38
rs765567899 Chr.22: 36523853-36523853 on Build GRCh38
rs773917881 Chr.22: 36523953-36523953 on Build GRCh38
rs925781504 Chr.22: 36524022-36524022 on Build GRCh38
rs760028301 Chr.22: 36523990-36523990 on Build GRCh38
rs1047172160 Chr.22: 36524118-36524118 on Build GRCh38
rs768334221 Chr.22: 36523944-36523944 on Build GRCh38
rs766563789 Chr.22: 36524162-36524162 on Build GRCh38
rs577705308 Chr.22: 36524100-36524100 on Build GRCh38
rs751231210 Chr.22: 36524143-36524143 on Build GRCh38
rs745742319 Chr.22: 36523873-36523873 on Build GRCh38
rs371704479 Chr.22: 36523986-36523986 on Build GRCh38
rs781446277 Chr.22: 36523871-36523871 on Build GRCh38
rs993253597 Chr.22: 36523924-36523924 on Build GRCh38
rs758633182 Chr.22: 36523860-36523860 on Build GRCh38
rs879030014 Chr.22: 36524142-36524142 on Build GRCh38
rs962167495 Chr.22: 36524224-36524224 on Build GRCh38
rs768709902 Chr.22: 36523983-36523983 on Build GRCh38
rs12165971 Chr.22: 36524052-36524052 on Build GRCh38

View Full Product Details