Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36561952-36562453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG2
Entrez Gene ID
10369
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 2
Gene Aliases
MRD10
UniGene
Hs.146766
Gene Chromosome Location
Chr.22: 36560869-36702645 on Build GRCh38
Transcripts
NM_006078.3 XM_017028531.1

SNP Detail

SNP ID Chromosome Location
rs1020800798 Chr.22: 36562136-36562136 on Build GRCh38
rs1041908868 Chr.22: 36562171-36562171 on Build GRCh38
rs1007871170 Chr.22: 36562062-36562062 on Build GRCh38
rs973842380 Chr.22: 36562350-36562350 on Build GRCh38
rs933415749 Chr.22: 36561995-36561995 on Build GRCh38
rs1009493028 Chr.22: 36562331-36562331 on Build GRCh38
rs896071498 Chr.22: 36562119-36562119 on Build GRCh38
rs571434211 Chr.22: 36562082-36562082 on Build GRCh38
rs764975190 Chr.22: 36562426-36562426 on Build GRCh38
rs73169693 Chr.22: 36562021-36562021 on Build GRCh38
rs142179335 Chr.22: 36562197-36562198 on Build GRCh38
rs933384467 Chr.22: 36562244-36562244 on Build GRCh38
rs982997829 Chr.22: 36562287-36562287 on Build GRCh38
rs775453133 Chr.22: 36562212-36562212 on Build GRCh38
rs73413764 Chr.22: 36562318-36562318 on Build GRCh38
rs950264008 Chr.22: 36562411-36562411 on Build GRCh38
rs560419751 Chr.22: 36562433-36562433 on Build GRCh38
rs187316728 Chr.22: 36562004-36562004 on Build GRCh38
rs917475091 Chr.22: 36562383-36562383 on Build GRCh38
rs561883297 Chr.22: 36562264-36562264 on Build GRCh38
rs759355845 Chr.22: 36562139-36562139 on Build GRCh38
rs756814994 Chr.22: 36562418-36562418 on Build GRCh38
rs992145765 Chr.22: 36562198-36562201 on Build GRCh38
rs941137246 Chr.22: 36562064-36562064 on Build GRCh38
rs7290792 Chr.22: 36562087-36562087 on Build GRCh38
rs909507006 Chr.22: 36562198-36562198 on Build GRCh38
rs753405842 Chr.22: 36562304-36562304 on Build GRCh38
rs796602992 Chr.22: 36562060-36562060 on Build GRCh38
rs544467547 Chr.22: 36562234-36562234 on Build GRCh38
rs899841837 Chr.22: 36562228-36562228 on Build GRCh38
rs908259284 Chr.22: 36562042-36562042 on Build GRCh38
rs866375502 Chr.22: 36562103-36562103 on Build GRCh38
rs937618830 Chr.22: 36562371-36562371 on Build GRCh38
rs112678315 Chr.22: 36562093-36562093 on Build GRCh38
rs1056052794 Chr.22: 36562106-36562106 on Build GRCh38
rs567607889 Chr.22: 36562035-36562035 on Build GRCh38
rs1000274261 Chr.22: 36562173-36562173 on Build GRCh38
rs535003460 Chr.22: 36562419-36562419 on Build GRCh38
rs573665875 Chr.22: 36562160-36562160 on Build GRCh38
rs897876013 Chr.22: 36561990-36561990 on Build GRCh38
rs572640538 Chr.22: 36562362-36562363 on Build GRCh38
rs189109844 Chr.22: 36562195-36562195 on Build GRCh38
rs796507900 Chr.22: 36562198-36562199 on Build GRCh38
rs575855301 Chr.22: 36562196-36562196 on Build GRCh38
rs1003283175 Chr.22: 36562406-36562406 on Build GRCh38
rs1015408618 Chr.22: 36562288-36562288 on Build GRCh38
rs1020769388 Chr.22: 36562343-36562343 on Build GRCh38
rs535914404 Chr.22: 36562172-36562172 on Build GRCh38
rs926396962 Chr.22: 36562367-36562367 on Build GRCh38
rs981868666 Chr.22: 36562150-36562150 on Build GRCh38
rs185800927 Chr.22: 36562265-36562265 on Build GRCh38
rs991830715 Chr.22: 36562382-36562382 on Build GRCh38
rs193046040 Chr.22: 36562235-36562235 on Build GRCh38
rs1051879918 Chr.22: 36562248-36562248 on Build GRCh38
rs906390563 Chr.22: 36562359-36562359 on Build GRCh38

View Full Product Details