Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37367871-37368370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELFN2
Entrez Gene ID
114794
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 2
Gene Aliases
LRRC62, PPP1R29
UniGene
Hs.344170
Gene Chromosome Location
Chr.22: 37340644-37427470 on Build GRCh38
Transcripts
NM_052906.4 NR_110512.1

SNP Detail

SNP ID Chromosome Location
rs935993328 Chr.22: 37367907-37367907 on Build GRCh38
rs151209180 Chr.22: 37368310-37368310 on Build GRCh38
rs999933783 Chr.22: 37367908-37367908 on Build GRCh38
rs6000692 Chr.22: 37368173-37368173 on Build GRCh38
rs763001525 Chr.22: 37368331-37368331 on Build GRCh38
rs1005588659 Chr.22: 37368035-37368035 on Build GRCh38
rs528004862 Chr.22: 37368196-37368196 on Build GRCh38
rs967172715 Chr.22: 37368120-37368120 on Build GRCh38
rs776954801 Chr.22: 37368123-37368123 on Build GRCh38
rs568617857 Chr.22: 37368311-37368311 on Build GRCh38
rs1000138849 Chr.22: 37368294-37368294 on Build GRCh38
rs916182927 Chr.22: 37368174-37368174 on Build GRCh38
rs762149591 Chr.22: 37368180-37368180 on Build GRCh38
rs911245260 Chr.22: 37368158-37368158 on Build GRCh38
rs964277776 Chr.22: 37368064-37368064 on Build GRCh38
rs3752518 Chr.22: 37368213-37368213 on Build GRCh38
rs980006029 Chr.22: 37368041-37368041 on Build GRCh38
rs1045999996 Chr.22: 37368220-37368220 on Build GRCh38
rs993946772 Chr.22: 37367932-37367932 on Build GRCh38
rs1013834746 Chr.22: 37367916-37367916 on Build GRCh38
rs909703333 Chr.22: 37368053-37368053 on Build GRCh38
rs577294134 Chr.22: 37367917-37367917 on Build GRCh38
rs1031870724 Chr.22: 37367911-37367911 on Build GRCh38
rs867831368 Chr.22: 37368189-37368189 on Build GRCh38
rs1051066819 Chr.22: 37368156-37368156 on Build GRCh38
rs115809786 Chr.22: 37368100-37368100 on Build GRCh38
rs936018761 Chr.22: 37368138-37368138 on Build GRCh38
rs369147486 Chr.22: 37368124-37368124 on Build GRCh38
rs952450300 Chr.22: 37367968-37367968 on Build GRCh38
rs1018336754 Chr.22: 37368326-37368326 on Build GRCh38
rs118187670 Chr.22: 37367912-37367912 on Build GRCh38
rs904011514 Chr.22: 37368262-37368262 on Build GRCh38
rs532003201 Chr.22: 37368299-37368299 on Build GRCh38
rs1018197193 Chr.22: 37368042-37368042 on Build GRCh38
rs76356417 Chr.22: 37368114-37368114 on Build GRCh38
rs969918529 Chr.22: 37367925-37367925 on Build GRCh38
rs62237489 Chr.22: 37367980-37367980 on Build GRCh38
rs564408460 Chr.22: 37368268-37368268 on Build GRCh38
rs904045348 Chr.22: 37367904-37367904 on Build GRCh38
rs1046848714 Chr.22: 37368295-37368295 on Build GRCh38
rs993661480 Chr.22: 37368265-37368265 on Build GRCh38
rs184498936 Chr.22: 37367953-37367953 on Build GRCh38
rs542099940 Chr.22: 37368164-37368164 on Build GRCh38
rs541133318 Chr.22: 37368004-37368004 on Build GRCh38
rs750521581 Chr.22: 37368330-37368330 on Build GRCh38

View Full Product Details