Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37373005-37373528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELFN2
Entrez Gene ID
114794
Gene Name
extracellular leucine rich repeat and fibronectin type III domain containing 2
Gene Aliases
LRRC62, PPP1R29
UniGene
Hs.344170
Gene Chromosome Location
Chr.22: 37340644-37427470 on Build GRCh38
Transcripts
NM_052906.4 NR_110512.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5967596 c.2079_2080insG p.I694fs*47 Chr.22: 37373455-37373456 on Build GRCh38
3842629 c.2434A>G p.K812E Chr.22: 37373101-37373101 on Build GRCh38
4608958 c.2133C>A p.S711R Chr.22: 37373402-37373402 on Build GRCh38
6854924 c.2325C>A p.P775P Chr.22: 37373210-37373210 on Build GRCh38
1033984 c.2114G>A p.C705Y Chr.22: 37373421-37373421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754045388 Chr.22: 37373309-37373309 on Build GRCh38
rs534286121 Chr.22: 37373212-37373212 on Build GRCh38
rs373874492 Chr.22: 37373046-37373046 on Build GRCh38
rs780348681 Chr.22: 37373163-37373163 on Build GRCh38
rs755839172 Chr.22: 37373417-37373417 on Build GRCh38
rs1005405928 Chr.22: 37373201-37373201 on Build GRCh38
rs761796191 Chr.22: 37373490-37373490 on Build GRCh38
rs750281373 Chr.22: 37373493-37373493 on Build GRCh38
rs781107766 Chr.22: 37373410-37373410 on Build GRCh38
rs904032962 Chr.22: 37373451-37373451 on Build GRCh38
rs780834506 Chr.22: 37373334-37373334 on Build GRCh38
rs544858839 Chr.22: 37373468-37373468 on Build GRCh38
rs753471829 Chr.22: 37373503-37373503 on Build GRCh38
rs148494870 Chr.22: 37373165-37373165 on Build GRCh38
rs766741566 Chr.22: 37373304-37373304 on Build GRCh38
rs751906828 Chr.22: 37373122-37373122 on Build GRCh38
rs371726939 Chr.22: 37373447-37373447 on Build GRCh38
rs745456971 Chr.22: 37373336-37373336 on Build GRCh38
rs144499416 Chr.22: 37373353-37373353 on Build GRCh38
rs776222892 Chr.22: 37373276-37373276 on Build GRCh38
rs771854807 Chr.22: 37373179-37373179 on Build GRCh38
rs962800042 Chr.22: 37373487-37373487 on Build GRCh38
rs749366380 Chr.22: 37373164-37373164 on Build GRCh38
rs983890678 Chr.22: 37373054-37373054 on Build GRCh38
rs763031057 Chr.22: 37373482-37373482 on Build GRCh38
rs770082574 Chr.22: 37373479-37373479 on Build GRCh38
rs777652287 Chr.22: 37373320-37373320 on Build GRCh38
rs759051934 Chr.22: 37373205-37373205 on Build GRCh38
rs776559734 Chr.22: 37373462-37373473 on Build GRCh38
rs550103003 Chr.22: 37373063-37373064 on Build GRCh38
rs762097569 Chr.22: 37373218-37373218 on Build GRCh38
rs752857902 Chr.22: 37373384-37373384 on Build GRCh38
rs941381736 Chr.22: 37373290-37373290 on Build GRCh38
rs368547931 Chr.22: 37373430-37373430 on Build GRCh38
rs878884083 Chr.22: 37373462-37373462 on Build GRCh38
rs768233564 Chr.22: 37373438-37373438 on Build GRCh38
rs753211802 Chr.22: 37373096-37373096 on Build GRCh38
rs573203265 Chr.22: 37373357-37373357 on Build GRCh38
rs372219343 Chr.22: 37373066-37373066 on Build GRCh38
rs767334560 Chr.22: 37373492-37373492 on Build GRCh38
rs781442509 Chr.22: 37373057-37373057 on Build GRCh38
rs1014197650 Chr.22: 37373467-37373467 on Build GRCh38
rs1051681029 Chr.22: 37373348-37373348 on Build GRCh38
rs200692409 Chr.22: 37373302-37373302 on Build GRCh38
rs778621356 Chr.22: 37373509-37373509 on Build GRCh38
rs201784349 Chr.22: 37373354-37373354 on Build GRCh38
rs747455234 Chr.22: 37373458-37373458 on Build GRCh38
rs375521751 Chr.22: 37373084-37373084 on Build GRCh38
rs888099326 Chr.22: 37373457-37373457 on Build GRCh38
rs144488226 Chr.22: 37373193-37373193 on Build GRCh38
rs199734718 Chr.22: 37373171-37373171 on Build GRCh38
rs943103344 Chr.22: 37373382-37373382 on Build GRCh38
rs779786440 Chr.22: 37373502-37373502 on Build GRCh38
rs368464304 Chr.22: 37373065-37373065 on Build GRCh38
rs776726552 Chr.22: 37373376-37373376 on Build GRCh38
rs762448443 Chr.22: 37373299-37373299 on Build GRCh38
rs894630686 Chr.22: 37373140-37373140 on Build GRCh38
rs1022436291 Chr.22: 37373445-37373445 on Build GRCh38
rs770606905 Chr.22: 37373264-37373264 on Build GRCh38
rs757599753 Chr.22: 37373123-37373123 on Build GRCh38
rs760551456 Chr.22: 37373149-37373149 on Build GRCh38
rs897326378 Chr.22: 37373071-37373071 on Build GRCh38
rs750614033 Chr.22: 37373141-37373141 on Build GRCh38
rs369540331 Chr.22: 37373215-37373215 on Build GRCh38
rs775523636 Chr.22: 37373480-37373480 on Build GRCh38
rs540600855 Chr.22: 37373433-37373433 on Build GRCh38
rs760259006 Chr.22: 37373187-37373187 on Build GRCh38
rs369093489 Chr.22: 37373038-37373038 on Build GRCh38
rs538457470 Chr.22: 37373278-37373278 on Build GRCh38
rs373783147 Chr.22: 37373183-37373183 on Build GRCh38
rs751438755 Chr.22: 37373405-37373405 on Build GRCh38
rs748063686 Chr.22: 37373250-37373250 on Build GRCh38
rs769378319 Chr.22: 37373285-37373285 on Build GRCh38
rs758428953 Chr.22: 37373395-37373395 on Build GRCh38
rs1037321628 Chr.22: 37373070-37373070 on Build GRCh38
rs764038571 Chr.22: 37373396-37373396 on Build GRCh38
rs969952157 Chr.22: 37373470-37373470 on Build GRCh38
rs755029989 Chr.22: 37373310-37373310 on Build GRCh38
rs763423434 Chr.22: 37373106-37373106 on Build GRCh38
rs141641460 Chr.22: 37373220-37373220 on Build GRCh38
rs755815232 Chr.22: 37373501-37373501 on Build GRCh38
rs368881782 Chr.22: 37373383-37373383 on Build GRCh38
rs756462935 Chr.22: 37373237-37373237 on Build GRCh38
rs752718592 Chr.22: 37373319-37373319 on Build GRCh38
rs150497338 Chr.22: 37373252-37373252 on Build GRCh38
rs761145702 Chr.22: 37373371-37373371 on Build GRCh38
rs750812504 Chr.22: 37373221-37373221 on Build GRCh38
rs761090800 Chr.22: 37373303-37373303 on Build GRCh38
rs148420515 Chr.22: 37373195-37373195 on Build GRCh38
rs771563240 Chr.22: 37373461-37373461 on Build GRCh38
rs760203893 Chr.22: 37373085-37373085 on Build GRCh38
rs746833926 Chr.22: 37373329-37373329 on Build GRCh38
rs766794867 Chr.22: 37373372-37373372 on Build GRCh38
rs780401854 Chr.22: 37373238-37373238 on Build GRCh38
rs755047462 Chr.22: 37373246-37373246 on Build GRCh38
rs768670879 Chr.22: 37373064-37373064 on Build GRCh38
rs764636886 Chr.22: 37373117-37373117 on Build GRCh38
rs376648337 Chr.22: 37373508-37373508 on Build GRCh38
rs368759580 Chr.22: 37373158-37373158 on Build GRCh38
rs754011945 Chr.22: 37373245-37373245 on Build GRCh38
rs994529397 Chr.22: 37373031-37373032 on Build GRCh38
rs781290326 Chr.22: 37373132-37373132 on Build GRCh38
rs778062441 Chr.22: 37373027-37373027 on Build GRCh38
rs577369562 Chr.22: 37373455-37373455 on Build GRCh38
rs200736226 Chr.22: 37373337-37373337 on Build GRCh38
rs956009997 Chr.22: 37373474-37373474 on Build GRCh38
rs146137658 Chr.22: 37373356-37373356 on Build GRCh38
rs1025517734 Chr.22: 37373037-37373037 on Build GRCh38
rs745636836 Chr.22: 37373412-37373412 on Build GRCh38
rs1031513097 Chr.22: 37373208-37373208 on Build GRCh38
rs569458468 Chr.22: 37373069-37373069 on Build GRCh38
rs538751449 Chr.22: 37373174-37373174 on Build GRCh38
rs765298109 Chr.22: 37373315-37373315 on Build GRCh38
rs756995270 Chr.22: 37373331-37373331 on Build GRCh38
rs769288474 Chr.22: 37373283-37373283 on Build GRCh38
rs770845915 Chr.22: 37373290-37373291 on Build GRCh38
rs759784030 Chr.22: 37373471-37373471 on Build GRCh38
rs61737924 Chr.22: 37373089-37373089 on Build GRCh38
rs200269334 Chr.22: 37373090-37373090 on Build GRCh38
rs200730758 Chr.22: 37373291-37373291 on Build GRCh38
rs139416895 Chr.22: 37373258-37373258 on Build GRCh38
rs757048659 Chr.22: 37373406-37373406 on Build GRCh38
rs373287422 Chr.22: 37373489-37373489 on Build GRCh38

View Full Product Details