Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37492560-37492959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD10
Entrez Gene ID
29775
Gene Name
caspase recruitment domain family member 10
Gene Aliases
BIMP1, CARMA3
UniGene
Hs.57973
Gene Chromosome Location
Chr.22: 37490362-37519203 on Build GRCh38
Transcripts
NM_014550.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4929681 c.2608C>T p.R870W Chr.22: 37492671-37492671 on Build GRCh38
6620056 c.2567C>T p.A856V Chr.22: 37492712-37492712 on Build GRCh38
1638060 c.2542G>A p.V848M Chr.22: 37492737-37492737 on Build GRCh38
6400328 c.1636A>G p.R546G Chr.22: 37492785-37492785 on Build GRCh38
4137432 c.2619C>A p.F873L Chr.22: 37492660-37492660 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375580111 Chr.22: 37492820-37492820 on Build GRCh38
rs377599502 Chr.22: 37492670-37492670 on Build GRCh38
rs1047056068 Chr.22: 37492900-37492900 on Build GRCh38
rs183467601 Chr.22: 37492845-37492845 on Build GRCh38
rs368352079 Chr.22: 37492799-37492799 on Build GRCh38
rs550505544 Chr.22: 37492588-37492588 on Build GRCh38
rs762848305 Chr.22: 37492796-37492796 on Build GRCh38
rs769750011 Chr.22: 37492793-37492793 on Build GRCh38
rs777349583 Chr.22: 37492635-37492635 on Build GRCh38
rs372431956 Chr.22: 37492738-37492738 on Build GRCh38
rs776727513 Chr.22: 37492706-37492706 on Build GRCh38
rs992487317 Chr.22: 37492634-37492635 on Build GRCh38
rs368566607 Chr.22: 37492821-37492821 on Build GRCh38
rs753293301 Chr.22: 37492830-37492830 on Build GRCh38
rs755453339 Chr.22: 37492743-37492743 on Build GRCh38
rs971814184 Chr.22: 37492918-37492918 on Build GRCh38
rs750030910 Chr.22: 37492816-37492816 on Build GRCh38
rs779530759 Chr.22: 37492746-37492746 on Build GRCh38
rs906710312 Chr.22: 37492929-37492929 on Build GRCh38
rs746651749 Chr.22: 37492638-37492638 on Build GRCh38
rs369567435 Chr.22: 37492656-37492656 on Build GRCh38
rs757570334 Chr.22: 37492854-37492854 on Build GRCh38
rs768464083 Chr.22: 37492615-37492615 on Build GRCh38
rs201794655 Chr.22: 37492794-37492794 on Build GRCh38
rs541283118 Chr.22: 37492841-37492841 on Build GRCh38
rs377214071 Chr.22: 37492685-37492685 on Build GRCh38
rs776581731 Chr.22: 37492782-37492782 on Build GRCh38
rs771240209 Chr.22: 37492780-37492780 on Build GRCh38
rs529758086 Chr.22: 37492628-37492628 on Build GRCh38
rs369846218 Chr.22: 37492930-37492931 on Build GRCh38
rs774583145 Chr.22: 37492676-37492676 on Build GRCh38
rs771180831 Chr.22: 37492699-37492699 on Build GRCh38
rs766558007 Chr.22: 37492623-37492623 on Build GRCh38
rs374574746 Chr.22: 37492678-37492678 on Build GRCh38
rs373917622 Chr.22: 37492595-37492595 on Build GRCh38
rs1040372062 Chr.22: 37492767-37492767 on Build GRCh38
rs757995738 Chr.22: 37492630-37492630 on Build GRCh38
rs148113688 Chr.22: 37492646-37492646 on Build GRCh38
rs774727612 Chr.22: 37492594-37492594 on Build GRCh38
rs572669921 Chr.22: 37492912-37492912 on Build GRCh38
rs371884305 Chr.22: 37492809-37492809 on Build GRCh38
rs746146158 Chr.22: 37492880-37492880 on Build GRCh38
rs199702373 Chr.22: 37492587-37492587 on Build GRCh38
rs201027754 Chr.22: 37492629-37492629 on Build GRCh38
rs200672370 Chr.22: 37492772-37492772 on Build GRCh38
rs749799640 Chr.22: 37492589-37492589 on Build GRCh38
rs945666761 Chr.22: 37492886-37492886 on Build GRCh38
rs1000038688 Chr.22: 37492800-37492800 on Build GRCh38
rs767098017 Chr.22: 37492811-37492811 on Build GRCh38
rs767712147 Chr.22: 37492605-37492605 on Build GRCh38
rs1009641311 Chr.22: 37492792-37492792 on Build GRCh38
rs370094065 Chr.22: 37492786-37492786 on Build GRCh38
rs759424958 Chr.22: 37492707-37492707 on Build GRCh38
rs377009692 Chr.22: 37492762-37492762 on Build GRCh38
rs141869325 Chr.22: 37492777-37492777 on Build GRCh38
rs562467144 Chr.22: 37492622-37492622 on Build GRCh38
rs756882892 Chr.22: 37492737-37492737 on Build GRCh38
rs537956524 Chr.22: 37492798-37492798 on Build GRCh38
rs1027817790 Chr.22: 37492929-37492930 on Build GRCh38
rs768943709 Chr.22: 37492592-37492592 on Build GRCh38
rs772601712 Chr.22: 37492763-37492763 on Build GRCh38
rs5756696 Chr.22: 37492861-37492861 on Build GRCh38
rs754919092 Chr.22: 37492625-37492625 on Build GRCh38
rs760946767 Chr.22: 37492698-37492698 on Build GRCh38
rs773438828 Chr.22: 37492616-37492616 on Build GRCh38
rs1046539117 Chr.22: 37492645-37492645 on Build GRCh38
rs769164076 Chr.22: 37492672-37492672 on Build GRCh38
rs778316775 Chr.22: 37492838-37492838 on Build GRCh38
rs140388457 Chr.22: 37492864-37492864 on Build GRCh38
rs751254949 Chr.22: 37492712-37492712 on Build GRCh38
rs56022423 Chr.22: 37492833-37492833 on Build GRCh38
rs112141463 Chr.22: 37492936-37492936 on Build GRCh38

View Full Product Details