Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37496320-37496822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD10
Entrez Gene ID
29775
Gene Name
caspase recruitment domain family member 10
Gene Aliases
BIMP1, CARMA3
UniGene
Hs.57973
Gene Chromosome Location
Chr.22: 37490362-37519203 on Build GRCh38
Transcripts
NM_014550.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7418919 c.1196C>A p.S399* Chr.22: 37496454-37496454 on Build GRCh38
5909868 c.2030C>T p.S677F Chr.22: 37496478-37496478 on Build GRCh38
4890660 c.1953G>T p.R651S Chr.22: 37496555-37496555 on Build GRCh38
3554178 c.2054C>T p.S685L Chr.22: 37496454-37496454 on Build GRCh38
3554177 c.2059+1G>A p.? Chr.22: 37496448-37496448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776672107 Chr.22: 37496563-37496563 on Build GRCh38
rs144994747 Chr.22: 37496522-37496522 on Build GRCh38
rs770909825 Chr.22: 37496559-37496559 on Build GRCh38
rs371707395 Chr.22: 37496428-37496428 on Build GRCh38
rs774751397 Chr.22: 37496438-37496438 on Build GRCh38
rs762603074 Chr.22: 37496605-37496605 on Build GRCh38
rs780289938 Chr.22: 37496424-37496424 on Build GRCh38
rs768665045 Chr.22: 37496483-37496484 on Build GRCh38
rs754598079 Chr.22: 37496406-37496406 on Build GRCh38
rs150364155 Chr.22: 37496730-37496730 on Build GRCh38
rs926427362 Chr.22: 37496576-37496576 on Build GRCh38
rs543000883 Chr.22: 37496690-37496690 on Build GRCh38
rs5756701 Chr.22: 37496363-37496363 on Build GRCh38
rs904333792 Chr.22: 37496655-37496655 on Build GRCh38
rs772296989 Chr.22: 37496450-37496450 on Build GRCh38
rs142113268 Chr.22: 37496454-37496454 on Build GRCh38
rs575823689 Chr.22: 37496643-37496643 on Build GRCh38
rs540908644 Chr.22: 37496758-37496758 on Build GRCh38
rs532713350 Chr.22: 37496800-37496800 on Build GRCh38
rs771831777 Chr.22: 37496792-37496792 on Build GRCh38
rs752343476 Chr.22: 37496413-37496413 on Build GRCh38
rs373196505 Chr.22: 37496481-37496481 on Build GRCh38
rs973567905 Chr.22: 37496791-37496791 on Build GRCh38
rs962869427 Chr.22: 37496763-37496763 on Build GRCh38
rs528521458 Chr.22: 37496734-37496734 on Build GRCh38
rs766281367 Chr.22: 37496464-37496464 on Build GRCh38
rs140404747 Chr.22: 37496396-37496396 on Build GRCh38
rs151150977 Chr.22: 37496469-37496469 on Build GRCh38
rs765039631 Chr.22: 37496598-37496598 on Build GRCh38
rs777296878 Chr.22: 37496497-37496497 on Build GRCh38
rs992453463 Chr.22: 37496657-37496657 on Build GRCh38
rs1037982694 Chr.22: 37496675-37496676 on Build GRCh38
rs1038035191 Chr.22: 37496395-37496395 on Build GRCh38
rs752877017 Chr.22: 37496574-37496577 on Build GRCh38
rs769110685 Chr.22: 37496528-37496528 on Build GRCh38
rs374827502 Chr.22: 37496412-37496412 on Build GRCh38
rs749725348 Chr.22: 37496516-37496516 on Build GRCh38
rs773438740 Chr.22: 37496546-37496546 on Build GRCh38
rs141109936 Chr.22: 37496475-37496475 on Build GRCh38
rs565470955 Chr.22: 37496775-37496775 on Build GRCh38
rs375211837 Chr.22: 37496429-37496429 on Build GRCh38
rs573410769 Chr.22: 37496350-37496350 on Build GRCh38
rs751008593 Chr.22: 37496499-37496499 on Build GRCh38
rs778516160 Chr.22: 37496621-37496621 on Build GRCh38
rs374348082 Chr.22: 37496416-37496416 on Build GRCh38
rs956073649 Chr.22: 37496675-37496675 on Build GRCh38
rs201152897 Chr.22: 37496543-37496543 on Build GRCh38
rs577751588 Chr.22: 37496529-37496529 on Build GRCh38
rs928981846 Chr.22: 37496801-37496801 on Build GRCh38
rs932365712 Chr.22: 37496702-37496702 on Build GRCh38
rs745664103 Chr.22: 37496782-37496782 on Build GRCh38
rs534915588 Chr.22: 37496440-37496440 on Build GRCh38
rs1003844052 Chr.22: 37496379-37496379 on Build GRCh38
rs1000691733 Chr.22: 37496663-37496663 on Build GRCh38
rs780670759 Chr.22: 37496515-37496515 on Build GRCh38
rs759371417 Chr.22: 37496582-37496582 on Build GRCh38
rs939203411 Chr.22: 37496653-37496653 on Build GRCh38
rs890360469 Chr.22: 37496585-37496585 on Build GRCh38
rs1031722247 Chr.22: 37496674-37496674 on Build GRCh38
rs746396361 Chr.22: 37496417-37496417 on Build GRCh38
rs760692759 Chr.22: 37496461-37496461 on Build GRCh38
rs377261206 Chr.22: 37496484-37496484 on Build GRCh38
rs980292708 Chr.22: 37496752-37496752 on Build GRCh38
rs368935726 Chr.22: 37496414-37496414 on Build GRCh38
rs770299322 Chr.22: 37496422-37496422 on Build GRCh38
rs756596755 Chr.22: 37496504-37496504 on Build GRCh38
rs760865621 Chr.22: 37496555-37496555 on Build GRCh38
rs757997262 Chr.22: 37496492-37496492 on Build GRCh38
rs372414450 Chr.22: 37496480-37496480 on Build GRCh38
rs1007428981 Chr.22: 37496604-37496604 on Build GRCh38
rs748408495 Chr.22: 37496536-37496536 on Build GRCh38
rs557461225 Chr.22: 37496602-37496602 on Build GRCh38
rs867411554 Chr.22: 37496714-37496714 on Build GRCh38
rs970270595 Chr.22: 37496738-37496738 on Build GRCh38

View Full Product Details