Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37566314-37566818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42EP1
Entrez Gene ID
11135
Gene Name
CDC42 effector protein 1
Gene Aliases
BORG5, CEP1, MSE55
UniGene
Hs.225356
Gene Chromosome Location
Chr.22: 37559778-37569405 on Build GRCh38
Transcripts
NM_152243.2 XM_006724107.1 XM_006724108.1 XM_006724109.1 XM_017028541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5981847 c.423C>A p.D141E Chr.22: 37566772-37566772 on Build GRCh38
3554186 c.243C>T p.F81F Chr.22: 37566592-37566592 on Build GRCh38
1416146 c.227G>A p.R76H Chr.22: 37566576-37566576 on Build GRCh38
5892462 c.305C>T p.P102L Chr.22: 37566654-37566654 on Build GRCh38
4582389 c.45G>A p.L15L Chr.22: 37566394-37566394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748248737 Chr.22: 37566551-37566551 on Build GRCh38
rs140945384 Chr.22: 37566628-37566628 on Build GRCh38
rs533958979 Chr.22: 37566790-37566790 on Build GRCh38
rs1011747026 Chr.22: 37566630-37566630 on Build GRCh38
rs995090347 Chr.22: 37566604-37566604 on Build GRCh38
rs111307192 Chr.22: 37566733-37566733 on Build GRCh38
rs754990384 Chr.22: 37566796-37566796 on Build GRCh38
rs899369484 Chr.22: 37566689-37566689 on Build GRCh38
rs747116423 Chr.22: 37566618-37566618 on Build GRCh38
rs370359827 Chr.22: 37566793-37566793 on Build GRCh38
rs376654024 Chr.22: 37566593-37566593 on Build GRCh38
rs969629486 Chr.22: 37566682-37566682 on Build GRCh38
rs773216227 Chr.22: 37566555-37566555 on Build GRCh38
rs956831361 Chr.22: 37566625-37566625 on Build GRCh38
rs142405513 Chr.22: 37566406-37566406 on Build GRCh38
rs199672437 Chr.22: 37566792-37566792 on Build GRCh38
rs763778405 Chr.22: 37566584-37566584 on Build GRCh38
rs748423187 Chr.22: 37566605-37566605 on Build GRCh38
rs375176513 Chr.22: 37566486-37566486 on Build GRCh38
rs1036116453 Chr.22: 37566422-37566422 on Build GRCh38
rs774551485 Chr.22: 37566475-37566475 on Build GRCh38
rs61739954 Chr.22: 37566576-37566576 on Build GRCh38
rs554614618 Chr.22: 37566460-37566460 on Build GRCh38
rs760004282 Chr.22: 37566365-37566365 on Build GRCh38
rs773879023 Chr.22: 37566688-37566688 on Build GRCh38
rs199905468 Chr.22: 37566662-37566662 on Build GRCh38
rs368492182 Chr.22: 37566405-37566405 on Build GRCh38
rs754729269 Chr.22: 37566433-37566433 on Build GRCh38
rs1034204798 Chr.22: 37566783-37566783 on Build GRCh38
rs769657329 Chr.22: 37566677-37566677 on Build GRCh38
rs759701710 Chr.22: 37566740-37566740 on Build GRCh38
rs147161865 Chr.22: 37566774-37566774 on Build GRCh38
rs889355431 Chr.22: 37566498-37566498 on Build GRCh38
rs765059393 Chr.22: 37566567-37566567 on Build GRCh38
rs143551825 Chr.22: 37566770-37566770 on Build GRCh38
rs139353065 Chr.22: 37566639-37566639 on Build GRCh38
rs755947790 Chr.22: 37566408-37566408 on Build GRCh38
rs771780348 Chr.22: 37566753-37566753 on Build GRCh38
rs770227871 Chr.22: 37566461-37566461 on Build GRCh38
rs775782167 Chr.22: 37566469-37566469 on Build GRCh38
rs778680876 Chr.22: 37566441-37566441 on Build GRCh38
rs369640551 Chr.22: 37566394-37566394 on Build GRCh38
rs751206521 Chr.22: 37566632-37566632 on Build GRCh38
rs573067727 Chr.22: 37566772-37566772 on Build GRCh38
rs371962479 Chr.22: 37566638-37566638 on Build GRCh38
rs763119468 Chr.22: 37566379-37566379 on Build GRCh38
rs1002607029 Chr.22: 37566516-37566516 on Build GRCh38
rs768284217 Chr.22: 37566342-37566342 on Build GRCh38
rs769736700 Chr.22: 37566626-37566626 on Build GRCh38
rs779105093 Chr.22: 37566597-37566597 on Build GRCh38
rs778519457 Chr.22: 37566344-37566344 on Build GRCh38
rs376012796 Chr.22: 37566355-37566355 on Build GRCh38
rs758778225 Chr.22: 37566655-37566655 on Build GRCh38
rs768905815 Chr.22: 37566473-37566473 on Build GRCh38
rs1015613673 Chr.22: 37566420-37566420 on Build GRCh38
rs149573903 Chr.22: 37566559-37566559 on Build GRCh38
rs1019471800 Chr.22: 37566623-37566624 on Build GRCh38
rs752233975 Chr.22: 37566512-37566512 on Build GRCh38
rs916954381 Chr.22: 37566658-37566658 on Build GRCh38
rs572970148 Chr.22: 37566504-37566504 on Build GRCh38
rs745922297 Chr.22: 37566676-37566676 on Build GRCh38
rs190518121 Chr.22: 37566532-37566532 on Build GRCh38
rs775196114 Chr.22: 37566627-37566627 on Build GRCh38
rs750109725 Chr.22: 37566377-37566377 on Build GRCh38
rs373648442 Chr.22: 37566586-37566586 on Build GRCh38
rs145680167 Chr.22: 37566563-37566563 on Build GRCh38
rs1036124428 Chr.22: 37566654-37566654 on Build GRCh38
rs754285888 Chr.22: 37566587-37566587 on Build GRCh38
rs113668714 Chr.22: 37566507-37566507 on Build GRCh38
rs370815357 Chr.22: 37566472-37566472 on Build GRCh38
rs776700659 Chr.22: 37566562-37566562 on Build GRCh38
rs199733984 Chr.22: 37566376-37566376 on Build GRCh38
rs756627353 Chr.22: 37566531-37566531 on Build GRCh38
rs768436129 Chr.22: 37566686-37566686 on Build GRCh38
rs370072833 Chr.22: 37566623-37566623 on Build GRCh38
rs779925168 Chr.22: 37566409-37566409 on Build GRCh38
rs376901875 Chr.22: 37566734-37566734 on Build GRCh38
rs774081807 Chr.22: 37566748-37566748 on Build GRCh38
rs138108680 Chr.22: 37566749-37566749 on Build GRCh38
rs757940883 Chr.22: 37566582-37566582 on Build GRCh38
rs536131927 Chr.22: 37566380-37566380 on Build GRCh38
rs201283947 Chr.22: 37566635-37566635 on Build GRCh38
rs757931238 Chr.22: 37566513-37566513 on Build GRCh38
rs377383839 Chr.22: 37566458-37566458 on Build GRCh38
rs778789525 Chr.22: 37566797-37566797 on Build GRCh38
rs761766893 Chr.22: 37566401-37566401 on Build GRCh38
rs201030338 Chr.22: 37566709-37566709 on Build GRCh38
rs751855498 Chr.22: 37566728-37566728 on Build GRCh38
rs758694944 Chr.22: 37566718-37566718 on Build GRCh38
rs146834922 Chr.22: 37566424-37566424 on Build GRCh38
rs200018136 Chr.22: 37566636-37566636 on Build GRCh38
rs1021549691 Chr.22: 37566511-37566511 on Build GRCh38
rs201388894 Chr.22: 37566534-37566534 on Build GRCh38
rs73418177 Chr.22: 37566550-37566550 on Build GRCh38
rs369211517 Chr.22: 37566585-37566585 on Build GRCh38
rs746406204 Chr.22: 37566527-37566527 on Build GRCh38
rs991988332 Chr.22: 37566560-37566560 on Build GRCh38
rs767434715 Chr.22: 37566476-37566476 on Build GRCh38
rs757834412 Chr.22: 37566454-37566454 on Build GRCh38
rs773202470 Chr.22: 37566477-37566477 on Build GRCh38
rs762710520 Chr.22: 37566683-37566683 on Build GRCh38
rs769024875 Chr.22: 37566356-37566356 on Build GRCh38
rs571270968 Chr.22: 37566669-37566669 on Build GRCh38
rs752013277 Chr.22: 37566781-37566781 on Build GRCh38
rs202138466 Chr.22: 37566703-37566703 on Build GRCh38
rs753569701 Chr.22: 37566419-37566419 on Build GRCh38
rs953392403 Chr.22: 37566610-37566610 on Build GRCh38
rs768991551 Chr.22: 37566543-37566543 on Build GRCh38
rs777109425 Chr.22: 37566523-37566523 on Build GRCh38
rs765724113 Chr.22: 37566705-37566705 on Build GRCh38
rs757605019 Chr.22: 37566730-37566730 on Build GRCh38
rs774440829 Chr.22: 37566631-37566632 on Build GRCh38
rs772224448 Chr.22: 37566607-37566607 on Build GRCh38
rs771080796 Chr.22: 37566621-37566621 on Build GRCh38
rs767836939 Chr.22: 37566680-37566703 on Build GRCh38
rs148893156 Chr.22: 37566692-37566692 on Build GRCh38
rs747642308 Chr.22: 37566350-37566350 on Build GRCh38
rs373084609 Chr.22: 37566670-37566670 on Build GRCh38
rs781374875 Chr.22: 37566671-37566671 on Build GRCh38
rs771513416 Chr.22: 37566353-37566353 on Build GRCh38
rs186346005 Chr.22: 37566356-37566356 on Build GRCh38
rs772839791 Chr.22: 37566769-37566769 on Build GRCh38
rs775490487 Chr.22: 37566678-37566678 on Build GRCh38
rs778052874 Chr.22: 37566617-37566617 on Build GRCh38
rs764490162 Chr.22: 37566778-37566778 on Build GRCh38
rs762035931 Chr.22: 37566632-37566632 on Build GRCh38
rs764483507 Chr.22: 37566719-37566719 on Build GRCh38
rs150186371 Chr.22: 37566631-37566631 on Build GRCh38
rs759509615 Chr.22: 37566624-37566624 on Build GRCh38
rs901627049 Chr.22: 37566427-37566427 on Build GRCh38
rs528047239 Chr.22: 37566704-37566704 on Build GRCh38
rs1050604887 Chr.22: 37566784-37566784 on Build GRCh38
rs753729994 Chr.22: 37566495-37566495 on Build GRCh38
rs1029006626 Chr.22: 37566439-37566439 on Build GRCh38
rs374889292 Chr.22: 37566444-37566444 on Build GRCh38
rs757689044 Chr.22: 37566782-37566782 on Build GRCh38
rs760780041 Chr.22: 37566556-37566556 on Build GRCh38
rs368470734 Chr.22: 37566492-37566492 on Build GRCh38
rs947968701 Chr.22: 37566575-37566575 on Build GRCh38

View Full Product Details