Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37568030-37568526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42EP1
Entrez Gene ID
11135
Gene Name
CDC42 effector protein 1
Gene Aliases
BORG5, CEP1, MSE55
UniGene
Hs.225356
Gene Chromosome Location
Chr.22: 37559778-37569405 on Build GRCh38
Transcripts
NM_152243.2 XM_006724107.1 XM_006724108.1 XM_006724109.1 XM_017028541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3390217 c.505T>C p.S169P Chr.22: 37568149-37568149 on Build GRCh38
1291243 c.869T>A p.V290E Chr.22: 37568513-37568513 on Build GRCh38
7448495 c.817C>G p.P273A Chr.22: 37568461-37568461 on Build GRCh38
3150496 c.741C>G p.A247A Chr.22: 37568385-37568385 on Build GRCh38
7322546 c.649C>A p.P217T Chr.22: 37568293-37568293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779661388 Chr.22: 37568200-37568200 on Build GRCh38
rs368647335 Chr.22: 37568203-37568203 on Build GRCh38
rs75025069 Chr.22: 37568134-37568134 on Build GRCh38
rs150706378 Chr.22: 37568182-37568182 on Build GRCh38
rs200344981 Chr.22: 37568382-37568382 on Build GRCh38
rs918610738 Chr.22: 37568156-37568156 on Build GRCh38
rs1013911951 Chr.22: 37568462-37568462 on Build GRCh38
rs767922622 Chr.22: 37568072-37568072 on Build GRCh38
rs746292047 Chr.22: 37568361-37568361 on Build GRCh38
rs756986704 Chr.22: 37568187-37568187 on Build GRCh38
rs759375037 Chr.22: 37568446-37568446 on Build GRCh38
rs767073773 Chr.22: 37568438-37568438 on Build GRCh38
rs749736100 Chr.22: 37568390-37568410 on Build GRCh38
rs372840111 Chr.22: 37568356-37568356 on Build GRCh38
rs955399481 Chr.22: 37568317-37568317 on Build GRCh38
rs777498992 Chr.22: 37568130-37568130 on Build GRCh38
rs1051192080 Chr.22: 37568162-37568162 on Build GRCh38
rs752564269 Chr.22: 37568119-37568119 on Build GRCh38
rs757020201 Chr.22: 37568422-37568422 on Build GRCh38
rs768914801 Chr.22: 37568394-37568394 on Build GRCh38
rs763936071 Chr.22: 37568408-37568427 on Build GRCh38
rs779016798 Chr.22: 37568483-37568483 on Build GRCh38
rs574213191 Chr.22: 37568470-37568470 on Build GRCh38
rs959628757 Chr.22: 37568148-37568148 on Build GRCh38
rs764859910 Chr.22: 37568455-37568455 on Build GRCh38
rs760914018 Chr.22: 37568160-37568160 on Build GRCh38
rs766487152 Chr.22: 37568092-37568092 on Build GRCh38
rs753989263 Chr.22: 37568093-37568093 on Build GRCh38
rs780455676 Chr.22: 37568386-37568427 on Build GRCh38
rs138703284 Chr.22: 37568138-37568138 on Build GRCh38
rs201024113 Chr.22: 37568147-37568147 on Build GRCh38
rs751408705 Chr.22: 37568421-37568422 on Build GRCh38
rs144367023 Chr.22: 37568339-37568339 on Build GRCh38
rs769457147 Chr.22: 37568199-37568199 on Build GRCh38
rs755568564 Chr.22: 37568369-37568389 on Build GRCh38
rs750866053 Chr.22: 37568400-37568400 on Build GRCh38
rs868730861 Chr.22: 37568503-37568503 on Build GRCh38
rs868496440 Chr.22: 37568323-37568323 on Build GRCh38
rs201200905 Chr.22: 37568384-37568384 on Build GRCh38
rs770970250 Chr.22: 37568444-37568444 on Build GRCh38
rs200195385 Chr.22: 37568402-37568422 on Build GRCh38
rs757916796 Chr.22: 37568395-37568395 on Build GRCh38
rs202036648 Chr.22: 37568380-37568380 on Build GRCh38
rs756660653 Chr.22: 37568473-37568473 on Build GRCh38
rs766504638 Chr.22: 37568404-37568412 on Build GRCh38
rs761921446 Chr.22: 37568381-37568381 on Build GRCh38
rs756499308 Chr.22: 37568405-37568405 on Build GRCh38
rs776909370 Chr.22: 37568294-37568294 on Build GRCh38
rs62235034 Chr.22: 37568422-37568422 on Build GRCh38
rs750705839 Chr.22: 37568073-37568073 on Build GRCh38
rs753475671 Chr.22: 37568345-37568345 on Build GRCh38
rs765288471 Chr.22: 37568289-37568291 on Build GRCh38
rs750032816 Chr.22: 37568500-37568501 on Build GRCh38
rs962980028 Chr.22: 37568133-37568133 on Build GRCh38
rs751300981 Chr.22: 37568335-37568375 on Build GRCh38
rs775307114 Chr.22: 37568406-37568406 on Build GRCh38
rs375221026 Chr.22: 37568234-37568234 on Build GRCh38
rs774998622 Chr.22: 37568151-37568151 on Build GRCh38
rs200631887 Chr.22: 37568207-37568207 on Build GRCh38
rs754221625 Chr.22: 37568476-37568476 on Build GRCh38
rs772130894 Chr.22: 37568434-37568434 on Build GRCh38
rs141161720 Chr.22: 37568167-37568167 on Build GRCh38
rs115843472 Chr.22: 37568196-37568196 on Build GRCh38
rs66468174 Chr.22: 37568406-37568426 on Build GRCh38
rs13055845 Chr.22: 37568406-37568406 on Build GRCh38
rs77417880 Chr.22: 37568407-37568407 on Build GRCh38
rs752645135 Chr.22: 37568326-37568367 on Build GRCh38
rs762377855 Chr.22: 37568464-37568464 on Build GRCh38
rs767175635 Chr.22: 37568438-37568438 on Build GRCh38
rs370872947 Chr.22: 37568320-37568320 on Build GRCh38
rs984581915 Chr.22: 37568399-37568399 on Build GRCh38
rs768799950 Chr.22: 37568378-37568378 on Build GRCh38
rs143174040 Chr.22: 37568069-37568069 on Build GRCh38
rs554282751 Chr.22: 37568280-37568280 on Build GRCh38
rs556416144 Chr.22: 37568209-37568209 on Build GRCh38
rs73418181 Chr.22: 37568091-37568091 on Build GRCh38
rs199910466 Chr.22: 37568173-37568173 on Build GRCh38
rs10427607 Chr.22: 37568376-37568376 on Build GRCh38
rs771349775 Chr.22: 37568293-37568293 on Build GRCh38
rs775592056 Chr.22: 37568225-37568225 on Build GRCh38
rs2281098 Chr.22: 37568292-37568292 on Build GRCh38
rs13055511 Chr.22: 37568433-37568433 on Build GRCh38
rs918544047 Chr.22: 37568311-37568311 on Build GRCh38
rs781722526 Chr.22: 37568360-37568360 on Build GRCh38
rs367706200 Chr.22: 37568233-37568233 on Build GRCh38
rs372858376 Chr.22: 37568144-37568144 on Build GRCh38
rs972707593 Chr.22: 37568146-37568146 on Build GRCh38
rs199535461 Chr.22: 37568220-37568220 on Build GRCh38
rs752219652 Chr.22: 37568394-37568394 on Build GRCh38
rs761116224 Chr.22: 37568210-37568210 on Build GRCh38
rs772587344 Chr.22: 37568159-37568159 on Build GRCh38
rs772473187 Chr.22: 37568401-37568422 on Build GRCh38
rs756968657 Chr.22: 37568345-37568386 on Build GRCh38
rs372439509 Chr.22: 37568250-37568250 on Build GRCh38
rs754520744 Chr.22: 37568275-37568275 on Build GRCh38
rs748494440 Chr.22: 37568398-37568416 on Build GRCh38
rs146373289 Chr.22: 37568268-37568268 on Build GRCh38
rs763894185 Chr.22: 37568334-37568334 on Build GRCh38
rs778836625 Chr.22: 37568113-37568113 on Build GRCh38
rs766190951 Chr.22: 37568341-37568341 on Build GRCh38
rs773314401 Chr.22: 37568440-37568440 on Build GRCh38
rs760816253 Chr.22: 37568217-37568218 on Build GRCh38
rs374489029 Chr.22: 37568223-37568223 on Build GRCh38
rs779190430 Chr.22: 37568427-37568427 on Build GRCh38
rs200534442 Chr.22: 37568389-37568389 on Build GRCh38
rs770089711 Chr.22: 37568362-37568362 on Build GRCh38
rs755967235 Chr.22: 37568340-37568340 on Build GRCh38
rs187761157 Chr.22: 37568417-37568417 on Build GRCh38
rs73418183 Chr.22: 37568224-37568224 on Build GRCh38
rs746204120 Chr.22: 37568296-37568296 on Build GRCh38
rs13057024 Chr.22: 37568454-37568454 on Build GRCh38
rs770073258 Chr.22: 37568326-37568326 on Build GRCh38
rs961996966 Chr.22: 37568333-37568333 on Build GRCh38
rs868388731 Chr.22: 37568344-37568344 on Build GRCh38
rs779820977 Chr.22: 37568267-37568267 on Build GRCh38
rs757076420 Chr.22: 37568244-37568244 on Build GRCh38
rs774834159 Chr.22: 37568395-37568414 on Build GRCh38
rs374180424 Chr.22: 37568174-37568174 on Build GRCh38
rs758382006 Chr.22: 37568180-37568180 on Build GRCh38
rs905590724 Chr.22: 37568496-37568496 on Build GRCh38
rs763550797 Chr.22: 37568466-37568466 on Build GRCh38
rs557993271 Chr.22: 37568350-37568350 on Build GRCh38
rs760869338 Chr.22: 37568403-37568422 on Build GRCh38
rs760558616 Chr.22: 37568387-37568387 on Build GRCh38
rs755340068 Chr.22: 37568401-37568402 on Build GRCh38
rs778238046 Chr.22: 37568351-37568351 on Build GRCh38
rs755001482 Chr.22: 37568101-37568101 on Build GRCh38
rs546212609 Chr.22: 37568336-37568336 on Build GRCh38
rs541568053 Chr.22: 37568188-37568188 on Build GRCh38
rs780505310 Chr.22: 37568475-37568475 on Build GRCh38
rs749469140 Chr.22: 37568375-37568375 on Build GRCh38
rs774372611 Chr.22: 37568338-37568338 on Build GRCh38
rs780789435 Chr.22: 37568348-37568368 on Build GRCh38
rs987241599 Chr.22: 37568325-37568325 on Build GRCh38
rs867254270 Chr.22: 37568354-37568354 on Build GRCh38
rs759667566 Chr.22: 37568168-37568168 on Build GRCh38
rs758136839 Chr.22: 37568331-37568331 on Build GRCh38
rs753569685 Chr.22: 37568390-37568390 on Build GRCh38
rs752340748 Chr.22: 37568461-37568461 on Build GRCh38
rs767172943 Chr.22: 37568245-37568245 on Build GRCh38
rs376281327 Chr.22: 37568150-37568150 on Build GRCh38
rs750109544 Chr.22: 37568247-37568247 on Build GRCh38
rs759160666 Chr.22: 37568392-37568392 on Build GRCh38
rs755354219 Chr.22: 37568481-37568481 on Build GRCh38
rs970244213 Chr.22: 37568258-37568258 on Build GRCh38
rs776609121 Chr.22: 37568445-37568445 on Build GRCh38
rs755879390 Chr.22: 37568266-37568266 on Build GRCh38
rs758221624 Chr.22: 37568126-37568126 on Build GRCh38
rs760642517 Chr.22: 37568443-37568443 on Build GRCh38
rs763008855 Chr.22: 37568407-37568427 on Build GRCh38
rs1017135033 Chr.22: 37568277-37568277 on Build GRCh38
rs748335297 Chr.22: 37568492-37568492 on Build GRCh38
rs62235033 Chr.22: 37568410-37568410 on Build GRCh38
rs202036105 Chr.22: 37568386-37568386 on Build GRCh38
rs745428305 Chr.22: 37568365-37568406 on Build GRCh38
rs200781970 Chr.22: 37568385-37568385 on Build GRCh38
rs757748503 Chr.22: 37568358-37568358 on Build GRCh38

View Full Product Details