Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38299816-38300326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1E
Entrez Gene ID
1454
Gene Name
casein kinase 1 epsilon
Gene Aliases
CKIepsilon, HCKIE
UniGene
Hs.474833
Gene Chromosome Location
Chr.22: 38290691-38318084 on Build GRCh38
Transcripts
NM_001894.4 NM_152221.2 NM_001289912.1

Gene Symbol
LOC400927-CSNK1E
Entrez Gene ID
102800317
Gene Name
LOC400927-CSNK1E readthrough
Gene Aliases
-
UniGene
Hs.720223
Gene Chromosome Location
Chr.22: 38290691-38398522 on Build GRCh38
Transcripts
NM_001894.4 NM_152221.2 NM_001289912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1416234 c.680G>A p.R227Q Chr.22: 38299951-38299951 on Build GRCh38
4675138 c.634C>T p.P212S Chr.22: 38299997-38299997 on Build GRCh38
5859366 c.715G>A p.V239I Chr.22: 38299916-38299916 on Build GRCh38
3151213 c.604G>A p.V202M Chr.22: 38300027-38300027 on Build GRCh38
1034117 c.705G>A p.T235T Chr.22: 38299926-38299926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201784219 Chr.22: 38299882-38299882 on Build GRCh38
rs564949394 Chr.22: 38300145-38300145 on Build GRCh38
rs762388387 Chr.22: 38299971-38299971 on Build GRCh38
rs765797254 Chr.22: 38299998-38299998 on Build GRCh38
rs552285492 Chr.22: 38300230-38300230 on Build GRCh38
rs770924331 Chr.22: 38299842-38299842 on Build GRCh38
rs764636536 Chr.22: 38299849-38299849 on Build GRCh38
rs1016811039 Chr.22: 38300194-38300194 on Build GRCh38
rs779946187 Chr.22: 38299853-38299854 on Build GRCh38
rs754385642 Chr.22: 38299854-38299854 on Build GRCh38
rs6001090 Chr.22: 38300173-38300173 on Build GRCh38
rs950334984 Chr.22: 38299860-38299860 on Build GRCh38
rs754542465 Chr.22: 38299864-38299864 on Build GRCh38
rs974858897 Chr.22: 38300204-38300204 on Build GRCh38
rs765082140 Chr.22: 38300189-38300189 on Build GRCh38
rs911139066 Chr.22: 38300254-38300254 on Build GRCh38
rs757907710 Chr.22: 38299858-38299858 on Build GRCh38
rs146245501 Chr.22: 38299866-38299866 on Build GRCh38
rs115297880 Chr.22: 38299847-38299847 on Build GRCh38
rs1013875122 Chr.22: 38300174-38300174 on Build GRCh38
rs776853410 Chr.22: 38300089-38300089 on Build GRCh38
rs777517365 Chr.22: 38300095-38300095 on Build GRCh38
rs780653845 Chr.22: 38300028-38300028 on Build GRCh38
rs774242774 Chr.22: 38299920-38299920 on Build GRCh38
rs201174621 Chr.22: 38299868-38299868 on Build GRCh38
rs972104935 Chr.22: 38300206-38300206 on Build GRCh38
rs1803339 Chr.22: 38299926-38299926 on Build GRCh38
rs754690883 Chr.22: 38300017-38300018 on Build GRCh38
rs554586329 Chr.22: 38300114-38300114 on Build GRCh38
rs148020976 Chr.22: 38300136-38300136 on Build GRCh38
rs200192255 Chr.22: 38300108-38300108 on Build GRCh38
rs544367014 Chr.22: 38300163-38300163 on Build GRCh38
rs754525242 Chr.22: 38300016-38300016 on Build GRCh38
rs1030316222 Chr.22: 38300207-38300207 on Build GRCh38
rs1003499185 Chr.22: 38299850-38299850 on Build GRCh38
rs750047552 Chr.22: 38300193-38300193 on Build GRCh38
rs751130741 Chr.22: 38299863-38299863 on Build GRCh38
rs748988647 Chr.22: 38299934-38299936 on Build GRCh38
rs761035222 Chr.22: 38299938-38299938 on Build GRCh38
rs755801722 Chr.22: 38300073-38300073 on Build GRCh38
rs933933402 Chr.22: 38300101-38300101 on Build GRCh38
rs141492244 Chr.22: 38300172-38300172 on Build GRCh38
rs555620365 Chr.22: 38299917-38299917 on Build GRCh38
rs56360388 Chr.22: 38300148-38300148 on Build GRCh38
rs754296072 Chr.22: 38299963-38299963 on Build GRCh38
rs370345548 Chr.22: 38299944-38299944 on Build GRCh38
rs577612792 Chr.22: 38300188-38300188 on Build GRCh38
rs567069334 Chr.22: 38300218-38300218 on Build GRCh38
rs964267969 Chr.22: 38300295-38300295 on Build GRCh38
rs923839798 Chr.22: 38300268-38300268 on Build GRCh38
rs1010000104 Chr.22: 38300276-38300276 on Build GRCh38
rs775792943 Chr.22: 38299928-38299928 on Build GRCh38
rs749189607 Chr.22: 38299903-38299903 on Build GRCh38
rs778643708 Chr.22: 38300069-38300070 on Build GRCh38
rs759692009 Chr.22: 38299922-38299922 on Build GRCh38
rs752339218 Chr.22: 38300053-38300053 on Build GRCh38
rs750997703 Chr.22: 38300002-38300002 on Build GRCh38
rs554808914 Chr.22: 38300130-38300130 on Build GRCh38
rs368022599 Chr.22: 38300122-38300122 on Build GRCh38
rs962169841 Chr.22: 38300195-38300195 on Build GRCh38
rs78340378 Chr.22: 38300221-38300221 on Build GRCh38
rs777609057 Chr.22: 38299896-38299896 on Build GRCh38
rs934647524 Chr.22: 38300078-38300078 on Build GRCh38
rs186833023 Chr.22: 38300131-38300131 on Build GRCh38
rs752963822 Chr.22: 38299846-38299846 on Build GRCh38

View Full Product Details