Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38656522-38657030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBY1
Entrez Gene ID
25776
Gene Name
chibby family member 1, beta catenin antagonist
Gene Aliases
C22orf2, CBY, HS508I15A, PGEA1, PIGEA-14, PIGEA14, arb1
UniGene
Hs.334911
Gene Chromosome Location
Chr.22: 38656653-38673850 on Build GRCh38
Transcripts
NM_001002880.1 NM_015373.3 NM_001013647.1 NM_001291030.1 XM_006724291.2 XM_006724292.2 XM_006724293.2 XM_006724294.2 XM_006724295.2 XM_011530309.2 XM_011530310.1 XM_011530311.1 XM_011530314.1

Gene Symbol
FAM227A
Entrez Gene ID
646851
Gene Name
family with sequence similarity 227 member A
Gene Aliases
-
UniGene
Hs.542707
Gene Chromosome Location
Chr.22: 38578120-38656629 on Build GRCh38
Transcripts
NM_001002880.1 NM_015373.3 NM_001013647.1 NM_001291030.1 XM_006724291.2 XM_006724292.2 XM_006724293.2 XM_006724294.2 XM_006724295.2 XM_011530309.2 XM_011530310.1 XM_011530311.1 XM_011530314.1

SNP Detail

SNP ID Chromosome Location
rs536694074 Chr.22: 38656750-38656750 on Build GRCh38
rs1011380101 Chr.22: 38656611-38656611 on Build GRCh38
rs987719735 Chr.22: 38656877-38656877 on Build GRCh38
rs765174215 Chr.22: 38656598-38656598 on Build GRCh38
rs890634653 Chr.22: 38656670-38656670 on Build GRCh38
rs946702550 Chr.22: 38657001-38657001 on Build GRCh38
rs931863336 Chr.22: 38656738-38656738 on Build GRCh38
rs370567486 Chr.22: 38656871-38656871 on Build GRCh38
rs553510833 Chr.22: 38656880-38656880 on Build GRCh38
rs921735865 Chr.22: 38656950-38656950 on Build GRCh38
rs907861739 Chr.22: 38656548-38656548 on Build GRCh38
rs371568106 Chr.22: 38656717-38656717 on Build GRCh38
rs547532492 Chr.22: 38656665-38656665 on Build GRCh38
rs1045085723 Chr.22: 38656963-38656963 on Build GRCh38
rs1034816378 Chr.22: 38656765-38656765 on Build GRCh38
rs893456301 Chr.22: 38656710-38656710 on Build GRCh38
rs902726354 Chr.22: 38656799-38656799 on Build GRCh38
rs907616397 Chr.22: 38656935-38656935 on Build GRCh38
rs775747724 Chr.22: 38656829-38656829 on Build GRCh38
rs991447982 Chr.22: 38656862-38656862 on Build GRCh38
rs1003655739 Chr.22: 38656800-38656800 on Build GRCh38
rs578137359 Chr.22: 38656922-38656922 on Build GRCh38
rs963508274 Chr.22: 38656825-38656825 on Build GRCh38
rs982371428 Chr.22: 38656644-38656644 on Build GRCh38
rs944550125 Chr.22: 38656937-38656937 on Build GRCh38
rs1021456792 Chr.22: 38656791-38656791 on Build GRCh38
rs1054741559 Chr.22: 38656961-38656961 on Build GRCh38
rs1049766209 Chr.22: 38656763-38656763 on Build GRCh38
rs999225557 Chr.22: 38656695-38656695 on Build GRCh38
rs765973239 Chr.22: 38656972-38656972 on Build GRCh38
rs879257236 Chr.22: 38656946-38656946 on Build GRCh38
rs959062078 Chr.22: 38656652-38656652 on Build GRCh38
rs1052176770 Chr.22: 38656700-38656700 on Build GRCh38
rs1040348639 Chr.22: 38656728-38656728 on Build GRCh38
rs921959061 Chr.22: 38656852-38656852 on Build GRCh38
rs192953203 Chr.22: 38656719-38656719 on Build GRCh38
rs773113365 Chr.22: 38656610-38656610 on Build GRCh38
rs577918519 Chr.22: 38656578-38656578 on Build GRCh38
rs1021131843 Chr.22: 38656632-38656632 on Build GRCh38
rs931925808 Chr.22: 38656951-38656951 on Build GRCh38
rs565755339 Chr.22: 38656694-38656694 on Build GRCh38
rs113231583 Chr.22: 38656911-38656911 on Build GRCh38
rs539698122 Chr.22: 38656718-38656718 on Build GRCh38
rs112364075 Chr.22: 38656561-38656561 on Build GRCh38
rs907703781 Chr.22: 38656712-38656712 on Build GRCh38
rs569299523 Chr.22: 38656729-38656729 on Build GRCh38
rs761830514 Chr.22: 38656584-38656584 on Build GRCh38
rs567321749 Chr.22: 38656815-38656815 on Build GRCh38
rs9622836 Chr.22: 38656562-38656562 on Build GRCh38
rs561317614 Chr.22: 38656635-38656635 on Build GRCh38
rs1047899282 Chr.22: 38656699-38656699 on Build GRCh38
rs990479924 Chr.22: 38656663-38656663 on Build GRCh38
rs928213547 Chr.22: 38656649-38656649 on Build GRCh38
rs16999186 Chr.22: 38656828-38656828 on Build GRCh38
rs893765168 Chr.22: 38656778-38656778 on Build GRCh38
rs543666551 Chr.22: 38656786-38656786 on Build GRCh38
rs367736890 Chr.22: 38656656-38656656 on Build GRCh38
rs930421303 Chr.22: 38656693-38656693 on Build GRCh38
rs951761933 Chr.22: 38656899-38656899 on Build GRCh38
rs1051841073 Chr.22: 38656648-38656648 on Build GRCh38
rs554954403 Chr.22: 38656784-38656784 on Build GRCh38
rs959631939 Chr.22: 38656846-38656846 on Build GRCh38

View Full Product Details