Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39083642-39084135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3G
Entrez Gene ID
60489
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3G
Gene Aliases
A3G, ARCD, ARP-9, ARP9, CEM-15, CEM15, MDS019, bK150C2.7, dJ494G10.1
UniGene
Hs.660143
Gene Chromosome Location
Chr.22: 39077005-39087743 on Build GRCh38
Transcripts
NM_021822.3 XM_006724290.1 XM_017028903.1 XM_017028904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1172774 c.612C>G p.F204L Chr.22: 39083761-39083761 on Build GRCh38
1034251 c.712C>T p.R238C Chr.22: 39083861-39083861 on Build GRCh38
6095347 c.709C>A p.Q237K Chr.22: 39083858-39083858 on Build GRCh38
4502250 c.608C>T p.T203I Chr.22: 39083757-39083757 on Build GRCh38
1682271 c.661T>C p.C221R Chr.22: 39083810-39083810 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769013198 Chr.22: 39083870-39083870 on Build GRCh38
rs376831638 Chr.22: 39083816-39083816 on Build GRCh38
rs201410551 Chr.22: 39083862-39083862 on Build GRCh38
rs367576342 Chr.22: 39083738-39083738 on Build GRCh38
rs754838478 Chr.22: 39083696-39083696 on Build GRCh38
rs758587862 Chr.22: 39083926-39083935 on Build GRCh38
rs752409833 Chr.22: 39083898-39083898 on Build GRCh38
rs1033359097 Chr.22: 39083805-39083805 on Build GRCh38
rs372186666 Chr.22: 39083763-39083763 on Build GRCh38
rs552054727 Chr.22: 39083825-39083825 on Build GRCh38
rs201223458 Chr.22: 39083724-39083724 on Build GRCh38
rs761018617 Chr.22: 39083759-39083759 on Build GRCh38
rs376343533 Chr.22: 39083893-39083893 on Build GRCh38
rs749405831 Chr.22: 39083720-39083720 on Build GRCh38
rs747458446 Chr.22: 39083864-39083864 on Build GRCh38
rs199763152 Chr.22: 39083861-39083861 on Build GRCh38
rs779806845 Chr.22: 39083687-39083687 on Build GRCh38
rs745975590 Chr.22: 39083905-39083905 on Build GRCh38
rs755954429 Chr.22: 39083834-39083834 on Build GRCh38
rs1001919152 Chr.22: 39083741-39083741 on Build GRCh38
rs773857650 Chr.22: 39083886-39083886 on Build GRCh38
rs762359640 Chr.22: 39083876-39083876 on Build GRCh38
rs369268224 Chr.22: 39084074-39084074 on Build GRCh38
rs913418341 Chr.22: 39083921-39083921 on Build GRCh38
rs147805045 Chr.22: 39083851-39083851 on Build GRCh38
rs1043705675 Chr.22: 39084030-39084030 on Build GRCh38
rs758673416 Chr.22: 39083909-39083909 on Build GRCh38
rs780465264 Chr.22: 39083917-39083917 on Build GRCh38
rs201701375 Chr.22: 39083903-39083903 on Build GRCh38
rs911716535 Chr.22: 39083958-39083958 on Build GRCh38
rs368345672 Chr.22: 39083857-39083857 on Build GRCh38
rs749293157 Chr.22: 39083849-39083849 on Build GRCh38
rs757807279 Chr.22: 39083792-39083792 on Build GRCh38
rs757940716 Chr.22: 39083680-39083680 on Build GRCh38
rs535069398 Chr.22: 39084043-39084043 on Build GRCh38
rs779263309 Chr.22: 39083858-39083858 on Build GRCh38
rs375643135 Chr.22: 39083713-39083713 on Build GRCh38
rs747330904 Chr.22: 39083923-39083923 on Build GRCh38
rs749879946 Chr.22: 39083772-39083772 on Build GRCh38
rs921831233 Chr.22: 39083824-39083824 on Build GRCh38
rs936727227 Chr.22: 39084084-39084084 on Build GRCh38
rs906153986 Chr.22: 39084048-39084048 on Build GRCh38
rs200230305 Chr.22: 39083737-39083737 on Build GRCh38
rs752523113 Chr.22: 39083828-39083828 on Build GRCh38
rs762565003 Chr.22: 39083776-39083776 on Build GRCh38
rs368721163 Chr.22: 39083719-39083719 on Build GRCh38
rs777812545 Chr.22: 39083841-39083841 on Build GRCh38
rs771617770 Chr.22: 39083944-39083944 on Build GRCh38
rs373770107 Chr.22: 39083904-39083904 on Build GRCh38
rs574077421 Chr.22: 39083710-39083710 on Build GRCh38
rs371945683 Chr.22: 39083883-39083883 on Build GRCh38
rs892740165 Chr.22: 39083726-39083726 on Build GRCh38
rs975514453 Chr.22: 39084050-39084050 on Build GRCh38
rs1057083334 Chr.22: 39084108-39084109 on Build GRCh38
rs141223892 Chr.22: 39083873-39083873 on Build GRCh38
rs944158909 Chr.22: 39083691-39083691 on Build GRCh38
rs1009815921 Chr.22: 39083730-39083730 on Build GRCh38
rs567856443 Chr.22: 39084022-39084022 on Build GRCh38
rs781098159 Chr.22: 39083707-39083707 on Build GRCh38
rs370686830 Chr.22: 39083694-39083694 on Build GRCh38
rs749737101 Chr.22: 39083663-39083663 on Build GRCh38
rs950726356 Chr.22: 39083939-39083939 on Build GRCh38
rs752942820 Chr.22: 39083763-39083770 on Build GRCh38
rs751275292 Chr.22: 39083801-39083801 on Build GRCh38
rs1053821319 Chr.22: 39084086-39084086 on Build GRCh38
rs780901950 Chr.22: 39083826-39083826 on Build GRCh38

View Full Product Details