Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39086930-39087434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3G
Entrez Gene ID
60489
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3G
Gene Aliases
A3G, ARCD, ARP-9, ARP9, CEM-15, CEM15, MDS019, bK150C2.7, dJ494G10.1
UniGene
Hs.660143
Gene Chromosome Location
Chr.22: 39077005-39087743 on Build GRCh38
Transcripts
NM_021822.3 XM_006724290.1 XM_017028903.1 XM_017028904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6162248 c.1030A>T p.K344* Chr.22: 39087016-39087016 on Build GRCh38
5456959 c.1126C>T p.R376W Chr.22: 39087112-39087112 on Build GRCh38
5930081 c.1027T>A p.F343I Chr.22: 39087013-39087013 on Build GRCh38
6994512 c.1137C>A p.L379L Chr.22: 39087123-39087123 on Build GRCh38
5797390 c.1052T>C p.V351A Chr.22: 39087038-39087038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948958583 Chr.22: 39087228-39087228 on Build GRCh38
rs980328918 Chr.22: 39087230-39087230 on Build GRCh38
rs542618653 Chr.22: 39086990-39086990 on Build GRCh38
rs1006810583 Chr.22: 39087368-39087368 on Build GRCh38
rs763455077 Chr.22: 39087134-39087134 on Build GRCh38
rs756936602 Chr.22: 39087064-39087064 on Build GRCh38
rs772027464 Chr.22: 39086982-39086982 on Build GRCh38
rs868100936 Chr.22: 39087383-39087383 on Build GRCh38
rs756809176 Chr.22: 39087154-39087154 on Build GRCh38
rs763399550 Chr.22: 39087392-39087392 on Build GRCh38
rs776777572 Chr.22: 39086999-39086999 on Build GRCh38
rs1005835891 Chr.22: 39087120-39087120 on Build GRCh38
rs781085039 Chr.22: 39087357-39087357 on Build GRCh38
rs562177159 Chr.22: 39087369-39087369 on Build GRCh38
rs571551528 Chr.22: 39087327-39087327 on Build GRCh38
rs777612685 Chr.22: 39086966-39086966 on Build GRCh38
rs200261226 Chr.22: 39087172-39087172 on Build GRCh38
rs909990702 Chr.22: 39087352-39087352 on Build GRCh38
rs763628310 Chr.22: 39087149-39087149 on Build GRCh38
rs924148953 Chr.22: 39087030-39087030 on Build GRCh38
rs574850138 Chr.22: 39087161-39087161 on Build GRCh38
rs201660527 Chr.22: 39087160-39087160 on Build GRCh38
rs144459761 Chr.22: 39087121-39087121 on Build GRCh38
rs374774637 Chr.22: 39087138-39087138 on Build GRCh38
rs28425624 Chr.22: 39087188-39087188 on Build GRCh38
rs144179214 Chr.22: 39087092-39087092 on Build GRCh38
rs200761010 Chr.22: 39087354-39087354 on Build GRCh38
rs376834165 Chr.22: 39087150-39087150 on Build GRCh38
rs116198065 Chr.22: 39086965-39086965 on Build GRCh38
rs989752389 Chr.22: 39087058-39087058 on Build GRCh38
rs933669167 Chr.22: 39087336-39087336 on Build GRCh38
rs758785141 Chr.22: 39087033-39087033 on Build GRCh38
rs62228279 Chr.22: 39087103-39087103 on Build GRCh38
rs916727094 Chr.22: 39087224-39087224 on Build GRCh38
rs753370280 Chr.22: 39087062-39087062 on Build GRCh38
rs568940353 Chr.22: 39087390-39087390 on Build GRCh38
rs17537588 Chr.22: 39087319-39087319 on Build GRCh38
rs917520538 Chr.22: 39087223-39087223 on Build GRCh38
rs753435735 Chr.22: 39087367-39087367 on Build GRCh38
rs1005475449 Chr.22: 39086964-39086964 on Build GRCh38
rs374686174 Chr.22: 39087050-39087050 on Build GRCh38
rs527643869 Chr.22: 39087165-39087165 on Build GRCh38
rs773429303 Chr.22: 39087127-39087127 on Build GRCh38
rs11545130 Chr.22: 39087097-39087097 on Build GRCh38
rs761903353 Chr.22: 39087113-39087113 on Build GRCh38
rs369306100 Chr.22: 39087095-39087095 on Build GRCh38
rs532555016 Chr.22: 39087137-39087137 on Build GRCh38
rs55930989 Chr.22: 39087237-39087238 on Build GRCh38
rs201271449 Chr.22: 39087394-39087394 on Build GRCh38
rs374575022 Chr.22: 39087170-39087170 on Build GRCh38
rs868061842 Chr.22: 39087111-39087111 on Build GRCh38
rs554285739 Chr.22: 39086991-39086991 on Build GRCh38
rs752227439 Chr.22: 39087049-39087049 on Build GRCh38
rs573300046 Chr.22: 39087091-39087091 on Build GRCh38
rs1022542421 Chr.22: 39087166-39087166 on Build GRCh38
rs752179957 Chr.22: 39087282-39087282 on Build GRCh38
rs764268250 Chr.22: 39086993-39086993 on Build GRCh38
rs201264627 Chr.22: 39086977-39086977 on Build GRCh38
rs748312821 Chr.22: 39087362-39087362 on Build GRCh38
rs779838723 Chr.22: 39087169-39087169 on Build GRCh38
rs1014236678 Chr.22: 39087403-39087403 on Build GRCh38
rs573440801 Chr.22: 39087043-39087043 on Build GRCh38
rs760129468 Chr.22: 39087142-39087142 on Build GRCh38
rs777551336 Chr.22: 39087053-39087053 on Build GRCh38
rs993057196 Chr.22: 39087209-39087209 on Build GRCh38
rs527732972 Chr.22: 39087288-39087288 on Build GRCh38
rs998115571 Chr.22: 39087143-39087143 on Build GRCh38
rs1005288262 Chr.22: 39087301-39087321 on Build GRCh38
rs35205117 Chr.22: 39087342-39087342 on Build GRCh38
rs772537820 Chr.22: 39087134-39087136 on Build GRCh38
rs767045111 Chr.22: 39087046-39087046 on Build GRCh38
rs750831926 Chr.22: 39087027-39087027 on Build GRCh38
rs369048288 Chr.22: 39086976-39086976 on Build GRCh38
rs1047054123 Chr.22: 39087337-39087337 on Build GRCh38
rs1031682744 Chr.22: 39087186-39087186 on Build GRCh38
rs888245150 Chr.22: 39086954-39086954 on Build GRCh38
rs989643351 Chr.22: 39087203-39087203 on Build GRCh38
rs774857271 Chr.22: 39087405-39087405 on Build GRCh38
rs766755018 Chr.22: 39087166-39087166 on Build GRCh38
rs199683475 Chr.22: 39087069-39087069 on Build GRCh38
rs1047556830 Chr.22: 39087102-39087102 on Build GRCh38
rs148267053 Chr.22: 39087073-39087073 on Build GRCh38
rs6001422 Chr.22: 39087192-39087192 on Build GRCh38
rs755600900 Chr.22: 39087317-39087317 on Build GRCh38
rs910037066 Chr.22: 39087236-39087236 on Build GRCh38
rs371155567 Chr.22: 39087400-39087400 on Build GRCh38
rs557218170 Chr.22: 39087364-39087364 on Build GRCh38
rs765455917 Chr.22: 39087014-39087014 on Build GRCh38
rs776778876 Chr.22: 39087112-39087112 on Build GRCh38
rs1025492930 Chr.22: 39086952-39086952 on Build GRCh38
rs769878799 Chr.22: 39087380-39087380 on Build GRCh38
rs768613653 Chr.22: 39087173-39087173 on Build GRCh38
rs1000300520 Chr.22: 39087386-39087386 on Build GRCh38
rs1032146494 Chr.22: 39087398-39087398 on Build GRCh38

View Full Product Details