Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39087117-39087665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC3G
Entrez Gene ID
60489
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 3G
Gene Aliases
A3G, ARCD, ARP-9, ARP9, CEM-15, CEM15, MDS019, bK150C2.7, dJ494G10.1
UniGene
Hs.660143
Gene Chromosome Location
Chr.22: 39077005-39087743 on Build GRCh38
Transcripts
NM_021822.3 XM_006724290.1 XM_017028903.1 XM_017028904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6994512 c.1137C>A p.L379L Chr.22: 39087123-39087123 on Build GRCh38
3964292 c.1141-4C>T p.? Chr.22: 39087403-39087403 on Build GRCh38
5607797 c.1140G>A p.Q380Q Chr.22: 39087126-39087126 on Build GRCh38
6894498 c.1154G>C p.*385S Chr.22: 39087420-39087420 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs948958583 Chr.22: 39087228-39087228 on Build GRCh38
rs980328918 Chr.22: 39087230-39087230 on Build GRCh38
rs1006810583 Chr.22: 39087368-39087368 on Build GRCh38
rs999787314 Chr.22: 39087573-39087573 on Build GRCh38
rs756809176 Chr.22: 39087154-39087154 on Build GRCh38
rs868100936 Chr.22: 39087383-39087383 on Build GRCh38
rs763399550 Chr.22: 39087392-39087392 on Build GRCh38
rs942643493 Chr.22: 39087547-39087547 on Build GRCh38
rs781085039 Chr.22: 39087357-39087357 on Build GRCh38
rs562177159 Chr.22: 39087369-39087369 on Build GRCh38
rs571551528 Chr.22: 39087327-39087327 on Build GRCh38
rs910853548 Chr.22: 39087522-39087522 on Build GRCh38
rs200261226 Chr.22: 39087172-39087172 on Build GRCh38
rs909990702 Chr.22: 39087352-39087352 on Build GRCh38
rs763628310 Chr.22: 39087149-39087149 on Build GRCh38
rs768088704 Chr.22: 39087415-39087415 on Build GRCh38
rs370524223 Chr.22: 39087448-39087448 on Build GRCh38
rs963180062 Chr.22: 39087458-39087458 on Build GRCh38
rs574850138 Chr.22: 39087161-39087161 on Build GRCh38
rs201660527 Chr.22: 39087160-39087160 on Build GRCh38
rs28425624 Chr.22: 39087188-39087188 on Build GRCh38
rs200761010 Chr.22: 39087354-39087354 on Build GRCh38
rs376834165 Chr.22: 39087150-39087150 on Build GRCh38
rs750133248 Chr.22: 39087441-39087441 on Build GRCh38
rs933669167 Chr.22: 39087336-39087336 on Build GRCh38
rs1044009832 Chr.22: 39087555-39087555 on Build GRCh38
rs562757876 Chr.22: 39087584-39087584 on Build GRCh38
rs933721658 Chr.22: 39087492-39087492 on Build GRCh38
rs916727094 Chr.22: 39087224-39087224 on Build GRCh38
rs781233064 Chr.22: 39087465-39087465 on Build GRCh38
rs568940353 Chr.22: 39087390-39087390 on Build GRCh38
rs17537588 Chr.22: 39087319-39087319 on Build GRCh38
rs917520538 Chr.22: 39087223-39087223 on Build GRCh38
rs76917817 Chr.22: 39087454-39087454 on Build GRCh38
rs753435735 Chr.22: 39087367-39087367 on Build GRCh38
rs970321635 Chr.22: 39087429-39087429 on Build GRCh38
rs527643869 Chr.22: 39087165-39087165 on Build GRCh38
rs544370874 Chr.22: 39087529-39087529 on Build GRCh38
rs578180167 Chr.22: 39087628-39087628 on Build GRCh38
rs368454440 Chr.22: 39087485-39087485 on Build GRCh38
rs55930989 Chr.22: 39087237-39087238 on Build GRCh38
rs201271449 Chr.22: 39087394-39087394 on Build GRCh38
rs374575022 Chr.22: 39087170-39087170 on Build GRCh38
rs756994012 Chr.22: 39087426-39087426 on Build GRCh38
rs560677630 Chr.22: 39087629-39087629 on Build GRCh38
rs1022542421 Chr.22: 39087166-39087166 on Build GRCh38
rs752179957 Chr.22: 39087282-39087282 on Build GRCh38
rs1024261391 Chr.22: 39087609-39087609 on Build GRCh38
rs1014289003 Chr.22: 39087592-39087592 on Build GRCh38
rs748312821 Chr.22: 39087362-39087362 on Build GRCh38
rs1014236678 Chr.22: 39087403-39087403 on Build GRCh38
rs779838723 Chr.22: 39087169-39087169 on Build GRCh38
rs748176876 Chr.22: 39087469-39087469 on Build GRCh38
rs760129468 Chr.22: 39087142-39087142 on Build GRCh38
rs367547786 Chr.22: 39087433-39087433 on Build GRCh38
rs993057196 Chr.22: 39087209-39087209 on Build GRCh38
rs760078242 Chr.22: 39087414-39087414 on Build GRCh38
rs527732972 Chr.22: 39087288-39087288 on Build GRCh38
rs998115571 Chr.22: 39087143-39087143 on Build GRCh38
rs1005288262 Chr.22: 39087301-39087321 on Build GRCh38
rs35205117 Chr.22: 39087342-39087342 on Build GRCh38
rs1047054123 Chr.22: 39087337-39087337 on Build GRCh38
rs1031682744 Chr.22: 39087186-39087186 on Build GRCh38
rs989643351 Chr.22: 39087203-39087203 on Build GRCh38
rs571380577 Chr.22: 39087506-39087506 on Build GRCh38
rs530089610 Chr.22: 39087591-39087591 on Build GRCh38
rs774857271 Chr.22: 39087405-39087405 on Build GRCh38
rs766755018 Chr.22: 39087166-39087166 on Build GRCh38
rs909866862 Chr.22: 39087637-39087637 on Build GRCh38
rs6001422 Chr.22: 39087192-39087192 on Build GRCh38
rs754891412 Chr.22: 39087463-39087463 on Build GRCh38
rs112802713 Chr.22: 39087427-39087427 on Build GRCh38
rs910037066 Chr.22: 39087236-39087236 on Build GRCh38
rs755600900 Chr.22: 39087317-39087317 on Build GRCh38
rs150768394 Chr.22: 39087520-39087520 on Build GRCh38
rs371155567 Chr.22: 39087400-39087400 on Build GRCh38
rs982332473 Chr.22: 39087490-39087490 on Build GRCh38
rs557218170 Chr.22: 39087364-39087364 on Build GRCh38
rs1023769846 Chr.22: 39087459-39087459 on Build GRCh38
rs928244021 Chr.22: 39087500-39087500 on Build GRCh38
rs1000300520 Chr.22: 39087386-39087386 on Build GRCh38
rs768613653 Chr.22: 39087173-39087173 on Build GRCh38
rs769878799 Chr.22: 39087380-39087380 on Build GRCh38
rs1032146494 Chr.22: 39087398-39087398 on Build GRCh38
rs540261431 Chr.22: 39087481-39087481 on Build GRCh38
rs903700386 Chr.22: 39087560-39087560 on Build GRCh38
rs751379555 Chr.22: 39087461-39087461 on Build GRCh38

View Full Product Details