Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39130346-39130885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX7
Entrez Gene ID
23492
Gene Name
chromobox 7
Gene Aliases
-
UniGene
Hs.356416
Gene Chromosome Location
Chr.22: 39130772-39152661 on Build GRCh38
Transcripts
NM_175709.3 XM_005261413.4 XM_006724174.3 XM_006724175.3 XM_006724176.3 XM_006724177.3 XM_006724178.3 XM_011530025.2 XM_017028679.1

SNP Detail

SNP ID Chromosome Location
rs139382 Chr.22: 39130755-39130755 on Build GRCh38
rs921059620 Chr.22: 39130399-39130399 on Build GRCh38
rs948432407 Chr.22: 39130512-39130512 on Build GRCh38
rs145308362 Chr.22: 39130754-39130754 on Build GRCh38
rs887366791 Chr.22: 39130569-39130569 on Build GRCh38
rs564550017 Chr.22: 39130391-39130391 on Build GRCh38
rs746787251 Chr.22: 39130373-39130373 on Build GRCh38
rs942553796 Chr.22: 39130809-39130809 on Build GRCh38
rs1043804 Chr.22: 39130776-39130776 on Build GRCh38
rs918542729 Chr.22: 39130552-39130552 on Build GRCh38
rs992988822 Chr.22: 39130423-39130423 on Build GRCh38
rs955293278 Chr.22: 39130402-39130402 on Build GRCh38
rs61492279 Chr.22: 39130730-39130730 on Build GRCh38
rs990224811 Chr.22: 39130717-39130717 on Build GRCh38
rs868780122 Chr.22: 39130374-39130374 on Build GRCh38
rs1050938803 Chr.22: 39130561-39130561 on Build GRCh38
rs1016914128 Chr.22: 39130600-39130600 on Build GRCh38
rs562316222 Chr.22: 39130577-39130577 on Build GRCh38
rs1003699645 Chr.22: 39130534-39130534 on Build GRCh38
rs1000582245 Chr.22: 39130405-39130405 on Build GRCh38
rs911120600 Chr.22: 39130789-39130789 on Build GRCh38
rs1039139518 Chr.22: 39130613-39130613 on Build GRCh38
rs375166605 Chr.22: 39130422-39130422 on Build GRCh38
rs182591049 Chr.22: 39130663-39130663 on Build GRCh38
rs904568713 Chr.22: 39130635-39130635 on Build GRCh38
rs955949034 Chr.22: 39130705-39130705 on Build GRCh38
rs962790170 Chr.22: 39130607-39130607 on Build GRCh38
rs191180880 Chr.22: 39130530-39130530 on Build GRCh38
rs979862287 Chr.22: 39130497-39130497 on Build GRCh38
rs865871926 Chr.22: 39130514-39130514 on Build GRCh38
rs904638416 Chr.22: 39130834-39130834 on Build GRCh38
rs887367976 Chr.22: 39130764-39130764 on Build GRCh38
rs563963080 Chr.22: 39130776-39130777 on Build GRCh38
rs1018052697 Chr.22: 39130368-39130368 on Build GRCh38
rs1054120087 Chr.22: 39130496-39130496 on Build GRCh38
rs909594314 Chr.22: 39130509-39130509 on Build GRCh38
rs756420830 Chr.22: 39130856-39130856 on Build GRCh38
rs1038398226 Chr.22: 39130811-39130812 on Build GRCh38
rs1012785380 Chr.22: 39130739-39130739 on Build GRCh38
rs969802251 Chr.22: 39130463-39130463 on Build GRCh38
rs371373391 Chr.22: 39130386-39130386 on Build GRCh38
rs761847957 Chr.22: 39130750-39130750 on Build GRCh38
rs776426478 Chr.22: 39130695-39130695 on Build GRCh38
rs529844632 Chr.22: 39130585-39130585 on Build GRCh38
rs575040344 Chr.22: 39130814-39130814 on Build GRCh38
rs768583556 Chr.22: 39130686-39130686 on Build GRCh38
rs58031662 Chr.22: 39130638-39130639 on Build GRCh38
rs1053070947 Chr.22: 39130851-39130851 on Build GRCh38
rs1014179544 Chr.22: 39130857-39130857 on Build GRCh38
rs986647834 Chr.22: 39130681-39130681 on Build GRCh38
rs1031926821 Chr.22: 39130413-39130413 on Build GRCh38
rs529121321 Chr.22: 39130527-39130527 on Build GRCh38
rs569180837 Chr.22: 39130835-39130835 on Build GRCh38
rs963276671 Chr.22: 39130786-39130786 on Build GRCh38
rs933925703 Chr.22: 39130772-39130772 on Build GRCh38
rs1032079775 Chr.22: 39130645-39130645 on Build GRCh38
rs970212048 Chr.22: 39130720-39130720 on Build GRCh38
rs933895010 Chr.22: 39130557-39130557 on Build GRCh38
rs986731200 Chr.22: 39130784-39130784 on Build GRCh38
rs548230868 Chr.22: 39130640-39130640 on Build GRCh38
rs753045994 Chr.22: 39130850-39130850 on Build GRCh38
rs561828876 Chr.22: 39130506-39130506 on Build GRCh38
rs145462297 Chr.22: 39130722-39130722 on Build GRCh38
rs766420753 Chr.22: 39130751-39130751 on Build GRCh38
rs188165902 Chr.22: 39130725-39130725 on Build GRCh38
rs924532050 Chr.22: 39130434-39130434 on Build GRCh38
rs917410661 Chr.22: 39130432-39130432 on Build GRCh38
rs536062092 Chr.22: 39130843-39130843 on Build GRCh38
rs867798201 Chr.22: 39130726-39130726 on Build GRCh38
rs1007109436 Chr.22: 39130608-39130608 on Build GRCh38

View Full Product Details