Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39149543-39150052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX7
Entrez Gene ID
23492
Gene Name
chromobox 7
Gene Aliases
-
UniGene
Hs.356416
Gene Chromosome Location
Chr.22: 39130772-39152661 on Build GRCh38
Transcripts
NM_175709.3 XM_005261413.4 XM_006724174.3 XM_006724175.3 XM_006724176.3 XM_006724177.3 XM_006724178.3 XM_011530025.2 XM_017028679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4669583 c.105G>A p.W35* Chr.22: 39149797-39149797 on Build GRCh38
6347770 c.107C>A p.P36H Chr.22: 39149795-39149795 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1049709874 Chr.22: 39149770-39149770 on Build GRCh38
rs754869621 Chr.22: 39149848-39149848 on Build GRCh38
rs934117625 Chr.22: 39149648-39149648 on Build GRCh38
rs908667307 Chr.22: 39149686-39149686 on Build GRCh38
rs1040785304 Chr.22: 39149738-39149738 on Build GRCh38
rs987403690 Chr.22: 39149971-39149971 on Build GRCh38
rs769035012 Chr.22: 39149857-39149860 on Build GRCh38
rs1044479982 Chr.22: 39150021-39150021 on Build GRCh38
rs147104497 Chr.22: 39149824-39149824 on Build GRCh38
rs763279407 Chr.22: 39149774-39149775 on Build GRCh38
rs960068335 Chr.22: 39149905-39149905 on Build GRCh38
rs776024849 Chr.22: 39149764-39149765 on Build GRCh38
rs758146729 Chr.22: 39149826-39149826 on Build GRCh38
rs781124430 Chr.22: 39149857-39149857 on Build GRCh38
rs201114582 Chr.22: 39149767-39149767 on Build GRCh38
rs768420455 Chr.22: 39149867-39149867 on Build GRCh38
rs961552852 Chr.22: 39149990-39149990 on Build GRCh38
rs766676894 Chr.22: 39149757-39149757 on Build GRCh38
rs1051367806 Chr.22: 39149678-39149678 on Build GRCh38
rs77832508 Chr.22: 39149587-39149587 on Build GRCh38
rs139502152 Chr.22: 39149878-39149878 on Build GRCh38
rs535455978 Chr.22: 39149609-39149609 on Build GRCh38
rs549874511 Chr.22: 39149581-39149581 on Build GRCh38
rs779974021 Chr.22: 39149838-39149838 on Build GRCh38
rs746894182 Chr.22: 39149741-39149741 on Build GRCh38
rs1012860886 Chr.22: 39149911-39149911 on Build GRCh38
rs931532972 Chr.22: 39149671-39149671 on Build GRCh38
rs907060507 Chr.22: 39149814-39149814 on Build GRCh38
rs533859500 Chr.22: 39149627-39149627 on Build GRCh38
rs566744166 Chr.22: 39149617-39149617 on Build GRCh38
rs191985014 Chr.22: 39149574-39149574 on Build GRCh38
rs369984674 Chr.22: 39149776-39149776 on Build GRCh38
rs1035719521 Chr.22: 39149898-39149898 on Build GRCh38
rs780303710 Chr.22: 39149704-39149708 on Build GRCh38
rs147724211 Chr.22: 39149922-39149922 on Build GRCh38
rs1015977479 Chr.22: 39149930-39149930 on Build GRCh38
rs770890760 Chr.22: 39150006-39150006 on Build GRCh38
rs769824191 Chr.22: 39149873-39149873 on Build GRCh38
rs999374836 Chr.22: 39149881-39149881 on Build GRCh38
rs552064710 Chr.22: 39149919-39149919 on Build GRCh38
rs761477607 Chr.22: 39149963-39149963 on Build GRCh38
rs933038450 Chr.22: 39149965-39149965 on Build GRCh38
rs991726395 Chr.22: 39149618-39149618 on Build GRCh38
rs774152605 Chr.22: 39150011-39150011 on Build GRCh38
rs531476326 Chr.22: 39149974-39149974 on Build GRCh38
rs556289710 Chr.22: 39149915-39149915 on Build GRCh38
rs879456900 Chr.22: 39149900-39149900 on Build GRCh38
rs747692280 Chr.22: 39149746-39149746 on Build GRCh38
rs748038323 Chr.22: 39149866-39149866 on Build GRCh38
rs763192984 Chr.22: 39149753-39149753 on Build GRCh38
rs976520522 Chr.22: 39150008-39150008 on Build GRCh38
rs893133439 Chr.22: 39149775-39149775 on Build GRCh38
rs562569207 Chr.22: 39149748-39149748 on Build GRCh38
rs916159809 Chr.22: 39149652-39149652 on Build GRCh38
rs984363002 Chr.22: 39149945-39149945 on Build GRCh38
rs751966582 Chr.22: 39149762-39149762 on Build GRCh38
rs112314602 Chr.22: 39149709-39149709 on Build GRCh38
rs569975189 Chr.22: 39149753-39149754 on Build GRCh38
rs924794665 Chr.22: 39149977-39149977 on Build GRCh38
rs202019493 Chr.22: 39149783-39149783 on Build GRCh38
rs562564843 Chr.22: 39150004-39150004 on Build GRCh38
rs776544649 Chr.22: 39149742-39149742 on Build GRCh38
rs746862021 Chr.22: 39149846-39149846 on Build GRCh38
rs113053332 Chr.22: 39150025-39150025 on Build GRCh38
rs902315448 Chr.22: 39149869-39149869 on Build GRCh38
rs770387652 Chr.22: 39149754-39149757 on Build GRCh38
rs1026131150 Chr.22: 39149888-39149888 on Build GRCh38
rs780770897 Chr.22: 39149730-39149730 on Build GRCh38
rs553778046 Chr.22: 39149612-39149612 on Build GRCh38
rs921435381 Chr.22: 39149643-39149643 on Build GRCh38
rs117568079 Chr.22: 39149879-39149879 on Build GRCh38
rs773354807 Chr.22: 39149750-39149750 on Build GRCh38
rs754438293 Chr.22: 39149685-39149685 on Build GRCh38
rs749070997 Chr.22: 39149961-39149961 on Build GRCh38
rs200181829 Chr.22: 39149771-39149771 on Build GRCh38
rs377014158 Chr.22: 39149766-39149766 on Build GRCh38
rs908865530 Chr.22: 39149608-39149608 on Build GRCh38
rs541829398 Chr.22: 39150009-39150009 on Build GRCh38
rs773302817 Chr.22: 39149876-39149876 on Build GRCh38
rs777665687 Chr.22: 39149932-39149932 on Build GRCh38

View Full Product Details