Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39521158-39521657 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF4
Entrez Gene ID
468
Gene Name
activating transcription factor 4
Gene Aliases
CREB-2, CREB2, TAXREB67, TXREB
UniGene
Hs.496487
Gene Chromosome Location
Chr.22: 39519709-39522686 on Build GRCh38
Transcripts
NM_001675.4 NM_182810.2 XM_017028807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3152087 c.53C>T p.S18F Chr.22: 39521498-39521498 on Build GRCh38
3708264 c.83A>T p.E28V Chr.22: 39521528-39521528 on Build GRCh38
1161427 c.125A>G p.K42R Chr.22: 39521570-39521570 on Build GRCh38
4821278 c.99C>G p.L33L Chr.22: 39521544-39521544 on Build GRCh38
6390898 c.35T>A p.L12* Chr.22: 39521480-39521480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151149967 Chr.22: 39521345-39521345 on Build GRCh38
rs536034941 Chr.22: 39521622-39521622 on Build GRCh38
rs747272986 Chr.22: 39521585-39521585 on Build GRCh38
rs370467448 Chr.22: 39521542-39521542 on Build GRCh38
rs910997963 Chr.22: 39521255-39521255 on Build GRCh38
rs1001545052 Chr.22: 39521413-39521413 on Build GRCh38
rs184569610 Chr.22: 39521247-39521247 on Build GRCh38
rs779393729 Chr.22: 39521423-39521423 on Build GRCh38
rs776908104 Chr.22: 39521587-39521587 on Build GRCh38
rs367738767 Chr.22: 39521328-39521328 on Build GRCh38
rs902767391 Chr.22: 39521296-39521296 on Build GRCh38
rs371466949 Chr.22: 39521402-39521402 on Build GRCh38
rs780640405 Chr.22: 39521548-39521548 on Build GRCh38
rs1032908283 Chr.22: 39521312-39521312 on Build GRCh38
rs972646909 Chr.22: 39521232-39521232 on Build GRCh38
rs918503685 Chr.22: 39521234-39521234 on Build GRCh38
rs895305048 Chr.22: 39521420-39521420 on Build GRCh38
rs139077616 Chr.22: 39521369-39521369 on Build GRCh38
rs530230780 Chr.22: 39521432-39521432 on Build GRCh38
rs1009820191 Chr.22: 39521287-39521287 on Build GRCh38
rs781630310 Chr.22: 39521508-39521508 on Build GRCh38
rs201201452 Chr.22: 39521446-39521446 on Build GRCh38
rs374660871 Chr.22: 39521421-39521421 on Build GRCh38
rs746700301 Chr.22: 39521428-39521428 on Build GRCh38
rs942515805 Chr.22: 39521367-39521367 on Build GRCh38
rs868642940 Chr.22: 39521239-39521239 on Build GRCh38
rs73885241 Chr.22: 39521295-39521295 on Build GRCh38
rs764339006 Chr.22: 39521463-39521463 on Build GRCh38
rs193166285 Chr.22: 39521182-39521182 on Build GRCh38
rs752369664 Chr.22: 39521552-39521552 on Build GRCh38
rs759741126 Chr.22: 39521305-39521305 on Build GRCh38
rs189837769 Chr.22: 39521259-39521259 on Build GRCh38
rs760943141 Chr.22: 39521519-39521519 on Build GRCh38
rs970788338 Chr.22: 39521424-39521424 on Build GRCh38
rs552275262 Chr.22: 39521490-39521490 on Build GRCh38
rs749101924 Chr.22: 39521570-39521570 on Build GRCh38
rs972720499 Chr.22: 39521329-39521329 on Build GRCh38
rs770742676 Chr.22: 39521574-39521574 on Build GRCh38
rs372377952 Chr.22: 39521306-39521306 on Build GRCh38
rs140650335 Chr.22: 39521278-39521278 on Build GRCh38
rs372523612 Chr.22: 39521540-39521540 on Build GRCh38
rs747830080 Chr.22: 39521601-39521602 on Build GRCh38
rs544809451 Chr.22: 39521362-39521362 on Build GRCh38
rs141727778 Chr.22: 39521621-39521621 on Build GRCh38
rs990861611 Chr.22: 39521188-39521188 on Build GRCh38
rs149908720 Chr.22: 39521452-39521452 on Build GRCh38
rs768241118 Chr.22: 39521429-39521429 on Build GRCh38
rs778473174 Chr.22: 39521561-39521561 on Build GRCh38
rs931196007 Chr.22: 39521242-39521242 on Build GRCh38
rs762144715 Chr.22: 39521591-39521591 on Build GRCh38
rs753654252 Chr.22: 39521225-39521225 on Build GRCh38
rs754403045 Chr.22: 39521467-39521467 on Build GRCh38
rs768694488 Chr.22: 39521586-39521586 on Build GRCh38
rs765613696 Chr.22: 39521597-39521597 on Build GRCh38
rs756538551 Chr.22: 39521403-39521403 on Build GRCh38
rs111935226 Chr.22: 39521353-39521353 on Build GRCh38
rs149326054 Chr.22: 39521579-39521579 on Build GRCh38
rs751836565 Chr.22: 39521395-39521395 on Build GRCh38
rs773738739 Chr.22: 39521598-39521598 on Build GRCh38
rs780881509 Chr.22: 39521482-39521482 on Build GRCh38
rs768004395 Chr.22: 39521401-39521401 on Build GRCh38
rs775898432 Chr.22: 39521517-39521517 on Build GRCh38
rs148038848 Chr.22: 39521605-39521605 on Build GRCh38
rs759839365 Chr.22: 39521448-39521448 on Build GRCh38
rs754480311 Chr.22: 39521481-39521481 on Build GRCh38
rs181328727 Chr.22: 39521349-39521349 on Build GRCh38
rs1054005603 Chr.22: 39521417-39521417 on Build GRCh38
rs755663587 Chr.22: 39521555-39521555 on Build GRCh38
rs779630800 Chr.22: 39521471-39521471 on Build GRCh38
rs4894 Chr.22: 39521510-39521510 on Build GRCh38
rs779038575 Chr.22: 39521577-39521577 on Build GRCh38
rs762759423 Chr.22: 39521440-39521440 on Build GRCh38
rs74751854 Chr.22: 39521314-39521314 on Build GRCh38
rs1040741627 Chr.22: 39521280-39521280 on Build GRCh38
rs146354334 Chr.22: 39521449-39521449 on Build GRCh38
rs371528882 Chr.22: 39521336-39521336 on Build GRCh38
rs11556097 Chr.22: 39521431-39521431 on Build GRCh38
rs34854854 Chr.22: 39521498-39521498 on Build GRCh38
rs900855773 Chr.22: 39521284-39521284 on Build GRCh38
rs751006399 Chr.22: 39521472-39521472 on Build GRCh38
rs994266051 Chr.22: 39521612-39521612 on Build GRCh38
rs925614655 Chr.22: 39521573-39521573 on Build GRCh38
rs764544798 Chr.22: 39521521-39521521 on Build GRCh38
rs546014190 Chr.22: 39521496-39521496 on Build GRCh38
rs199649119 Chr.22: 39521511-39521511 on Build GRCh38
rs945751296 Chr.22: 39521392-39521392 on Build GRCh38
rs369962392 Chr.22: 39521437-39521437 on Build GRCh38
rs749260078 Chr.22: 39521500-39521500 on Build GRCh38
rs375843215 Chr.22: 39521556-39521556 on Build GRCh38
rs767028222 Chr.22: 39521618-39521618 on Build GRCh38
rs754096991 Chr.22: 39521525-39521525 on Build GRCh38
rs566834234 Chr.22: 39521192-39521192 on Build GRCh38
rs527712972 Chr.22: 39521480-39521480 on Build GRCh38
rs942463488 Chr.22: 39521262-39521262 on Build GRCh38
rs755200555 Chr.22: 39521400-39521400 on Build GRCh38
rs778398197 Chr.22: 39521419-39521419 on Build GRCh38
rs757658895 Chr.22: 39521468-39521468 on Build GRCh38
rs111719524 Chr.22: 39521582-39521582 on Build GRCh38
rs772253684 Chr.22: 39521514-39521514 on Build GRCh38
rs1001072408 Chr.22: 39521307-39521307 on Build GRCh38
rs563469938 Chr.22: 39521319-39521319 on Build GRCh38
rs200926784 Chr.22: 39521538-39521538 on Build GRCh38
rs762388843 Chr.22: 39521529-39521529 on Build GRCh38
rs577428565 Chr.22: 39521358-39521358 on Build GRCh38
rs769662388 Chr.22: 39521493-39521493 on Build GRCh38
rs948230107 Chr.22: 39521584-39521584 on Build GRCh38
rs144184225 Chr.22: 39521320-39521320 on Build GRCh38
rs769564982 Chr.22: 39521433-39521433 on Build GRCh38
rs527679261 Chr.22: 39521200-39521200 on Build GRCh38
rs376351770 Chr.22: 39521441-39521441 on Build GRCh38

View Full Product Details