Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39521690-39522185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF4
Entrez Gene ID
468
Gene Name
activating transcription factor 4
Gene Aliases
CREB-2, CREB2, TAXREB67, TXREB
UniGene
Hs.496487
Gene Chromosome Location
Chr.22: 39519709-39522686 on Build GRCh38
Transcripts
NM_001675.4 NM_182810.2 XM_017028807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4822584 c.430C>G p.L144V Chr.22: 39521976-39521976 on Build GRCh38
297659 c.441T>C p.S147S Chr.22: 39521987-39521987 on Build GRCh38
224099 c.302G>A p.G101D Chr.22: 39521848-39521848 on Build GRCh38
4977708 c.482A>T p.Q161L Chr.22: 39522028-39522028 on Build GRCh38
331238 c.631G>T p.E211* Chr.22: 39522177-39522177 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536072398 Chr.22: 39522087-39522087 on Build GRCh38
rs757892971 Chr.22: 39522036-39522036 on Build GRCh38
rs775770534 Chr.22: 39522138-39522138 on Build GRCh38
rs149857296 Chr.22: 39521840-39521840 on Build GRCh38
rs369567094 Chr.22: 39521752-39521752 on Build GRCh38
rs750690606 Chr.22: 39521965-39521965 on Build GRCh38
rs868548256 Chr.22: 39521910-39521910 on Build GRCh38
rs141575018 Chr.22: 39521782-39521782 on Build GRCh38
rs761617273 Chr.22: 39522021-39522021 on Build GRCh38
rs747543814 Chr.22: 39521946-39521946 on Build GRCh38
rs760328559 Chr.22: 39521915-39521915 on Build GRCh38
rs369760669 Chr.22: 39521850-39521850 on Build GRCh38
rs765835344 Chr.22: 39522097-39522097 on Build GRCh38
rs867065698 Chr.22: 39521894-39521894 on Build GRCh38
rs771524268 Chr.22: 39521811-39521812 on Build GRCh38
rs765557801 Chr.22: 39521927-39521927 on Build GRCh38
rs140428747 Chr.22: 39521950-39521950 on Build GRCh38
rs141643322 Chr.22: 39522111-39522111 on Build GRCh38
rs114210419 Chr.22: 39521748-39521748 on Build GRCh38
rs1005988092 Chr.22: 39521879-39521879 on Build GRCh38
rs181790280 Chr.22: 39521922-39521922 on Build GRCh38
rs575170457 Chr.22: 39522077-39522077 on Build GRCh38
rs771259116 Chr.22: 39521743-39521743 on Build GRCh38
rs11556099 Chr.22: 39521856-39521856 on Build GRCh38
rs138640471 Chr.22: 39522066-39522066 on Build GRCh38
rs762005484 Chr.22: 39521962-39521962 on Build GRCh38
rs529375059 Chr.22: 39521926-39521926 on Build GRCh38
rs758837809 Chr.22: 39521942-39521942 on Build GRCh38
rs375266335 Chr.22: 39521750-39521750 on Build GRCh38
rs981100683 Chr.22: 39522082-39522082 on Build GRCh38
rs760553685 Chr.22: 39521919-39521919 on Build GRCh38
rs146204295 Chr.22: 39522125-39522125 on Build GRCh38
rs759246444 Chr.22: 39521842-39521842 on Build GRCh38
rs772801965 Chr.22: 39521889-39521889 on Build GRCh38
rs374484492 Chr.22: 39522129-39522129 on Build GRCh38
rs746637544 Chr.22: 39522038-39522038 on Build GRCh38
rs748131287 Chr.22: 39522045-39522045 on Build GRCh38
rs148568771 Chr.22: 39521859-39521859 on Build GRCh38
rs781046850 Chr.22: 39522106-39522106 on Build GRCh38
rs142911753 Chr.22: 39521875-39521875 on Build GRCh38
rs149522605 Chr.22: 39522017-39522017 on Build GRCh38
rs756172316 Chr.22: 39521775-39521775 on Build GRCh38
rs771555732 Chr.22: 39522009-39522009 on Build GRCh38
rs369838694 Chr.22: 39521822-39521822 on Build GRCh38
rs377487629 Chr.22: 39521985-39521985 on Build GRCh38
rs971085725 Chr.22: 39522072-39522072 on Build GRCh38
rs752888565 Chr.22: 39521771-39521771 on Build GRCh38
rs772487305 Chr.22: 39522132-39522132 on Build GRCh38
rs1012671336 Chr.22: 39522069-39522069 on Build GRCh38
rs754568653 Chr.22: 39522102-39522102 on Build GRCh38
rs375489476 Chr.22: 39521761-39521761 on Build GRCh38
rs766764304 Chr.22: 39522027-39522027 on Build GRCh38
rs768004321 Chr.22: 39522059-39522059 on Build GRCh38
rs572204009 Chr.22: 39522041-39522041 on Build GRCh38
rs774758731 Chr.22: 39521744-39521744 on Build GRCh38
rs371009074 Chr.22: 39522051-39522051 on Build GRCh38
rs767949359 Chr.22: 39522029-39522029 on Build GRCh38
rs575135566 Chr.22: 39521828-39521828 on Build GRCh38
rs544103363 Chr.22: 39522000-39522000 on Build GRCh38
rs770325317 Chr.22: 39521948-39521948 on Build GRCh38
rs1032859544 Chr.22: 39522061-39522061 on Build GRCh38
rs138075537 Chr.22: 39521981-39521981 on Build GRCh38
rs115111682 Chr.22: 39521786-39521786 on Build GRCh38
rs146469032 Chr.22: 39521944-39521944 on Build GRCh38
rs767181464 Chr.22: 39521912-39521912 on Build GRCh38
rs758265085 Chr.22: 39521716-39521716 on Build GRCh38
rs753266567 Chr.22: 39522030-39522030 on Build GRCh38
rs773862346 Chr.22: 39521906-39521906 on Build GRCh38
rs186327285 Chr.22: 39521924-39521924 on Build GRCh38
rs566582294 Chr.22: 39521997-39521997 on Build GRCh38
rs142422504 Chr.22: 39521939-39521939 on Build GRCh38
rs188265360 Chr.22: 39521720-39521720 on Build GRCh38
rs747519249 Chr.22: 39521764-39521764 on Build GRCh38
rs767548425 Chr.22: 39521848-39521848 on Build GRCh38
rs748952205 Chr.22: 39521891-39521891 on Build GRCh38
rs775433091 Chr.22: 39521918-39521918 on Build GRCh38
rs147877175 Chr.22: 39521947-39521947 on Build GRCh38
rs761566773 Chr.22: 39521749-39521749 on Build GRCh38
rs773109981 Chr.22: 39522024-39522024 on Build GRCh38
rs371958399 Chr.22: 39521736-39521736 on Build GRCh38
rs754079438 Chr.22: 39521854-39521854 on Build GRCh38
rs747948440 Chr.22: 39522108-39522108 on Build GRCh38
rs761467344 Chr.22: 39521801-39521801 on Build GRCh38
rs746089688 Chr.22: 39522131-39522131 on Build GRCh38
rs888535148 Chr.22: 39521864-39521864 on Build GRCh38
rs367700144 Chr.22: 39522151-39522151 on Build GRCh38
rs777156994 Chr.22: 39521880-39521882 on Build GRCh38
rs749969874 Chr.22: 39522034-39522034 on Build GRCh38
rs570395027 Chr.22: 39522031-39522031 on Build GRCh38
rs773985132 Chr.22: 39522057-39522059 on Build GRCh38
rs61737791 Chr.22: 39522146-39522146 on Build GRCh38
rs746282616 Chr.22: 39521923-39521923 on Build GRCh38
rs775860856 Chr.22: 39521799-39521799 on Build GRCh38
rs765888817 Chr.22: 39522152-39522152 on Build GRCh38
rs899844959 Chr.22: 39521986-39521987 on Build GRCh38
rs374864123 Chr.22: 39521712-39521712 on Build GRCh38
rs760997915 Chr.22: 39522142-39522142 on Build GRCh38
rs748330323 Chr.22: 39521725-39521725 on Build GRCh38
rs778487966 Chr.22: 39522033-39522033 on Build GRCh38
rs769553378 Chr.22: 39522050-39522050 on Build GRCh38
rs763038389 Chr.22: 39521967-39521967 on Build GRCh38
rs764169884 Chr.22: 39521921-39521921 on Build GRCh38
rs766100228 Chr.22: 39521832-39521832 on Build GRCh38
rs751006341 Chr.22: 39522155-39522155 on Build GRCh38
rs542420026 Chr.22: 39521852-39521852 on Build GRCh38
rs368914495 Chr.22: 39522109-39522109 on Build GRCh38
rs145847419 Chr.22: 39521858-39521858 on Build GRCh38
rs755054646 Chr.22: 39521721-39521721 on Build GRCh38
rs773709050 Chr.22: 39521742-39521742 on Build GRCh38
rs11556101 Chr.22: 39522014-39522014 on Build GRCh38
rs770192882 Chr.22: 39521945-39521946 on Build GRCh38
rs780198861 Chr.22: 39521719-39521719 on Build GRCh38
rs373032649 Chr.22: 39521759-39521759 on Build GRCh38
rs765201376 Chr.22: 39521964-39521964 on Build GRCh38
rs770681729 Chr.22: 39521905-39521905 on Build GRCh38
rs760045225 Chr.22: 39521747-39521747 on Build GRCh38
rs746376611 Chr.22: 39521796-39521796 on Build GRCh38
rs763178109 Chr.22: 39522143-39522143 on Build GRCh38
rs780918174 Chr.22: 39522044-39522044 on Build GRCh38
rs201312683 Chr.22: 39522052-39522052 on Build GRCh38
rs769461525 Chr.22: 39521888-39521888 on Build GRCh38
rs754482333 Chr.22: 39522158-39522158 on Build GRCh38
rs547944682 Chr.22: 39521972-39521972 on Build GRCh38
rs916847510 Chr.22: 39521807-39521807 on Build GRCh38
rs760612945 Chr.22: 39521952-39521952 on Build GRCh38
rs775634104 Chr.22: 39522020-39522022 on Build GRCh38
rs751236282 Chr.22: 39522098-39522098 on Build GRCh38
rs763082397 Chr.22: 39522026-39522026 on Build GRCh38
rs1022687030 Chr.22: 39522071-39522071 on Build GRCh38
rs867462896 Chr.22: 39521895-39521895 on Build GRCh38
rs775068459 Chr.22: 39521880-39521880 on Build GRCh38
rs759424161 Chr.22: 39521908-39521908 on Build GRCh38
rs753112995 Chr.22: 39521996-39521996 on Build GRCh38
rs779679328 Chr.22: 39522013-39522013 on Build GRCh38
rs777275974 Chr.22: 39521759-39521760 on Build GRCh38
rs2228181 Chr.22: 39522147-39522147 on Build GRCh38
rs774745103 Chr.22: 39522054-39522054 on Build GRCh38
rs767122150 Chr.22: 39522159-39522159 on Build GRCh38
rs373772666 Chr.22: 39521933-39521933 on Build GRCh38
rs368098688 Chr.22: 39522149-39522149 on Build GRCh38
rs201284334 Chr.22: 39522090-39522090 on Build GRCh38
rs150385569 Chr.22: 39521955-39521955 on Build GRCh38
rs769936180 Chr.22: 39521726-39521726 on Build GRCh38
rs1029503588 Chr.22: 39521902-39521902 on Build GRCh38
rs146185967 Chr.22: 39521785-39521785 on Build GRCh38
rs777609927 Chr.22: 39522110-39522110 on Build GRCh38
rs775205463 Chr.22: 39521951-39521951 on Build GRCh38
rs552162021 Chr.22: 39522028-39522028 on Build GRCh38
rs202175644 Chr.22: 39522022-39522022 on Build GRCh38
rs772567775 Chr.22: 39521797-39521797 on Build GRCh38
rs771103079 Chr.22: 39522053-39522053 on Build GRCh38
rs951648088 Chr.22: 39522118-39522118 on Build GRCh38
rs781282473 Chr.22: 39521722-39521722 on Build GRCh38
rs750757685 Chr.22: 39521992-39521992 on Build GRCh38
rs529870594 Chr.22: 39521960-39521960 on Build GRCh38
rs747488604 Chr.22: 39521883-39521883 on Build GRCh38
rs781776700 Chr.22: 39521987-39521987 on Build GRCh38
rs144769713 Chr.22: 39522058-39522058 on Build GRCh38

View Full Product Details